Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100630442-100630954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG7
Entrez Gene ID
84873
Gene Name
adhesion G protein-coupled receptor G7
Gene Aliases
GPR128
UniGene
Hs.334511
Gene Chromosome Location
Chr.3: 100609589-100696174 on Build GRCh38
Transcripts
NM_032787.2 XM_011513245.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5698421 c.233A>G p.N78S Chr.3: 100630708-100630708 on Build GRCh38
2914807 c.233A>T p.N78I Chr.3: 100630708-100630708 on Build GRCh38
5863087 c.295G>A p.G99R Chr.3: 100630770-100630770 on Build GRCh38
3408064 c.254A>G p.Y85C Chr.3: 100630729-100630729 on Build GRCh38
1035629 c.235T>G p.F79V Chr.3: 100630710-100630710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023856529 Chr.3: 100630473-100630473 on Build GRCh38
rs893948240 Chr.3: 100630787-100630787 on Build GRCh38
rs971417209 Chr.3: 100630808-100630808 on Build GRCh38
rs771139682 Chr.3: 100630889-100630889 on Build GRCh38
rs768263844 Chr.3: 100630707-100630707 on Build GRCh38
rs750134558 Chr.3: 100630685-100630685 on Build GRCh38
rs146877111 Chr.3: 100630731-100630731 on Build GRCh38
rs370982120 Chr.3: 100630534-100630534 on Build GRCh38
rs753192855 Chr.3: 100630726-100630726 on Build GRCh38
rs887574160 Chr.3: 100630754-100630754 on Build GRCh38
rs988641583 Chr.3: 100630582-100630582 on Build GRCh38
rs960752880 Chr.3: 100630559-100630559 on Build GRCh38
rs771263196 Chr.3: 100630834-100630834 on Build GRCh38
rs1023872406 Chr.3: 100630798-100630798 on Build GRCh38
rs759660175 Chr.3: 100630749-100630749 on Build GRCh38
rs764077151 Chr.3: 100630662-100630662 on Build GRCh38
rs982545414 Chr.3: 100630505-100630505 on Build GRCh38
rs546735067 Chr.3: 100630669-100630669 on Build GRCh38
rs778139978 Chr.3: 100630827-100630827 on Build GRCh38
rs902929678 Chr.3: 100630761-100630761 on Build GRCh38
rs761829747 Chr.3: 100630773-100630773 on Build GRCh38
rs748868553 Chr.3: 100630704-100630704 on Build GRCh38
rs747431853 Chr.3: 100630829-100630829 on Build GRCh38
rs374423340 Chr.3: 100630717-100630717 on Build GRCh38
rs1002426472 Chr.3: 100630910-100630910 on Build GRCh38
rs568584391 Chr.3: 100630679-100630679 on Build GRCh38
rs779782148 Chr.3: 100630698-100630698 on Build GRCh38
rs529603725 Chr.3: 100630491-100630491 on Build GRCh38
rs757041751 Chr.3: 100630675-100630675 on Build GRCh38
rs41272969 Chr.3: 100630530-100630530 on Build GRCh38
rs753059092 Chr.3: 100630488-100630488 on Build GRCh38
rs755147264 Chr.3: 100630703-100630703 on Build GRCh38
rs779330429 Chr.3: 100630709-100630709 on Build GRCh38
rs755837949 Chr.3: 100630686-100630686 on Build GRCh38
rs114127284 Chr.3: 100630766-100630766 on Build GRCh38
rs914014963 Chr.3: 100630588-100630588 on Build GRCh38
rs1040040660 Chr.3: 100630694-100630694 on Build GRCh38
rs747555940 Chr.3: 100630800-100630800 on Build GRCh38
rs187509287 Chr.3: 100630793-100630793 on Build GRCh38
rs1012420970 Chr.3: 100630470-100630470 on Build GRCh38
rs969645395 Chr.3: 100630510-100630510 on Build GRCh38
rs755963422 Chr.3: 100630785-100630785 on Build GRCh38
rs759784773 Chr.3: 100630836-100630836 on Build GRCh38
rs370823404 Chr.3: 100630501-100630501 on Build GRCh38
rs907942694 Chr.3: 100630507-100630507 on Build GRCh38
rs371702261 Chr.3: 100630824-100630824 on Build GRCh38
rs570007876 Chr.3: 100630835-100630835 on Build GRCh38
rs775650184 Chr.3: 100630843-100630843 on Build GRCh38
rs911821307 Chr.3: 100630688-100630688 on Build GRCh38
rs750234175 Chr.3: 100630777-100630777 on Build GRCh38
rs1034985176 Chr.3: 100630546-100630546 on Build GRCh38
rs910439653 Chr.3: 100630626-100630626 on Build GRCh38
rs552459084 Chr.3: 100630621-100630621 on Build GRCh38
rs183518500 Chr.3: 100630592-100630592 on Build GRCh38
rs748723869 Chr.3: 100630714-100630714 on Build GRCh38
rs570524027 Chr.3: 100630647-100630647 on Build GRCh38
rs199532239 Chr.3: 100630841-100630841 on Build GRCh38
rs201198203 Chr.3: 100630851-100630851 on Build GRCh38
rs763068527 Chr.3: 100630775-100630775 on Build GRCh38
rs767903502 Chr.3: 100630725-100630725 on Build GRCh38
rs541471682 Chr.3: 100630496-100630496 on Build GRCh38
rs528290242 Chr.3: 100630663-100630663 on Build GRCh38
rs778508765 Chr.3: 100630796-100630796 on Build GRCh38
rs753512578 Chr.3: 100630792-100630792 on Build GRCh38
rs367805466 Chr.3: 100630744-100630744 on Build GRCh38
rs1025782623 Chr.3: 100630487-100630487 on Build GRCh38
rs767554641 Chr.3: 100630854-100630854 on Build GRCh38
rs771209708 Chr.3: 100630729-100630729 on Build GRCh38
rs199749116 Chr.3: 100630721-100630721 on Build GRCh38
rs779906546 Chr.3: 100630790-100630790 on Build GRCh38
rs765268571 Chr.3: 100630760-100630760 on Build GRCh38
rs376681454 Chr.3: 100630725-100630725 on Build GRCh38
rs962474773 Chr.3: 100630508-100630508 on Build GRCh38

View Full Product Details