Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108161742-108162251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT57
Entrez Gene ID
55081
Gene Name
intraflagellar transport 57
Gene Aliases
ESRRBL1, HIPPI, MHS4R2
UniGene
Hs.412196
Gene Chromosome Location
Chr.3: 108160812-108222570 on Build GRCh38
Transcripts
NM_018010.3

SNP Detail

SNP ID Chromosome Location
rs924654180 Chr.3: 108162004-108162004 on Build GRCh38
rs979298909 Chr.3: 108161853-108161853 on Build GRCh38
rs879214309 Chr.3: 108161973-108161974 on Build GRCh38
rs932364212 Chr.3: 108161907-108161907 on Build GRCh38
rs2035 Chr.3: 108162072-108162072 on Build GRCh38
rs139677905 Chr.3: 108162174-108162174 on Build GRCh38
rs578026911 Chr.3: 108162203-108162203 on Build GRCh38
rs1027414003 Chr.3: 108161908-108161908 on Build GRCh38
rs968952396 Chr.3: 108161852-108161852 on Build GRCh38
rs905906279 Chr.3: 108161786-108161786 on Build GRCh38
rs960007862 Chr.3: 108161824-108161824 on Build GRCh38
rs989089614 Chr.3: 108161831-108161831 on Build GRCh38
rs1035761874 Chr.3: 108162011-108162011 on Build GRCh38
rs1000382043 Chr.3: 108162047-108162047 on Build GRCh38
rs904840095 Chr.3: 108161834-108161834 on Build GRCh38
rs953554396 Chr.3: 108162096-108162096 on Build GRCh38
rs147812604 Chr.3: 108161841-108161841 on Build GRCh38
rs77845940 Chr.3: 108161981-108161981 on Build GRCh38
rs993369194 Chr.3: 108162185-108162185 on Build GRCh38
rs758847908 Chr.3: 108162013-108162013 on Build GRCh38
rs905962581 Chr.3: 108161984-108161984 on Build GRCh38
rs951917016 Chr.3: 108161926-108161926 on Build GRCh38
rs931165416 Chr.3: 108161966-108161966 on Build GRCh38
rs985205159 Chr.3: 108162129-108162129 on Build GRCh38
rs763879876 Chr.3: 108162166-108162166 on Build GRCh38
rs572653475 Chr.3: 108162019-108162019 on Build GRCh38
rs1020538975 Chr.3: 108161843-108161843 on Build GRCh38
rs925063304 Chr.3: 108161867-108161867 on Build GRCh38
rs948888007 Chr.3: 108162029-108162029 on Build GRCh38
rs1020550309 Chr.3: 108162032-108162032 on Build GRCh38
rs535996029 Chr.3: 108161894-108161894 on Build GRCh38
rs376511377 Chr.3: 108161818-108161818 on Build GRCh38
rs558785047 Chr.3: 108162014-108162014 on Build GRCh38
rs951864506 Chr.3: 108162221-108162221 on Build GRCh38
rs752833038 Chr.3: 108161821-108161821 on Build GRCh38
rs1003647972 Chr.3: 108161842-108161842 on Build GRCh38
rs962244113 Chr.3: 108161856-108161856 on Build GRCh38
rs985110727 Chr.3: 108161925-108161925 on Build GRCh38
rs146708114 Chr.3: 108161974-108161974 on Build GRCh38
rs569960683 Chr.3: 108161933-108161933 on Build GRCh38
rs897499975 Chr.3: 108162146-108162146 on Build GRCh38
rs574952706 Chr.3: 108162012-108162012 on Build GRCh38
rs57535746 Chr.3: 108162130-108162130 on Build GRCh38
rs1027569517 Chr.3: 108162217-108162217 on Build GRCh38
rs965055878 Chr.3: 108162169-108162169 on Build GRCh38
rs897062550 Chr.3: 108161954-108161954 on Build GRCh38
rs1048670847 Chr.3: 108161969-108161969 on Build GRCh38
rs534988981 Chr.3: 108162042-108162042 on Build GRCh38
rs776109449 Chr.3: 108161932-108161932 on Build GRCh38
rs549939460 Chr.3: 108161927-108161927 on Build GRCh38
rs538936021 Chr.3: 108162008-108162008 on Build GRCh38
rs184964779 Chr.3: 108161802-108161802 on Build GRCh38

View Full Product Details