Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108162150-108162660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT57
Entrez Gene ID
55081
Gene Name
intraflagellar transport 57
Gene Aliases
ESRRBL1, HIPPI, MHS4R2
UniGene
Hs.412196
Gene Chromosome Location
Chr.3: 108160812-108222570 on Build GRCh38
Transcripts
NM_018010.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3800854 c.1280G>C p.G427A Chr.3: 108162487-108162487 on Build GRCh38
479191 c.1177A>G p.R393G Chr.3: 108162590-108162590 on Build GRCh38
4112044 c.1198A>G p.T400A Chr.3: 108162569-108162569 on Build GRCh38
6203077 c.1128T>A p.I376I Chr.3: 108162639-108162639 on Build GRCh38
4112045 c.1148T>C p.L383P Chr.3: 108162619-108162619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761041032 Chr.3: 108162572-108162572 on Build GRCh38
rs111640670 Chr.3: 108162343-108162343 on Build GRCh38
rs369854616 Chr.3: 108162449-108162449 on Build GRCh38
rs762267856 Chr.3: 108162443-108162443 on Build GRCh38
rs35713185 Chr.3: 108162535-108162535 on Build GRCh38
rs1057274113 Chr.3: 108162298-108162298 on Build GRCh38
rs928820204 Chr.3: 108162410-108162410 on Build GRCh38
rs764838033 Chr.3: 108162461-108162461 on Build GRCh38
rs918520704 Chr.3: 108162242-108162242 on Build GRCh38
rs749424747 Chr.3: 108162434-108162434 on Build GRCh38
rs1048245380 Chr.3: 108162284-108162284 on Build GRCh38
rs547351126 Chr.3: 108162447-108162447 on Build GRCh38
rs759082556 Chr.3: 108162605-108162605 on Build GRCh38
rs578026911 Chr.3: 108162203-108162203 on Build GRCh38
rs139677905 Chr.3: 108162174-108162174 on Build GRCh38
rs527604264 Chr.3: 108162423-108162423 on Build GRCh38
rs972387970 Chr.3: 108162467-108162467 on Build GRCh38
rs763826129 Chr.3: 108162628-108162628 on Build GRCh38
rs952387530 Chr.3: 108162550-108162550 on Build GRCh38
rs773792727 Chr.3: 108162446-108162446 on Build GRCh38
rs768927703 Chr.3: 108162518-108162518 on Build GRCh38
rs768803214 Chr.3: 108162512-108162512 on Build GRCh38
rs1056253528 Chr.3: 108162437-108162437 on Build GRCh38
rs201830101 Chr.3: 108162476-108162476 on Build GRCh38
rs780354610 Chr.3: 108162481-108162482 on Build GRCh38
rs75763868 Chr.3: 108162496-108162496 on Build GRCh38
rs1048954746 Chr.3: 108162520-108162520 on Build GRCh38
rs11034 Chr.3: 108162314-108162314 on Build GRCh38
rs953581508 Chr.3: 108162402-108162402 on Build GRCh38
rs773660613 Chr.3: 108162557-108162557 on Build GRCh38
rs375757410 Chr.3: 108162580-108162580 on Build GRCh38
rs779269200 Chr.3: 108162530-108162530 on Build GRCh38
rs777549747 Chr.3: 108162486-108162486 on Build GRCh38
rs146677431 Chr.3: 108162616-108162616 on Build GRCh38
rs373228018 Chr.3: 108162621-108162621 on Build GRCh38
rs993369194 Chr.3: 108162185-108162185 on Build GRCh38
rs1000878732 Chr.3: 108162390-108162390 on Build GRCh38
rs1007450385 Chr.3: 108162546-108162546 on Build GRCh38
rs371511443 Chr.3: 108162558-108162558 on Build GRCh38
rs58273586 Chr.3: 108162305-108162305 on Build GRCh38
rs776821550 Chr.3: 108162597-108162598 on Build GRCh38
rs1044558551 Chr.3: 108162403-108162403 on Build GRCh38
rs758176688 Chr.3: 108162481-108162481 on Build GRCh38
rs759015407 Chr.3: 108162453-108162453 on Build GRCh38
rs948961179 Chr.3: 108162398-108162398 on Build GRCh38
rs866781501 Chr.3: 108162527-108162527 on Build GRCh38
rs751413320 Chr.3: 108162491-108162491 on Build GRCh38
rs747262059 Chr.3: 108162482-108162482 on Build GRCh38
rs1009019245 Chr.3: 108162407-108162407 on Build GRCh38
rs749359150 Chr.3: 108162515-108162515 on Build GRCh38
rs781042192 Chr.3: 108162497-108162497 on Build GRCh38
rs990382305 Chr.3: 108162369-108162370 on Build GRCh38
rs372611427 Chr.3: 108162348-108162348 on Build GRCh38
rs768904097 Chr.3: 108162437-108162437 on Build GRCh38
rs951864506 Chr.3: 108162221-108162221 on Build GRCh38
rs572198055 Chr.3: 108162603-108162603 on Build GRCh38
rs904791739 Chr.3: 108162379-108162379 on Build GRCh38
rs764786941 Chr.3: 108162615-108162615 on Build GRCh38
rs774757325 Chr.3: 108162442-108162442 on Build GRCh38
rs775672395 Chr.3: 108162426-108162426 on Build GRCh38
rs202056159 Chr.3: 108162577-108162577 on Build GRCh38
rs751345291 Chr.3: 108162633-108162633 on Build GRCh38
rs931196826 Chr.3: 108162247-108162247 on Build GRCh38
rs774201233 Chr.3: 108162392-108162392 on Build GRCh38
rs772626580 Chr.3: 108162444-108162444 on Build GRCh38
rs1027569517 Chr.3: 108162217-108162217 on Build GRCh38
rs529101351 Chr.3: 108162436-108162437 on Build GRCh38
rs956104167 Chr.3: 108162362-108162362 on Build GRCh38
rs751323403 Chr.3: 108162285-108162285 on Build GRCh38
rs984454235 Chr.3: 108162600-108162600 on Build GRCh38
rs139626836 Chr.3: 108162533-108162533 on Build GRCh38
rs1135937 Chr.3: 108162291-108162291 on Build GRCh38
rs200599808 Chr.3: 108162540-108162540 on Build GRCh38
rs966503472 Chr.3: 108162301-108162301 on Build GRCh38

View Full Product Details