Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113649100-113649612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USF3
Entrez Gene ID
205717
Gene Name
upstream transcription factor family member 3
Gene Aliases
KIAA2018
UniGene
Hs.632570
Gene Chromosome Location
Chr.3: 113648385-113696657 on Build GRCh38
Transcripts
NM_001009899.3 NR_111981.1 XM_005247208.4 XM_017005871.1 XM_017005872.1

SNP Detail

SNP ID Chromosome Location
rs1015930138 Chr.3: 113649391-113649391 on Build GRCh38
rs928106021 Chr.3: 113649241-113649241 on Build GRCh38
rs569314027 Chr.3: 113649181-113649181 on Build GRCh38
rs998495854 Chr.3: 113649251-113649251 on Build GRCh38
rs1049610110 Chr.3: 113649385-113649385 on Build GRCh38
rs143414907 Chr.3: 113649389-113649389 on Build GRCh38
rs192803501 Chr.3: 113649355-113649355 on Build GRCh38
rs939651775 Chr.3: 113649264-113649264 on Build GRCh38
rs373551487 Chr.3: 113649536-113649536 on Build GRCh38
rs530426240 Chr.3: 113649204-113649204 on Build GRCh38
rs538486961 Chr.3: 113649459-113649459 on Build GRCh38
rs1045278340 Chr.3: 113649232-113649232 on Build GRCh38
rs749076999 Chr.3: 113649179-113649179 on Build GRCh38
rs930779753 Chr.3: 113649377-113649377 on Build GRCh38
rs998756045 Chr.3: 113649486-113649486 on Build GRCh38
rs567310802 Chr.3: 113649341-113649341 on Build GRCh38
rs79162957 Chr.3: 113649528-113649528 on Build GRCh38
rs930489889 Chr.3: 113649125-113649125 on Build GRCh38
rs969821085 Chr.3: 113649504-113649504 on Build GRCh38
rs557121133 Chr.3: 113649392-113649392 on Build GRCh38
rs12492542 Chr.3: 113649325-113649325 on Build GRCh38
rs769258375 Chr.3: 113649500-113649503 on Build GRCh38
rs747736520 Chr.3: 113649365-113649365 on Build GRCh38
rs901519732 Chr.3: 113649216-113649216 on Build GRCh38
rs779776913 Chr.3: 113649139-113649139 on Build GRCh38
rs1051463 Chr.3: 113649517-113649517 on Build GRCh38
rs1031858993 Chr.3: 113649261-113649262 on Build GRCh38
rs980990635 Chr.3: 113649295-113649295 on Build GRCh38
rs551171857 Chr.3: 113649170-113649170 on Build GRCh38
rs761153532 Chr.3: 113649558-113649558 on Build GRCh38
rs962690180 Chr.3: 113649166-113649166 on Build GRCh38
rs952101886 Chr.3: 113649552-113649552 on Build GRCh38
rs371964877 Chr.3: 113649374-113649374 on Build GRCh38
rs892784271 Chr.3: 113649499-113649500 on Build GRCh38
rs1026354725 Chr.3: 113649535-113649535 on Build GRCh38
rs1053112074 Chr.3: 113649266-113649266 on Build GRCh38
rs551327269 Chr.3: 113649517-113649518 on Build GRCh38
rs973749681 Chr.3: 113649488-113649488 on Build GRCh38
rs552976828 Chr.3: 113649361-113649361 on Build GRCh38
rs1010867435 Chr.3: 113649289-113649289 on Build GRCh38
rs962588797 Chr.3: 113649440-113649440 on Build GRCh38
rs905376404 Chr.3: 113649500-113649504 on Build GRCh38
rs948424843 Chr.3: 113649184-113649184 on Build GRCh38
rs892677571 Chr.3: 113649286-113649286 on Build GRCh38
rs1029527460 Chr.3: 113649384-113649384 on Build GRCh38
rs1051464 Chr.3: 113649510-113649510 on Build GRCh38
rs1052815216 Chr.3: 113649492-113649492 on Build GRCh38
rs1011363357 Chr.3: 113649500-113649500 on Build GRCh38
rs981588704 Chr.3: 113649518-113649518 on Build GRCh38
rs542946486 Chr.3: 113649572-113649572 on Build GRCh38
rs969956448 Chr.3: 113649292-113649292 on Build GRCh38
rs1004172350 Chr.3: 113649390-113649390 on Build GRCh38
rs928192700 Chr.3: 113649526-113649526 on Build GRCh38
rs888940685 Chr.3: 113649169-113649169 on Build GRCh38
rs943203492 Chr.3: 113649573-113649573 on Build GRCh38
rs771586082 Chr.3: 113649503-113649503 on Build GRCh38
rs77076594 Chr.3: 113649364-113649364 on Build GRCh38

View Full Product Details