Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129458457-129458980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT122
Entrez Gene ID
55764
Gene Name
intraflagellar transport 122
Gene Aliases
CED, CED1, SPG, WDR10, WDR10p, WDR140
UniGene
Hs.655284
Gene Chromosome Location
Chr.3: 129440036-129520507 on Build GRCh38
Transcripts
NM_001280541.1 NM_001280545.1 NM_001280546.1 NM_018262.3 NM_052985.3 NM_052989.2 NM_052990.2 XM_005247609.1 XM_006713689.1 XM_006713691.2 XM_006713692.2 XM_006713695.2 XM_011512972.2 XM_017006817.1 XM_017006818.1 XM_017006819.1 XM_017006820.1 XM_017006821.1 XM_017006822.1 XM_017006823.1 XM_017006824.1 XM_017006825.1 XM_017006826.1 XM_017006827.1 XM_017006828.1 XM_017006829.1 XM_017006830.1 XM_017006831.1 XM_017006832.1 XM_017006833.1 XM_017006834.1 XM_017006835.1 XM_017006836.1 XM_017006838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1418823 c.237C>G p.I79M Chr.3: 129458642-129458642 on Build GRCh38
4636444 c.220G>A p.D74N Chr.3: 129458625-129458625 on Build GRCh38
5796275 c.272+1G>A p.? Chr.3: 129458678-129458678 on Build GRCh38
4644490 c.228C>T p.S76S Chr.3: 129458633-129458633 on Build GRCh38
203923 c.200G>A p.R67H Chr.3: 129458605-129458605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188207730 Chr.3: 129458930-129458930 on Build GRCh38
rs372702964 Chr.3: 129458595-129458595 on Build GRCh38
rs989176064 Chr.3: 129458627-129458627 on Build GRCh38
rs554654606 Chr.3: 129458873-129458873 on Build GRCh38
rs1012943423 Chr.3: 129458632-129458632 on Build GRCh38
rs1037237931 Chr.3: 129458915-129458915 on Build GRCh38
rs779703788 Chr.3: 129458565-129458565 on Build GRCh38
rs181971625 Chr.3: 129458604-129458604 on Build GRCh38
rs181000640 Chr.3: 129458938-129458938 on Build GRCh38
rs1037684097 Chr.3: 129458802-129458802 on Build GRCh38
rs547691100 Chr.3: 129458699-129458699 on Build GRCh38
rs376819101 Chr.3: 129458717-129458717 on Build GRCh38
rs369525803 Chr.3: 129458634-129458634 on Build GRCh38
rs977549771 Chr.3: 129458635-129458637 on Build GRCh38
rs775693589 Chr.3: 129458656-129458656 on Build GRCh38
rs1030082247 Chr.3: 129458832-129458832 on Build GRCh38
rs755976988 Chr.3: 129458719-129458719 on Build GRCh38
rs767510838 Chr.3: 129458702-129458703 on Build GRCh38
rs928796428 Chr.3: 129458648-129458648 on Build GRCh38
rs532583426 Chr.3: 129458677-129458677 on Build GRCh38
rs969011558 Chr.3: 129458657-129458657 on Build GRCh38
rs941289316 Chr.3: 129458863-129458863 on Build GRCh38
rs1057268089 Chr.3: 129458580-129458580 on Build GRCh38
rs939205700 Chr.3: 129458926-129458926 on Build GRCh38
rs751713457 Chr.3: 129458690-129458690 on Build GRCh38
rs749137520 Chr.3: 129458610-129458610 on Build GRCh38
rs955757483 Chr.3: 129458765-129458765 on Build GRCh38
rs953695771 Chr.3: 129458824-129458824 on Build GRCh38
rs759016671 Chr.3: 129458686-129458688 on Build GRCh38
rs537066272 Chr.3: 129458947-129458947 on Build GRCh38
rs75111418 Chr.3: 129458772-129458772 on Build GRCh38
rs926701737 Chr.3: 129458623-129458623 on Build GRCh38
rs372483083 Chr.3: 129458678-129458678 on Build GRCh38
rs758585414 Chr.3: 129458549-129458549 on Build GRCh38
rs773902277 Chr.3: 129458634-129458643 on Build GRCh38
rs563499183 Chr.3: 129458506-129458506 on Build GRCh38
rs779711516 Chr.3: 129458605-129458605 on Build GRCh38
rs895931100 Chr.3: 129458950-129458950 on Build GRCh38
rs144140226 Chr.3: 129458619-129458619 on Build GRCh38
rs774104621 Chr.3: 129458613-129458613 on Build GRCh38
rs758705902 Chr.3: 129458538-129458538 on Build GRCh38
rs375738861 Chr.3: 129458556-129458556 on Build GRCh38
rs114519508 Chr.3: 129458805-129458805 on Build GRCh38
rs868486162 Chr.3: 129458920-129458920 on Build GRCh38
rs985172799 Chr.3: 129458835-129458835 on Build GRCh38
rs776191615 Chr.3: 129458766-129458766 on Build GRCh38
rs993589780 Chr.3: 129458578-129458578 on Build GRCh38
rs530936539 Chr.3: 129458514-129458514 on Build GRCh38
rs751803328 Chr.3: 129458588-129458588 on Build GRCh38
rs993550643 Chr.3: 129458818-129458818 on Build GRCh38
rs781338240 Chr.3: 129458603-129458603 on Build GRCh38
rs909765986 Chr.3: 129458850-129458850 on Build GRCh38
rs139945948 Chr.3: 129458643-129458643 on Build GRCh38
rs771422778 Chr.3: 129458624-129458624 on Build GRCh38
rs2285349 Chr.3: 129458503-129458503 on Build GRCh38
rs370660486 Chr.3: 129458705-129458705 on Build GRCh38
rs772835552 Chr.3: 129458633-129458633 on Build GRCh38
rs190278519 Chr.3: 129458532-129458532 on Build GRCh38

View Full Product Details