Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141778064-141778565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK7
Entrez Gene ID
131890
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
GPRK7
UniGene
Hs.680654
Gene Chromosome Location
Chr.3: 141763127-141817373 on Build GRCh38
Transcripts
NM_139209.2 XM_017005717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
261197 c.199C>T p.R67C Chr.3: 141778483-141778483 on Build GRCh38
6096508 c.54G>T p.Q18H Chr.3: 141778338-141778338 on Build GRCh38
1752946 c.43G>A p.A15T Chr.3: 141778327-141778327 on Build GRCh38
7267198 c.12delG p.A6fs*5 Chr.3: 141778296-141778296 on Build GRCh38
4319547 c.210C>T p.F70F Chr.3: 141778494-141778494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35145570 Chr.3: 141778264-141778265 on Build GRCh38
rs150651806 Chr.3: 141778477-141778477 on Build GRCh38
rs563165279 Chr.3: 141778322-141778322 on Build GRCh38
rs569108851 Chr.3: 141778356-141778356 on Build GRCh38
rs761208239 Chr.3: 141778253-141778253 on Build GRCh38
rs113260364 Chr.3: 141778099-141778099 on Build GRCh38
rs143776792 Chr.3: 141778389-141778389 on Build GRCh38
rs866339926 Chr.3: 141778291-141778291 on Build GRCh38
rs377464259 Chr.3: 141778301-141778301 on Build GRCh38
rs200671434 Chr.3: 141778336-141778336 on Build GRCh38
rs142109851 Chr.3: 141778327-141778327 on Build GRCh38
rs764098462 Chr.3: 141778378-141778378 on Build GRCh38
rs141897427 Chr.3: 141778465-141778465 on Build GRCh38
rs779099651 Chr.3: 141778387-141778387 on Build GRCh38
rs147427020 Chr.3: 141778520-141778520 on Build GRCh38
rs11715715 Chr.3: 141778231-141778231 on Build GRCh38
rs143028766 Chr.3: 141778518-141778518 on Build GRCh38
rs757437128 Chr.3: 141778386-141778386 on Build GRCh38
rs769188536 Chr.3: 141778338-141778338 on Build GRCh38
rs774924835 Chr.3: 141778235-141778235 on Build GRCh38
rs780012984 Chr.3: 141778433-141778433 on Build GRCh38
rs756073341 Chr.3: 141778376-141778376 on Build GRCh38
rs760533722 Chr.3: 141778455-141778455 on Build GRCh38
rs34537072 Chr.3: 141778426-141778426 on Build GRCh38
rs770457746 Chr.3: 141778416-141778416 on Build GRCh38
rs369135654 Chr.3: 141778273-141778273 on Build GRCh38
rs554122518 Chr.3: 141778226-141778226 on Build GRCh38
rs149923467 Chr.3: 141778535-141778535 on Build GRCh38
rs758755551 Chr.3: 141778480-141778480 on Build GRCh38
rs949959698 Chr.3: 141778360-141778360 on Build GRCh38
rs571411294 Chr.3: 141778194-141778194 on Build GRCh38
rs752524519 Chr.3: 141778366-141778366 on Build GRCh38
rs377053600 Chr.3: 141778100-141778100 on Build GRCh38
rs774940551 Chr.3: 141778442-141778442 on Build GRCh38
rs759147321 Chr.3: 141778359-141778359 on Build GRCh38
rs201493770 Chr.3: 141778495-141778495 on Build GRCh38
rs1533499 Chr.3: 141778132-141778132 on Build GRCh38
rs765257624 Chr.3: 141778533-141778533 on Build GRCh38
rs747391734 Chr.3: 141778399-141778399 on Build GRCh38
rs532690516 Chr.3: 141778283-141778283 on Build GRCh38
rs904956167 Chr.3: 141778091-141778091 on Build GRCh38
rs552090044 Chr.3: 141778178-141778178 on Build GRCh38
rs1037865881 Chr.3: 141778341-141778341 on Build GRCh38
rs534266012 Chr.3: 141778162-141778162 on Build GRCh38
rs758675614 Chr.3: 141778392-141778392 on Build GRCh38
rs868864461 Chr.3: 141778317-141778317 on Build GRCh38
rs186188558 Chr.3: 141778202-141778202 on Build GRCh38
rs778452393 Chr.3: 141778516-141778516 on Build GRCh38
rs772751848 Chr.3: 141778268-141778268 on Build GRCh38
rs1011157208 Chr.3: 141778212-141778212 on Build GRCh38
rs761814269 Chr.3: 141778474-141778474 on Build GRCh38
rs112016084 Chr.3: 141778410-141778410 on Build GRCh38
rs968794698 Chr.3: 141778197-141778197 on Build GRCh38
rs750048804 Chr.3: 141778376-141778377 on Build GRCh38
rs189817337 Chr.3: 141778221-141778221 on Build GRCh38
rs762526012 Chr.3: 141778352-141778352 on Build GRCh38
rs750522033 Chr.3: 141778479-141778479 on Build GRCh38
rs548619860 Chr.3: 141778129-141778129 on Build GRCh38
rs1017585708 Chr.3: 141778427-141778427 on Build GRCh38
rs761904077 Chr.3: 141778527-141778527 on Build GRCh38
rs373761431 Chr.3: 141778307-141778307 on Build GRCh38
rs565619405 Chr.3: 141778196-141778196 on Build GRCh38
rs745801577 Chr.3: 141778323-141778323 on Build GRCh38
rs181765052 Chr.3: 141778201-141778201 on Build GRCh38
rs1002306047 Chr.3: 141778180-141778180 on Build GRCh38
rs375407859 Chr.3: 141778272-141778272 on Build GRCh38
rs182834100 Chr.3: 141778248-141778248 on Build GRCh38
rs954695259 Chr.3: 141778175-141778175 on Build GRCh38
rs35998945 Chr.3: 141778391-141778391 on Build GRCh38
rs542940579 Chr.3: 141778222-141778222 on Build GRCh38
rs148101386 Chr.3: 141778420-141778420 on Build GRCh38
rs1533501 Chr.3: 141778308-141778308 on Build GRCh38
rs758070401 Chr.3: 141778229-141778229 on Build GRCh38
rs1055657448 Chr.3: 141778430-141778430 on Build GRCh38
rs781727846 Chr.3: 141778402-141778402 on Build GRCh38
rs370871734 Chr.3: 141778425-141778425 on Build GRCh38
rs988391890 Chr.3: 141778496-141778496 on Build GRCh38
rs554482696 Chr.3: 141778525-141778525 on Build GRCh38
rs1006369977 Chr.3: 141778098-141778098 on Build GRCh38
rs764698179 Chr.3: 141778259-141778259 on Build GRCh38
rs34429284 Chr.3: 141778526-141778526 on Build GRCh38
rs55712018 Chr.3: 141778326-141778326 on Build GRCh38
rs768833186 Chr.3: 141778401-141778401 on Build GRCh38
rs890718052 Chr.3: 141778408-141778408 on Build GRCh38
rs755390654 Chr.3: 141778486-141778486 on Build GRCh38
rs199734533 Chr.3: 141778331-141778331 on Build GRCh38
rs763749436 Chr.3: 141778462-141778462 on Build GRCh38
rs762567903 Chr.3: 141778270-141778270 on Build GRCh38
rs34592817 Chr.3: 141778393-141778393 on Build GRCh38
rs759986205 Chr.3: 141778245-141778245 on Build GRCh38
rs759235194 Chr.3: 141778421-141778421 on Build GRCh38
rs772660757 Chr.3: 141778342-141778342 on Build GRCh38
rs774012349 Chr.3: 141778358-141778358 on Build GRCh38
rs371087666 Chr.3: 141778316-141778316 on Build GRCh38
rs1533500 Chr.3: 141778278-141778278 on Build GRCh38
rs767249385 Chr.3: 141778282-141778282 on Build GRCh38
rs527727725 Chr.3: 141778362-141778362 on Build GRCh38
rs773818685 Chr.3: 141778418-141778418 on Build GRCh38
rs1030745612 Chr.3: 141778147-141778147 on Build GRCh38
rs1051988672 Chr.3: 141778379-141778379 on Build GRCh38
rs79064253 Chr.3: 141778498-141778498 on Build GRCh38
rs756047832 Chr.3: 141778296-141778296 on Build GRCh38
rs780618575 Chr.3: 141778396-141778396 on Build GRCh38
rs751671473 Chr.3: 141778483-141778483 on Build GRCh38

View Full Product Details