Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141778453-141778948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK7
Entrez Gene ID
131890
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
GPRK7
UniGene
Hs.680654
Gene Chromosome Location
Chr.3: 141763127-141817373 on Build GRCh38
Transcripts
NM_139209.2 XM_017005717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
261197 c.199C>T p.R67C Chr.3: 141778483-141778483 on Build GRCh38
6203255 c.589C>T p.L197L Chr.3: 141778873-141778873 on Build GRCh38
6038678 c.335C>A p.T112N Chr.3: 141778619-141778619 on Build GRCh38
4319547 c.210C>T p.F70F Chr.3: 141778494-141778494 on Build GRCh38
1039255 c.319C>T p.Q107* Chr.3: 141778603-141778603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778266106 Chr.3: 141778596-141778596 on Build GRCh38
rs150651806 Chr.3: 141778477-141778477 on Build GRCh38
rs912098049 Chr.3: 141778855-141778855 on Build GRCh38
rs753046646 Chr.3: 141778576-141778576 on Build GRCh38
rs764488767 Chr.3: 141778738-141778738 on Build GRCh38
rs781657147 Chr.3: 141778575-141778575 on Build GRCh38
rs1040642711 Chr.3: 141778704-141778704 on Build GRCh38
rs56199819 Chr.3: 141778551-141778551 on Build GRCh38
rs150445233 Chr.3: 141778728-141778728 on Build GRCh38
rs770761010 Chr.3: 141778692-141778692 on Build GRCh38
rs757955543 Chr.3: 141778605-141778605 on Build GRCh38
rs754472954 Chr.3: 141778777-141778777 on Build GRCh38
rs370316169 Chr.3: 141778552-141778552 on Build GRCh38
rs752439966 Chr.3: 141778792-141778792 on Build GRCh38
rs145001481 Chr.3: 141778561-141778561 on Build GRCh38
rs760050500 Chr.3: 141778858-141778858 on Build GRCh38
rs765538885 Chr.3: 141778880-141778880 on Build GRCh38
rs763488179 Chr.3: 141778853-141778853 on Build GRCh38
rs1034234897 Chr.3: 141778919-141778919 on Build GRCh38
rs944950438 Chr.3: 141778691-141778691 on Build GRCh38
rs55707760 Chr.3: 141778871-141778871 on Build GRCh38
rs780812559 Chr.3: 141778782-141778782 on Build GRCh38
rs774273604 Chr.3: 141778640-141778640 on Build GRCh38
rs147427020 Chr.3: 141778520-141778520 on Build GRCh38
rs773113276 Chr.3: 141778672-141778672 on Build GRCh38
rs777770174 Chr.3: 141778797-141778797 on Build GRCh38
rs143028766 Chr.3: 141778518-141778518 on Build GRCh38
rs747965154 Chr.3: 141778625-141778625 on Build GRCh38
rs773607331 Chr.3: 141778916-141778916 on Build GRCh38
rs546694683 Chr.3: 141778887-141778887 on Build GRCh38
rs770098637 Chr.3: 141778911-141778911 on Build GRCh38
rs142108127 Chr.3: 141778641-141778641 on Build GRCh38
rs779566185 Chr.3: 141778724-141778725 on Build GRCh38
rs776745087 Chr.3: 141778897-141778897 on Build GRCh38
rs775522873 Chr.3: 141778812-141778812 on Build GRCh38
rs766522493 Chr.3: 141778634-141778634 on Build GRCh38
rs149923467 Chr.3: 141778535-141778535 on Build GRCh38
rs758755551 Chr.3: 141778480-141778480 on Build GRCh38
rs777130690 Chr.3: 141778851-141778851 on Build GRCh38
rs778988214 Chr.3: 141778671-141778671 on Build GRCh38
rs918991548 Chr.3: 141778789-141778789 on Build GRCh38
rs201493770 Chr.3: 141778495-141778495 on Build GRCh38
rs367747374 Chr.3: 141778803-141778803 on Build GRCh38
rs774557918 Chr.3: 141778710-141778710 on Build GRCh38
rs756491318 Chr.3: 141778648-141778648 on Build GRCh38
rs34769632 Chr.3: 141778627-141778627 on Build GRCh38
rs765257624 Chr.3: 141778533-141778533 on Build GRCh38
rs753481177 Chr.3: 141778872-141778872 on Build GRCh38
rs145262857 Chr.3: 141778666-141778666 on Build GRCh38
rs758007042 Chr.3: 141778558-141778558 on Build GRCh38
rs763615719 Chr.3: 141778921-141778921 on Build GRCh38
rs745778241 Chr.3: 141778802-141778802 on Build GRCh38
rs111567991 Chr.3: 141778846-141778846 on Build GRCh38
rs35318124 Chr.3: 141778664-141778664 on Build GRCh38
rs765662924 Chr.3: 141778763-141778763 on Build GRCh38
rs149627012 Chr.3: 141778770-141778770 on Build GRCh38
rs143277568 Chr.3: 141778828-141778828 on Build GRCh38
rs756403402 Chr.3: 141778587-141778587 on Build GRCh38
rs778452393 Chr.3: 141778516-141778516 on Build GRCh38
rs761814269 Chr.3: 141778474-141778474 on Build GRCh38
rs202140465 Chr.3: 141778800-141778800 on Build GRCh38
rs200056259 Chr.3: 141778650-141778650 on Build GRCh38
rs773308032 Chr.3: 141778628-141778628 on Build GRCh38
rs766990220 Chr.3: 141778923-141778923 on Build GRCh38
rs750522033 Chr.3: 141778479-141778479 on Build GRCh38
rs761904077 Chr.3: 141778527-141778527 on Build GRCh38
rs780171476 Chr.3: 141778890-141778890 on Build GRCh38
rs762058612 Chr.3: 141778904-141778904 on Build GRCh38
rs750928230 Chr.3: 141778849-141778849 on Build GRCh38
rs779669917 Chr.3: 141778652-141778652 on Build GRCh38
rs200708791 Chr.3: 141778609-141778609 on Build GRCh38
rs760853907 Chr.3: 141778734-141778734 on Build GRCh38
rs112167264 Chr.3: 141778811-141778811 on Build GRCh38
rs550296305 Chr.3: 141778863-141778863 on Build GRCh38
rs988391890 Chr.3: 141778496-141778496 on Build GRCh38
rs773146855 Chr.3: 141778655-141778655 on Build GRCh38
rs759547557 Chr.3: 141778711-141778711 on Build GRCh38
rs554482696 Chr.3: 141778525-141778525 on Build GRCh38
rs141991963 Chr.3: 141778827-141778827 on Build GRCh38
rs781008881 Chr.3: 141778665-141778665 on Build GRCh38
rs138267509 Chr.3: 141778758-141778758 on Build GRCh38
rs34429284 Chr.3: 141778526-141778526 on Build GRCh38
rs747092764 Chr.3: 141778894-141778894 on Build GRCh38
rs755807570 Chr.3: 141778795-141778795 on Build GRCh38
rs56019094 Chr.3: 141778870-141778870 on Build GRCh38
rs755390654 Chr.3: 141778486-141778486 on Build GRCh38
rs779397723 Chr.3: 141778608-141778608 on Build GRCh38
rs770693714 Chr.3: 141778801-141778801 on Build GRCh38
rs760942018 Chr.3: 141778816-141778816 on Build GRCh38
rs865797477 Chr.3: 141778607-141778607 on Build GRCh38
rs146703848 Chr.3: 141778649-141778649 on Build GRCh38
rs752858372 Chr.3: 141778642-141778642 on Build GRCh38
rs749744947 Chr.3: 141778601-141778601 on Build GRCh38
rs778737262 Chr.3: 141778877-141778877 on Build GRCh38
rs576881901 Chr.3: 141778573-141778573 on Build GRCh38
rs73872327 Chr.3: 141778685-141778685 on Build GRCh38
rs576789062 Chr.3: 141778818-141778818 on Build GRCh38
rs757033589 Chr.3: 141778873-141778873 on Build GRCh38
rs768177089 Chr.3: 141778613-141778613 on Build GRCh38
rs755170134 Chr.3: 141778631-141778631 on Build GRCh38
rs140605792 Chr.3: 141778712-141778712 on Build GRCh38
rs771982384 Chr.3: 141778882-141778882 on Build GRCh38
rs79064253 Chr.3: 141778498-141778498 on Build GRCh38
rs56070798 Chr.3: 141778623-141778623 on Build GRCh38
rs754185819 Chr.3: 141778747-141778747 on Build GRCh38
rs751671473 Chr.3: 141778483-141778483 on Build GRCh38

View Full Product Details