Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141807792-141808305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK7
Entrez Gene ID
131890
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
GPRK7
UniGene
Hs.680654
Gene Chromosome Location
Chr.3: 141763127-141817373 on Build GRCh38
Transcripts
NM_139209.2 XM_017005717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1291619 c.1250A>G p.D417G Chr.3: 141807844-141807844 on Build GRCh38
5355190 c.1220C>T p.T407I Chr.3: 141807814-141807814 on Build GRCh38
4437041 c.1273G>C p.D425H Chr.3: 141807867-141807867 on Build GRCh38
6863938 c.1236_1237insA p.F414fs*4 Chr.3: 141807830-141807831 on Build GRCh38
729007 c.1222C>G p.L408V Chr.3: 141807816-141807816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150028271 Chr.3: 141808161-141808161 on Build GRCh38
rs13317804 Chr.3: 141808153-141808153 on Build GRCh38
rs767919973 Chr.3: 141807824-141807824 on Build GRCh38
rs28546331 Chr.3: 141808125-141808125 on Build GRCh38
rs759819824 Chr.3: 141807942-141807942 on Build GRCh38
rs199949873 Chr.3: 141807930-141807930 on Build GRCh38
rs200924184 Chr.3: 141807825-141807825 on Build GRCh38
rs919091883 Chr.3: 141808178-141808178 on Build GRCh38
rs570540445 Chr.3: 141808208-141808208 on Build GRCh38
rs982083968 Chr.3: 141808238-141808238 on Build GRCh38
rs779581517 Chr.3: 141807883-141807883 on Build GRCh38
rs749210636 Chr.3: 141807917-141807917 on Build GRCh38
rs747946920 Chr.3: 141807969-141807969 on Build GRCh38
rs201761300 Chr.3: 141807885-141807885 on Build GRCh38
rs909868552 Chr.3: 141808152-141808152 on Build GRCh38
rs772820959 Chr.3: 141807966-141807966 on Build GRCh38
rs769383075 Chr.3: 141808048-141808048 on Build GRCh38
rs547559166 Chr.3: 141808017-141808017 on Build GRCh38
rs185869215 Chr.3: 141808015-141808015 on Build GRCh38
rs764521169 Chr.3: 141807856-141807856 on Build GRCh38
rs929177304 Chr.3: 141808217-141808217 on Build GRCh38
rs754667711 Chr.3: 141807891-141807891 on Build GRCh38
rs757873502 Chr.3: 141807862-141807862 on Build GRCh38
rs762356540 Chr.3: 141807964-141807964 on Build GRCh38
rs761050087 Chr.3: 141807957-141807957 on Build GRCh38
rs1042026563 Chr.3: 141807935-141807935 on Build GRCh38
rs937117539 Chr.3: 141807858-141807858 on Build GRCh38
rs760603900 Chr.3: 141807841-141807843 on Build GRCh38
rs769578596 Chr.3: 141807908-141807908 on Build GRCh38
rs747965204 Chr.3: 141807907-141807907 on Build GRCh38
rs1027712296 Chr.3: 141808151-141808151 on Build GRCh38
rs775631304 Chr.3: 141807954-141807954 on Build GRCh38
rs754338338 Chr.3: 141807960-141807960 on Build GRCh38
rs777595232 Chr.3: 141807916-141807916 on Build GRCh38
rs754321266 Chr.3: 141807861-141807861 on Build GRCh38
rs970333285 Chr.3: 141808097-141808097 on Build GRCh38
rs774394310 Chr.3: 141807938-141807938 on Build GRCh38
rs530687886 Chr.3: 141808016-141808018 on Build GRCh38
rs781203350 Chr.3: 141807906-141807906 on Build GRCh38
rs971702936 Chr.3: 141808187-141808187 on Build GRCh38
rs180894394 Chr.3: 141807988-141807988 on Build GRCh38
rs1051545497 Chr.3: 141808258-141808258 on Build GRCh38
rs900482707 Chr.3: 141808051-141808051 on Build GRCh38
rs751303405 Chr.3: 141807884-141807884 on Build GRCh38
rs368907384 Chr.3: 141807945-141807945 on Build GRCh38
rs760960407 Chr.3: 141807842-141807842 on Build GRCh38
rs139307968 Chr.3: 141808106-141808106 on Build GRCh38

View Full Product Details