Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149027365-149027869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYG1
Entrez Gene ID
2992
Gene Name
glycogenin 1
Gene Aliases
GSD15, GYG
UniGene
Hs.477892
Gene Chromosome Location
Chr.3: 148991408-149027669 on Build GRCh38
Transcripts
NM_001184720.1 NM_001184721.1 NM_004130.3 XM_017006275.1 XM_017006276.1

SNP Detail

SNP ID Chromosome Location
rs1029272757 Chr.3: 149027663-149027663 on Build GRCh38
rs1031587240 Chr.3: 149027639-149027642 on Build GRCh38
rs574568513 Chr.3: 149027648-149027648 on Build GRCh38
rs878943184 Chr.3: 149027644-149027644 on Build GRCh38
rs755953405 Chr.3: 149027593-149027593 on Build GRCh38
rs909542265 Chr.3: 149027500-149027500 on Build GRCh38
rs940782689 Chr.3: 149027729-149027731 on Build GRCh38
rs990159302 Chr.3: 149027657-149027657 on Build GRCh38
rs752476168 Chr.3: 149027545-149027545 on Build GRCh38
rs777656062 Chr.3: 149027777-149027777 on Build GRCh38
rs563768369 Chr.3: 149027780-149027780 on Build GRCh38
rs770296525 Chr.3: 149027840-149027840 on Build GRCh38
rs879787830 Chr.3: 149027642-149027642 on Build GRCh38
rs3347 Chr.3: 149027449-149027449 on Build GRCh38
rs1051470504 Chr.3: 149027735-149027735 on Build GRCh38
rs577915466 Chr.3: 149027486-149027486 on Build GRCh38
rs1045462143 Chr.3: 149027838-149027838 on Build GRCh38
rs917485718 Chr.3: 149027404-149027404 on Build GRCh38
rs997999603 Chr.3: 149027586-149027586 on Build GRCh38
rs911890907 Chr.3: 149027718-149027718 on Build GRCh38
rs866297796 Chr.3: 149027706-149027706 on Build GRCh38
rs1020337466 Chr.3: 149027824-149027824 on Build GRCh38
rs572380867 Chr.3: 149027407-149027407 on Build GRCh38
rs967152093 Chr.3: 149027623-149027624 on Build GRCh38
rs995170824 Chr.3: 149027506-149027506 on Build GRCh38
rs998600689 Chr.3: 149027625-149027625 on Build GRCh38
rs558145373 Chr.3: 149027483-149027483 on Build GRCh38
rs941150799 Chr.3: 149027553-149027553 on Build GRCh38
rs543900782 Chr.3: 149027773-149027773 on Build GRCh38
rs376685571 Chr.3: 149027772-149027772 on Build GRCh38
rs953746952 Chr.3: 149027687-149027687 on Build GRCh38
rs148445852 Chr.3: 149027830-149027831 on Build GRCh38
rs898959823 Chr.3: 149027504-149027504 on Build GRCh38
rs749278437 Chr.3: 149027781-149027781 on Build GRCh38
rs998610553 Chr.3: 149027842-149027842 on Build GRCh38
rs899672290 Chr.3: 149027569-149027569 on Build GRCh38
rs1009480131 Chr.3: 149027810-149027810 on Build GRCh38
rs1056015568 Chr.3: 149027792-149027792 on Build GRCh38
rs537865633 Chr.3: 149027394-149027395 on Build GRCh38
rs1036194421 Chr.3: 149027496-149027496 on Build GRCh38
rs1023962820 Chr.3: 149027600-149027600 on Build GRCh38
rs987468792 Chr.3: 149027708-149027708 on Build GRCh38
rs993501890 Chr.3: 149027390-149027390 on Build GRCh38
rs112973519 Chr.3: 149027529-149027530 on Build GRCh38
rs1039415724 Chr.3: 149027568-149027568 on Build GRCh38

View Full Product Details