Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151438186-151438678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF10
Entrez Gene ID
285313
Gene Name
immunoglobulin superfamily member 10
Gene Aliases
CMF608
UniGene
Hs.58561
Gene Chromosome Location
Chr.3: 151435051-151619926 on Build GRCh38
Transcripts
NM_001178145.1 NM_001178146.1 NM_178822.4 XM_011512708.2 XM_011512709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6163784 c.6101G>T p.R2034I Chr.3: 151438460-151438460 on Build GRCh38
3915158 c.5980C>T p.H1994Y Chr.3: 151438581-151438581 on Build GRCh38
4553938 c.6069G>A p.G2023G Chr.3: 151438492-151438492 on Build GRCh38
4114500 c.6114C>T p.A2038A Chr.3: 151438447-151438447 on Build GRCh38
4774698 c.5982C>T p.H1994H Chr.3: 151438579-151438579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149524314 Chr.3: 151438471-151438471 on Build GRCh38
rs763368994 Chr.3: 151438631-151438632 on Build GRCh38
rs778522469 Chr.3: 151438224-151438224 on Build GRCh38
rs148237311 Chr.3: 151438576-151438576 on Build GRCh38
rs527825904 Chr.3: 151438324-151438324 on Build GRCh38
rs771352318 Chr.3: 151438344-151438344 on Build GRCh38
rs754352952 Chr.3: 151438459-151438459 on Build GRCh38
rs779485240 Chr.3: 151438551-151438551 on Build GRCh38
rs765668994 Chr.3: 151438628-151438628 on Build GRCh38
rs1054279356 Chr.3: 151438511-151438511 on Build GRCh38
rs967058408 Chr.3: 151438534-151438535 on Build GRCh38
rs199577194 Chr.3: 151438571-151438571 on Build GRCh38
rs138856486 Chr.3: 151438257-151438257 on Build GRCh38
rs751741616 Chr.3: 151438310-151438310 on Build GRCh38
rs779102895 Chr.3: 151438460-151438460 on Build GRCh38
rs374514917 Chr.3: 151438384-151438384 on Build GRCh38
rs760865266 Chr.3: 151438543-151438543 on Build GRCh38
rs764309430 Chr.3: 151438615-151438615 on Build GRCh38
rs374148063 Chr.3: 151438338-151438338 on Build GRCh38
rs780741423 Chr.3: 151438561-151438561 on Build GRCh38
rs760750544 Chr.3: 151438215-151438215 on Build GRCh38
rs747106942 Chr.3: 151438228-151438228 on Build GRCh38
rs140636759 Chr.3: 151438285-151438285 on Build GRCh38
rs78115932 Chr.3: 151438225-151438225 on Build GRCh38
rs750903543 Chr.3: 151438629-151438629 on Build GRCh38
rs75202100 Chr.3: 151438308-151438308 on Build GRCh38
rs747787349 Chr.3: 151438565-151438565 on Build GRCh38
rs768022610 Chr.3: 151438387-151438387 on Build GRCh38
rs764217271 Chr.3: 151438545-151438545 on Build GRCh38
rs753647662 Chr.3: 151438221-151438221 on Build GRCh38
rs775309595 Chr.3: 151438353-151438353 on Build GRCh38
rs780842125 Chr.3: 151438486-151438486 on Build GRCh38
rs747617244 Chr.3: 151438495-151438495 on Build GRCh38
rs369357367 Chr.3: 151438505-151438505 on Build GRCh38
rs761633551 Chr.3: 151438298-151438298 on Build GRCh38
rs753961925 Chr.3: 151438617-151438617 on Build GRCh38
rs762466411 Chr.3: 151438627-151438627 on Build GRCh38
rs759251237 Chr.3: 151438580-151438580 on Build GRCh38
rs370277399 Chr.3: 151438247-151438247 on Build GRCh38
rs776651746 Chr.3: 151438292-151438292 on Build GRCh38
rs770294032 Chr.3: 151438564-151438565 on Build GRCh38
rs975322538 Chr.3: 151438246-151438246 on Build GRCh38
rs760972781 Chr.3: 151438424-151438424 on Build GRCh38
rs768260632 Chr.3: 151438371-151438371 on Build GRCh38
rs368664622 Chr.3: 151438372-151438372 on Build GRCh38
rs749681801 Chr.3: 151438342-151438342 on Build GRCh38
rs752703973 Chr.3: 151438438-151438438 on Build GRCh38
rs748559170 Chr.3: 151438236-151438236 on Build GRCh38
rs750208612 Chr.3: 151438303-151438303 on Build GRCh38
rs201909176 Chr.3: 151438302-151438302 on Build GRCh38
rs756157146 Chr.3: 151438448-151438448 on Build GRCh38
rs146314482 Chr.3: 151438656-151438656 on Build GRCh38
rs570208083 Chr.3: 151438567-151438567 on Build GRCh38
rs149372803 Chr.3: 151438484-151438484 on Build GRCh38
rs371854370 Chr.3: 151438431-151438431 on Build GRCh38
rs757807362 Chr.3: 151438548-151438548 on Build GRCh38
rs776591826 Chr.3: 151438258-151438259 on Build GRCh38
rs1014505628 Chr.3: 151438466-151438466 on Build GRCh38
rs760957111 Chr.3: 151438605-151438605 on Build GRCh38
rs775593519 Chr.3: 151438602-151438602 on Build GRCh38
rs141482593 Chr.3: 151438489-151438489 on Build GRCh38
rs746593123 Chr.3: 151438362-151438362 on Build GRCh38
rs377142937 Chr.3: 151438226-151438226 on Build GRCh38
rs138224788 Chr.3: 151438428-151438428 on Build GRCh38
rs754946545 Chr.3: 151438230-151438230 on Build GRCh38
rs781113596 Chr.3: 151438318-151438318 on Build GRCh38
rs762858801 Chr.3: 151438517-151438517 on Build GRCh38
rs201261892 Chr.3: 151438583-151438583 on Build GRCh38
rs1051552779 Chr.3: 151438382-151438382 on Build GRCh38
rs141250073 Chr.3: 151438578-151438578 on Build GRCh38
rs771834714 Chr.3: 151438271-151438271 on Build GRCh38
rs886431262 Chr.3: 151438350-151438350 on Build GRCh38
rs764102480 Chr.3: 151438218-151438218 on Build GRCh38
rs757412694 Chr.3: 151438644-151438644 on Build GRCh38
rs773068128 Chr.3: 151438411-151438411 on Build GRCh38
rs762768763 Chr.3: 151438417-151438417 on Build GRCh38
rs767970971 Chr.3: 151438235-151438235 on Build GRCh38
rs774284539 Chr.3: 151438579-151438579 on Build GRCh38
rs746712810 Chr.3: 151438476-151438476 on Build GRCh38
rs752247874 Chr.3: 151438210-151438210 on Build GRCh38
rs768826828 Chr.3: 151438215-151438217 on Build GRCh38
rs758713177 Chr.3: 151438636-151438636 on Build GRCh38
rs778425302 Chr.3: 151438457-151438457 on Build GRCh38
rs375721071 Chr.3: 151438525-151438525 on Build GRCh38
rs145634427 Chr.3: 151438316-151438316 on Build GRCh38
rs34478732 Chr.3: 151438408-151438409 on Build GRCh38
rs758133735 Chr.3: 151438305-151438305 on Build GRCh38
rs371437973 Chr.3: 151438507-151438507 on Build GRCh38
rs767644999 Chr.3: 151438533-151438533 on Build GRCh38
rs774015729 Chr.3: 151438522-151438522 on Build GRCh38
rs962440009 Chr.3: 151438624-151438624 on Build GRCh38
rs982493163 Chr.3: 151438258-151438258 on Build GRCh38
rs1003877453 Chr.3: 151438586-151438586 on Build GRCh38
rs912530027 Chr.3: 151438386-151438386 on Build GRCh38
rs761837266 Chr.3: 151438466-151438467 on Build GRCh38
rs200998219 Chr.3: 151438222-151438222 on Build GRCh38
rs752390474 Chr.3: 151438649-151438649 on Build GRCh38
rs751861050 Chr.3: 151438426-151438426 on Build GRCh38
rs144167004 Chr.3: 151438555-151438555 on Build GRCh38
rs146146480 Chr.3: 151438613-151438614 on Build GRCh38
rs752894684 Chr.3: 151438539-151438539 on Build GRCh38
rs908711998 Chr.3: 151438277-151438277 on Build GRCh38
rs777237629 Chr.3: 151438573-151438573 on Build GRCh38
rs760233090 Chr.3: 151438279-151438279 on Build GRCh38
rs775221370 Chr.3: 151438273-151438273 on Build GRCh38
rs550459840 Chr.3: 151438562-151438562 on Build GRCh38
rs761874663 Chr.3: 151438381-151438381 on Build GRCh38
rs757717712 Chr.3: 151438470-151438470 on Build GRCh38
rs759863150 Chr.3: 151438532-151438532 on Build GRCh38
rs150254783 Chr.3: 151438231-151438231 on Build GRCh38
rs922497375 Chr.3: 151438261-151438261 on Build GRCh38
rs1015385031 Chr.3: 151438609-151438609 on Build GRCh38
rs773684022 Chr.3: 151438253-151438253 on Build GRCh38
rs770756297 Chr.3: 151438519-151438519 on Build GRCh38

View Full Product Details