Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160268082-160268613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT80
Entrez Gene ID
57560
Gene Name
intraflagellar transport 80
Gene Aliases
ATD2, SRTD2, WDR56
UniGene
Hs.478095
Gene Chromosome Location
Chr.3: 160256986-160399532 on Build GRCh38
Transcripts
NM_001190241.1 NM_001190242.1 NM_020800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7108186 c.2139T>A p.D713E Chr.3: 160268497-160268497 on Build GRCh38
6409512 c.2125A>G p.K709E Chr.3: 160268511-160268511 on Build GRCh38
4843147 c.2188G>A p.E730K Chr.3: 160268448-160268448 on Build GRCh38
6420035 c.2113G>T p.A705S Chr.3: 160268523-160268523 on Build GRCh38
2155317 c.2100G>A p.R700R Chr.3: 160268536-160268536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764764460 Chr.3: 160268481-160268481 on Build GRCh38
rs147761181 Chr.3: 160268584-160268584 on Build GRCh38
rs751917876 Chr.3: 160268494-160268494 on Build GRCh38
rs760611602 Chr.3: 160268448-160268448 on Build GRCh38
rs1039272217 Chr.3: 160268409-160268409 on Build GRCh38
rs564220393 Chr.3: 160268249-160268249 on Build GRCh38
rs746088343 Chr.3: 160268557-160268557 on Build GRCh38
rs370294182 Chr.3: 160268487-160268487 on Build GRCh38
rs886058129 Chr.3: 160268431-160268431 on Build GRCh38
rs1026272699 Chr.3: 160268229-160268229 on Build GRCh38
rs552042466 Chr.3: 160268147-160268147 on Build GRCh38
rs115849906 Chr.3: 160268393-160268393 on Build GRCh38
rs766079000 Chr.3: 160268490-160268490 on Build GRCh38
rs754140449 Chr.3: 160268551-160268551 on Build GRCh38
rs1003398267 Chr.3: 160268347-160268347 on Build GRCh38
rs770228492 Chr.3: 160268173-160268173 on Build GRCh38
rs553145696 Chr.3: 160268171-160268171 on Build GRCh38
rs777972752 Chr.3: 160268533-160268533 on Build GRCh38
rs1051491053 Chr.3: 160268133-160268133 on Build GRCh38
rs769856006 Chr.3: 160268398-160268398 on Build GRCh38
rs754746882 Chr.3: 160268504-160268505 on Build GRCh38
rs749975436 Chr.3: 160268485-160268485 on Build GRCh38
rs943036641 Chr.3: 160268156-160268156 on Build GRCh38
rs751433239 Chr.3: 160268406-160268409 on Build GRCh38
rs780364698 Chr.3: 160268374-160268374 on Build GRCh38
rs774644990 Chr.3: 160268540-160268540 on Build GRCh38
rs931581142 Chr.3: 160268550-160268550 on Build GRCh38
rs551983792 Chr.3: 160268480-160268480 on Build GRCh38
rs898758043 Chr.3: 160268204-160268204 on Build GRCh38
rs201820395 Chr.3: 160268478-160268478 on Build GRCh38
rs530781513 Chr.3: 160268138-160268138 on Build GRCh38
rs563445545 Chr.3: 160268244-160268244 on Build GRCh38
rs755428490 Chr.3: 160268383-160268383 on Build GRCh38
rs16831157 Chr.3: 160268497-160268497 on Build GRCh38
rs919051681 Chr.3: 160268297-160268297 on Build GRCh38
rs1008457044 Chr.3: 160268123-160268123 on Build GRCh38
rs763798508 Chr.3: 160268460-160268460 on Build GRCh38
rs758076802 Chr.3: 160268362-160268362 on Build GRCh38
rs137853116 Chr.3: 160268535-160268535 on Build GRCh38
rs995766910 Chr.3: 160268225-160268225 on Build GRCh38
rs969072753 Chr.3: 160268438-160268438 on Build GRCh38
rs372855267 Chr.3: 160268377-160268377 on Build GRCh38
rs977361551 Chr.3: 160268457-160268457 on Build GRCh38
rs372711710 Chr.3: 160268406-160268406 on Build GRCh38
rs781425551 Chr.3: 160268504-160268504 on Build GRCh38
rs992279246 Chr.3: 160268283-160268283 on Build GRCh38
rs112382107 Chr.3: 160268336-160268336 on Build GRCh38
rs933102548 Chr.3: 160268105-160268105 on Build GRCh38
rs767350088 Chr.3: 160268530-160268531 on Build GRCh38
rs888841443 Chr.3: 160268134-160268134 on Build GRCh38
rs1037333271 Chr.3: 160268172-160268172 on Build GRCh38
rs116072970 Chr.3: 160268360-160268360 on Build GRCh38
rs901815416 Chr.3: 160268427-160268427 on Build GRCh38
rs535067699 Chr.3: 160268423-160268423 on Build GRCh38
rs375941259 Chr.3: 160268436-160268436 on Build GRCh38
rs997566753 Chr.3: 160268462-160268462 on Build GRCh38
rs1049253652 Chr.3: 160268372-160268372 on Build GRCh38
rs532099377 Chr.3: 160268271-160268271 on Build GRCh38
rs748643921 Chr.3: 160268529-160268529 on Build GRCh38
rs980650284 Chr.3: 160268503-160268503 on Build GRCh38
rs756615065 Chr.3: 160268530-160268530 on Build GRCh38
rs373671362 Chr.3: 160268536-160268536 on Build GRCh38
rs776319625 Chr.3: 160268587-160268587 on Build GRCh38
rs943067730 Chr.3: 160268520-160268520 on Build GRCh38

View Full Product Details