Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160279081-160279583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT80
Entrez Gene ID
57560
Gene Name
intraflagellar transport 80
Gene Aliases
ATD2, SRTD2, WDR56
UniGene
Hs.478095
Gene Chromosome Location
Chr.3: 160256986-160399532 on Build GRCh38
Transcripts
NM_001190241.1 NM_001190242.1 NM_020800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6684099 c.1786G>A p.V596I Chr.3: 160279243-160279243 on Build GRCh38
3846635 c.1729G>A p.D577N Chr.3: 160279300-160279300 on Build GRCh38
445875 c.1707T>G p.N569K Chr.3: 160279322-160279322 on Build GRCh38
3915237 c.1736C>T p.S579F Chr.3: 160279293-160279293 on Build GRCh38
6684098 c.1811C>T p.A604V Chr.3: 160279218-160279218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036351934 Chr.3: 160279459-160279459 on Build GRCh38
rs142518115 Chr.3: 160279258-160279258 on Build GRCh38
rs1013567354 Chr.3: 160279485-160279485 on Build GRCh38
rs760832088 Chr.3: 160279414-160279414 on Build GRCh38
rs864622337 Chr.3: 160279276-160279276 on Build GRCh38
rs976753508 Chr.3: 160279321-160279321 on Build GRCh38
rs202145480 Chr.3: 160279351-160279351 on Build GRCh38
rs766857775 Chr.3: 160279260-160279260 on Build GRCh38
rs373850496 Chr.3: 160279148-160279148 on Build GRCh38
rs747841807 Chr.3: 160279344-160279344 on Build GRCh38
rs768427720 Chr.3: 160279214-160279214 on Build GRCh38
rs759541752 Chr.3: 160279264-160279264 on Build GRCh38
rs1046749084 Chr.3: 160279489-160279489 on Build GRCh38
rs773593306 Chr.3: 160279256-160279256 on Build GRCh38
rs146409420 Chr.3: 160279139-160279148 on Build GRCh38
rs186192085 Chr.3: 160279204-160279204 on Build GRCh38
rs769040891 Chr.3: 160279260-160279260 on Build GRCh38
rs753630151 Chr.3: 160279166-160279166 on Build GRCh38
rs771343257 Chr.3: 160279388-160279388 on Build GRCh38
rs533814246 Chr.3: 160279491-160279496 on Build GRCh38
rs371192509 Chr.3: 160279448-160279448 on Build GRCh38
rs776932992 Chr.3: 160279369-160279370 on Build GRCh38
rs774900986 Chr.3: 160279389-160279389 on Build GRCh38
rs371520687 Chr.3: 160279312-160279312 on Build GRCh38
rs967072083 Chr.3: 160279216-160279216 on Build GRCh38
rs764974699 Chr.3: 160279275-160279275 on Build GRCh38
rs755211694 Chr.3: 160279142-160279142 on Build GRCh38
rs1001996844 Chr.3: 160279185-160279185 on Build GRCh38
rs942101445 Chr.3: 160279457-160279457 on Build GRCh38
rs767664338 Chr.3: 160279160-160279160 on Build GRCh38
rs776493983 Chr.3: 160279218-160279218 on Build GRCh38
rs778749946 Chr.3: 160279179-160279179 on Build GRCh38
rs751538474 Chr.3: 160279314-160279314 on Build GRCh38
rs774272889 Chr.3: 160279491-160279492 on Build GRCh38
rs769406658 Chr.3: 160279370-160279371 on Build GRCh38
rs923523379 Chr.3: 160279347-160279347 on Build GRCh38
rs147387605 Chr.3: 160279268-160279268 on Build GRCh38
rs772778746 Chr.3: 160279371-160279371 on Build GRCh38
rs540494495 Chr.3: 160279203-160279203 on Build GRCh38
rs758317567 Chr.3: 160279193-160279193 on Build GRCh38
rs747849008 Chr.3: 160279370-160279370 on Build GRCh38
rs746031570 Chr.3: 160279526-160279526 on Build GRCh38
rs908547988 Chr.3: 160279376-160279376 on Build GRCh38
rs373116334 Chr.3: 160279188-160279188 on Build GRCh38
rs898550833 Chr.3: 160279524-160279524 on Build GRCh38
rs200508245 Chr.3: 160279483-160279483 on Build GRCh38
rs533163415 Chr.3: 160279274-160279274 on Build GRCh38
rs139849328 Chr.3: 160279338-160279338 on Build GRCh38
rs750185428 Chr.3: 160279291-160279291 on Build GRCh38
rs780858175 Chr.3: 160279394-160279394 on Build GRCh38
rs777732974 Chr.3: 160279170-160279170 on Build GRCh38
rs967683781 Chr.3: 160279290-160279290 on Build GRCh38
rs770104467 Chr.3: 160279237-160279237 on Build GRCh38
rs373859634 Chr.3: 160279140-160279149 on Build GRCh38
rs6778728 Chr.3: 160279273-160279273 on Build GRCh38
rs754592412 Chr.3: 160279316-160279316 on Build GRCh38
rs747788766 Chr.3: 160279235-160279235 on Build GRCh38
rs758028408 Chr.3: 160279135-160279135 on Build GRCh38
rs909341353 Chr.3: 160279433-160279433 on Build GRCh38
rs772182976 Chr.3: 160279408-160279408 on Build GRCh38
rs778019318 Chr.3: 160279363-160279363 on Build GRCh38
rs775510674 Chr.3: 160279409-160279409 on Build GRCh38
rs897835116 Chr.3: 160279460-160279460 on Build GRCh38
rs760018090 Chr.3: 160279407-160279407 on Build GRCh38
rs749606176 Chr.3: 160279379-160279379 on Build GRCh38

View Full Product Details