Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160280580-160281088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT80
Entrez Gene ID
57560
Gene Name
intraflagellar transport 80
Gene Aliases
ATD2, SRTD2, WDR56
UniGene
Hs.478095
Gene Chromosome Location
Chr.3: 160256986-160399532 on Build GRCh38
Transcripts
NM_001190241.1 NM_001190242.1 NM_020800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5760694 c.1527G>A p.V509V Chr.3: 160280804-160280804 on Build GRCh38
4763204 c.1627G>T p.D543Y Chr.3: 160280704-160280704 on Build GRCh38
6096692 c.1521A>T p.T507T Chr.3: 160280810-160280810 on Build GRCh38
479642 c.1549A>G p.T517A Chr.3: 160280782-160280782 on Build GRCh38
6582085 c.1621G>A p.D541N Chr.3: 160280710-160280710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572407989 Chr.3: 160280978-160280978 on Build GRCh38
rs924439848 Chr.3: 160280866-160280866 on Build GRCh38
rs538068198 Chr.3: 160280833-160280833 on Build GRCh38
rs1027265275 Chr.3: 160280706-160280706 on Build GRCh38
rs368030836 Chr.3: 160280632-160280632 on Build GRCh38
rs777413519 Chr.3: 160280669-160280669 on Build GRCh38
rs1016138018 Chr.3: 160280636-160280636 on Build GRCh38
rs560378935 Chr.3: 160280966-160280966 on Build GRCh38
rs762359374 Chr.3: 160280617-160280621 on Build GRCh38
rs756866740 Chr.3: 160280791-160280791 on Build GRCh38
rs550922501 Chr.3: 160280748-160280748 on Build GRCh38
rs776969647 Chr.3: 160280730-160280730 on Build GRCh38
rs149274965 Chr.3: 160280721-160280721 on Build GRCh38
rs760046151 Chr.3: 160280823-160280823 on Build GRCh38
rs574214763 Chr.3: 160280756-160280756 on Build GRCh38
rs542616153 Chr.3: 160281014-160281014 on Build GRCh38
rs768828850 Chr.3: 160280810-160280810 on Build GRCh38
rs431905497 Chr.3: 160280684-160280686 on Build GRCh38
rs1037995917 Chr.3: 160281029-160281029 on Build GRCh38
rs906880499 Chr.3: 160281003-160281003 on Build GRCh38
rs1005211350 Chr.3: 160280997-160280997 on Build GRCh38
rs538991665 Chr.3: 160280964-160280964 on Build GRCh38
rs1042643447 Chr.3: 160280839-160280839 on Build GRCh38
rs775551789 Chr.3: 160280861-160280861 on Build GRCh38
rs183316666 Chr.3: 160281028-160281028 on Build GRCh38
rs187515125 Chr.3: 160280709-160280709 on Build GRCh38
rs752150146 Chr.3: 160280758-160280758 on Build GRCh38
rs915202399 Chr.3: 160280701-160280701 on Build GRCh38
rs761918122 Chr.3: 160280743-160280743 on Build GRCh38
rs948658410 Chr.3: 160280970-160280970 on Build GRCh38
rs754934331 Chr.3: 160280637-160280637 on Build GRCh38
rs748453055 Chr.3: 160280923-160280923 on Build GRCh38
rs934545824 Chr.3: 160280910-160280910 on Build GRCh38
rs766317378 Chr.3: 160280744-160280744 on Build GRCh38
rs766262685 Chr.3: 160280628-160280628 on Build GRCh38
rs764959356 Chr.3: 160280676-160280676 on Build GRCh38
rs376003348 Chr.3: 160280786-160280786 on Build GRCh38
rs556364921 Chr.3: 160280856-160280856 on Build GRCh38
rs187862036 Chr.3: 160281030-160281030 on Build GRCh38
rs776540844 Chr.3: 160280814-160280814 on Build GRCh38
rs993750897 Chr.3: 160281033-160281033 on Build GRCh38
rs747457756 Chr.3: 160280809-160280809 on Build GRCh38
rs1026951803 Chr.3: 160281062-160281062 on Build GRCh38
rs371765876 Chr.3: 160280723-160280723 on Build GRCh38
rs1044004525 Chr.3: 160280989-160280989 on Build GRCh38
rs746175699 Chr.3: 160280805-160280805 on Build GRCh38
rs748884261 Chr.3: 160280673-160280673 on Build GRCh38
rs758912357 Chr.3: 160280806-160280806 on Build GRCh38
rs572926587 Chr.3: 160280771-160280771 on Build GRCh38
rs748431748 Chr.3: 160280817-160280817 on Build GRCh38
rs752199749 Chr.3: 160280652-160280652 on Build GRCh38
rs965421270 Chr.3: 160280844-160280844 on Build GRCh38
rs765860493 Chr.3: 160280619-160280620 on Build GRCh38
rs192817903 Chr.3: 160280959-160280959 on Build GRCh38
rs755731425 Chr.3: 160280659-160280659 on Build GRCh38
rs779221410 Chr.3: 160280682-160280682 on Build GRCh38
rs1049473425 Chr.3: 160280924-160280924 on Build GRCh38
rs375597309 Chr.3: 160280617-160280617 on Build GRCh38
rs535870220 Chr.3: 160280704-160280704 on Build GRCh38
rs759566776 Chr.3: 160280751-160280751 on Build GRCh38
rs746408528 Chr.3: 160280703-160280703 on Build GRCh38
rs762873799 Chr.3: 160280620-160280620 on Build GRCh38
rs774299281 Chr.3: 160280852-160280852 on Build GRCh38
rs914519232 Chr.3: 160280927-160280927 on Build GRCh38
rs371952748 Chr.3: 160280651-160280651 on Build GRCh38
rs770091105 Chr.3: 160280849-160280849 on Build GRCh38
rs779274656 Chr.3: 160280796-160280796 on Build GRCh38

View Full Product Details