Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194397413-194397912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GP5
Entrez Gene ID
2814
Gene Name
glycoprotein V platelet
Gene Aliases
CD42d, GPV
UniGene
Hs.73734
Gene Chromosome Location
Chr.3: 194394821-194399266 on Build GRCh38
Transcripts
NM_004488.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1421762 c.714C>T p.F238F Chr.3: 194397569-194397569 on Build GRCh38
1421766 c.529T>C p.L177L Chr.3: 194397754-194397754 on Build GRCh38
729894 c.536G>A p.G179E Chr.3: 194397747-194397747 on Build GRCh38
1253416 c.654C>T p.G218G Chr.3: 194397629-194397629 on Build GRCh38
446203 c.585G>A p.K195K Chr.3: 194397698-194397698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777081348 Chr.3: 194397871-194397871 on Build GRCh38
rs113052158 Chr.3: 194397481-194397481 on Build GRCh38
rs773407579 Chr.3: 194397695-194397695 on Build GRCh38
rs760537446 Chr.3: 194397555-194397555 on Build GRCh38
rs768484317 Chr.3: 194397687-194397687 on Build GRCh38
rs1043740329 Chr.3: 194397435-194397435 on Build GRCh38
rs897065551 Chr.3: 194397885-194397885 on Build GRCh38
rs752054881 Chr.3: 194397614-194397614 on Build GRCh38
rs200310605 Chr.3: 194397477-194397477 on Build GRCh38
rs770606358 Chr.3: 194397486-194397486 on Build GRCh38
rs139517746 Chr.3: 194397434-194397434 on Build GRCh38
rs375963952 Chr.3: 194397503-194397503 on Build GRCh38
rs531464548 Chr.3: 194397840-194397840 on Build GRCh38
rs979816227 Chr.3: 194397709-194397709 on Build GRCh38
rs941308233 Chr.3: 194397854-194397854 on Build GRCh38
rs749575320 Chr.3: 194397696-194397696 on Build GRCh38
rs767870641 Chr.3: 194397735-194397735 on Build GRCh38
rs757961631 Chr.3: 194397806-194397806 on Build GRCh38
rs745390651 Chr.3: 194397664-194397664 on Build GRCh38
rs745851802 Chr.3: 194397492-194397492 on Build GRCh38
rs148797033 Chr.3: 194397628-194397628 on Build GRCh38
rs771091355 Chr.3: 194397835-194397835 on Build GRCh38
rs147926792 Chr.3: 194397841-194397841 on Build GRCh38
rs759111164 Chr.3: 194397840-194397841 on Build GRCh38
rs71316299 Chr.3: 194397836-194397836 on Build GRCh38
rs7631261 Chr.3: 194397641-194397641 on Build GRCh38
rs935357800 Chr.3: 194397768-194397768 on Build GRCh38
rs773982652 Chr.3: 194397803-194397803 on Build GRCh38
rs769709278 Chr.3: 194397465-194397466 on Build GRCh38
rs779830095 Chr.3: 194397809-194397809 on Build GRCh38
rs745618726 Chr.3: 194397461-194397461 on Build GRCh38
rs878894021 Chr.3: 194397675-194397675 on Build GRCh38
rs756816416 Chr.3: 194397650-194397650 on Build GRCh38
rs752108067 Chr.3: 194397452-194397452 on Build GRCh38
rs749857036 Chr.3: 194397800-194397800 on Build GRCh38
rs1045271957 Chr.3: 194397826-194397826 on Build GRCh38
rs776467393 Chr.3: 194397572-194397572 on Build GRCh38
rs753107434 Chr.3: 194397472-194397472 on Build GRCh38
rs866648305 Chr.3: 194397682-194397682 on Build GRCh38
rs202163032 Chr.3: 194397577-194397577 on Build GRCh38
rs562312596 Chr.3: 194397500-194397500 on Build GRCh38
rs766501150 Chr.3: 194397601-194397601 on Build GRCh38
rs765313464 Chr.3: 194397680-194397680 on Build GRCh38
rs777537024 Chr.3: 194397645-194397645 on Build GRCh38
rs964172806 Chr.3: 194397678-194397678 on Build GRCh38
rs1035218886 Chr.3: 194397512-194397512 on Build GRCh38
rs756533140 Chr.3: 194397629-194397629 on Build GRCh38
rs371868475 Chr.3: 194397796-194397796 on Build GRCh38
rs754760971 Chr.3: 194397819-194397819 on Build GRCh38
rs780066557 Chr.3: 194397670-194397670 on Build GRCh38
rs759683798 Chr.3: 194397729-194397729 on Build GRCh38
rs267599745 Chr.3: 194397679-194397679 on Build GRCh38
rs774236332 Chr.3: 194397723-194397723 on Build GRCh38
rs764613759 Chr.3: 194397743-194397743 on Build GRCh38
rs199511845 Chr.3: 194397573-194397573 on Build GRCh38
rs145481862 Chr.3: 194397564-194397564 on Build GRCh38
rs1037886809 Chr.3: 194397868-194397868 on Build GRCh38
rs747021716 Chr.3: 194397816-194397816 on Build GRCh38
rs768062632 Chr.3: 194397787-194397787 on Build GRCh38
rs777768347 Chr.3: 194397526-194397527 on Build GRCh38
rs201516422 Chr.3: 194397702-194397702 on Build GRCh38
rs747949677 Chr.3: 194397832-194397832 on Build GRCh38
rs770908656 Chr.3: 194397554-194397555 on Build GRCh38
rs758214797 Chr.3: 194397669-194397669 on Build GRCh38
rs775083744 Chr.3: 194397501-194397501 on Build GRCh38
rs780895842 Chr.3: 194397825-194397825 on Build GRCh38
rs761333814 Chr.3: 194397851-194397851 on Build GRCh38
rs752760235 Chr.3: 194397793-194397793 on Build GRCh38
rs753538809 Chr.3: 194397751-194397751 on Build GRCh38
rs372044750 Chr.3: 194397470-194397470 on Build GRCh38
rs756906495 Chr.3: 194397753-194397753 on Build GRCh38
rs866113497 Chr.3: 194397565-194397565 on Build GRCh38
rs993417033 Chr.3: 194397604-194397604 on Build GRCh38
rs775848940 Chr.3: 194397842-194397842 on Build GRCh38
rs749179860 Chr.3: 194397527-194397528 on Build GRCh38
rs778974879 Chr.3: 194397491-194397491 on Build GRCh38
rs904951541 Chr.3: 194397560-194397560 on Build GRCh38
rs777590156 Chr.3: 194397478-194397478 on Build GRCh38
rs761098935 Chr.3: 194397739-194397739 on Build GRCh38
rs925498188 Chr.3: 194397453-194397453 on Build GRCh38
rs762994127 Chr.3: 194397595-194397595 on Build GRCh38
rs552136303 Chr.3: 194397740-194397740 on Build GRCh38
rs753596185 Chr.3: 194397648-194397648 on Build GRCh38
rs200544767 Chr.3: 194397704-194397704 on Build GRCh38
rs896642230 Chr.3: 194397488-194397488 on Build GRCh38
rs776105867 Chr.3: 194397737-194397737 on Build GRCh38
rs564790004 Chr.3: 194397468-194397468 on Build GRCh38
rs932705672 Chr.3: 194397783-194397783 on Build GRCh38
rs764705809 Chr.3: 194397866-194397866 on Build GRCh38
rs764998085 Chr.3: 194397764-194397764 on Build GRCh38
rs778635870 Chr.3: 194397663-194397663 on Build GRCh38
rs949499114 Chr.3: 194397821-194397821 on Build GRCh38
rs1044799160 Chr.3: 194397524-194397524 on Build GRCh38
rs146555099 Chr.3: 194397608-194397608 on Build GRCh38
rs1021936902 Chr.3: 194397568-194397568 on Build GRCh38
rs764761485 Chr.3: 194397887-194397887 on Build GRCh38
rs773697033 Chr.3: 194397808-194397808 on Build GRCh38
rs373097680 Chr.3: 194397621-194397621 on Build GRCh38
rs369093144 Chr.3: 194397592-194397592 on Build GRCh38
rs73889857 Chr.3: 194397884-194397884 on Build GRCh38

View Full Product Details