Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55082398-55082922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDR
Entrez Gene ID
3791
Gene Name
kinase insert domain receptor
Gene Aliases
CD309, FLK1, VEGFR, VEGFR2
UniGene
Hs.479756
Gene Chromosome Location
Chr.4: 55078259-55125595 on Build GRCh38
Transcripts
NM_002253.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4135812 c.3690G>C p.K1230N Chr.4: 55082608-55082608 on Build GRCh38
365817 c.3700G>T p.V1234L Chr.4: 55082598-55082598 on Build GRCh38
6925141 c.3678C>A p.N1226K Chr.4: 55082620-55082620 on Build GRCh38
3917933 c.3692G>A p.S1231N Chr.4: 55082606-55082606 on Build GRCh38
6954233 c.3760G>C p.D1254H Chr.4: 55082538-55082538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985560183 Chr.4: 55082726-55082726 on Build GRCh38
rs923597399 Chr.4: 55082846-55082846 on Build GRCh38
rs76817373 Chr.4: 55082818-55082818 on Build GRCh38
rs763129470 Chr.4: 55082887-55082887 on Build GRCh38
rs746789315 Chr.4: 55082672-55082672 on Build GRCh38
rs948272810 Chr.4: 55082652-55082652 on Build GRCh38
rs757443823 Chr.4: 55082568-55082568 on Build GRCh38
rs912777456 Chr.4: 55082422-55082422 on Build GRCh38
rs967964674 Chr.4: 55082834-55082834 on Build GRCh38
rs778078301 Chr.4: 55082559-55082559 on Build GRCh38
rs201866429 Chr.4: 55082682-55082682 on Build GRCh38
rs998001986 Chr.4: 55082738-55082738 on Build GRCh38
rs770746793 Chr.4: 55082681-55082681 on Build GRCh38
rs1029908959 Chr.4: 55082460-55082460 on Build GRCh38
rs374982964 Chr.4: 55082527-55082527 on Build GRCh38
rs200648845 Chr.4: 55082638-55082638 on Build GRCh38
rs142120845 Chr.4: 55082541-55082541 on Build GRCh38
rs772902095 Chr.4: 55082599-55082599 on Build GRCh38
rs372769194 Chr.4: 55082665-55082665 on Build GRCh38
rs41520946 Chr.4: 55082426-55082426 on Build GRCh38
rs377109088 Chr.4: 55082594-55082594 on Build GRCh38
rs751902659 Chr.4: 55082620-55082620 on Build GRCh38
rs200085996 Chr.4: 55082657-55082657 on Build GRCh38
rs901892302 Chr.4: 55082735-55082735 on Build GRCh38
rs757575555 Chr.4: 55082622-55082622 on Build GRCh38
rs111307973 Chr.4: 55082543-55082543 on Build GRCh38
rs1029107559 Chr.4: 55082752-55082752 on Build GRCh38
rs368072993 Chr.4: 55082799-55082799 on Build GRCh38
rs191496526 Chr.4: 55082714-55082714 on Build GRCh38
rs1022191336 Chr.4: 55082791-55082791 on Build GRCh38
rs150847930 Chr.4: 55082570-55082570 on Build GRCh38
rs78801899 Chr.4: 55082528-55082528 on Build GRCh38
rs553564973 Chr.4: 55082687-55082687 on Build GRCh38
rs568921825 Chr.4: 55082577-55082577 on Build GRCh38
rs550836649 Chr.4: 55082437-55082437 on Build GRCh38
rs760219983 Chr.4: 55082506-55082506 on Build GRCh38
rs1015992221 Chr.4: 55082685-55082685 on Build GRCh38
rs773471186 Chr.4: 55082716-55082716 on Build GRCh38
rs763426023 Chr.4: 55082613-55082613 on Build GRCh38
rs912747671 Chr.4: 55082757-55082757 on Build GRCh38
rs1053451779 Chr.4: 55082895-55082895 on Build GRCh38
rs765747186 Chr.4: 55082538-55082538 on Build GRCh38
rs759104314 Chr.4: 55082608-55082608 on Build GRCh38
rs894977242 Chr.4: 55082542-55082542 on Build GRCh38
rs764614608 Chr.4: 55082614-55082614 on Build GRCh38
rs1027068452 Chr.4: 55082689-55082689 on Build GRCh38
rs561517653 Chr.4: 55082443-55082443 on Build GRCh38
rs1006742492 Chr.4: 55082774-55082774 on Build GRCh38
rs368229220 Chr.4: 55082603-55082603 on Build GRCh38
rs571774697 Chr.4: 55082873-55082873 on Build GRCh38
rs751858962 Chr.4: 55082563-55082563 on Build GRCh38
rs547194966 Chr.4: 55082497-55082497 on Build GRCh38
rs748096654 Chr.4: 55082595-55082595 on Build GRCh38
rs756332251 Chr.4: 55082635-55082635 on Build GRCh38
rs745420505 Chr.4: 55082521-55082521 on Build GRCh38
rs1004806880 Chr.4: 55082698-55082698 on Build GRCh38
rs767647176 Chr.4: 55082625-55082625 on Build GRCh38
rs772987575 Chr.4: 55082505-55082505 on Build GRCh38
rs960435281 Chr.4: 55082897-55082897 on Build GRCh38
rs781436645 Chr.4: 55082569-55082569 on Build GRCh38
rs564932535 Chr.4: 55082894-55082894 on Build GRCh38
rs974557653 Chr.4: 55082688-55082688 on Build GRCh38
rs750648058 Chr.4: 55082628-55082628 on Build GRCh38
rs185368292 Chr.4: 55082604-55082604 on Build GRCh38
rs150116423 Chr.4: 55082547-55082547 on Build GRCh38
rs530260334 Chr.4: 55082486-55082486 on Build GRCh38
rs977711852 Chr.4: 55082836-55082836 on Build GRCh38
rs180730202 Chr.4: 55082773-55082773 on Build GRCh38
rs978394255 Chr.4: 55082884-55082884 on Build GRCh38
rs902199058 Chr.4: 55082511-55082511 on Build GRCh38
rs776015468 Chr.4: 55082612-55082612 on Build GRCh38
rs764721295 Chr.4: 55082611-55082611 on Build GRCh38
rs906414824 Chr.4: 55082668-55082668 on Build GRCh38
rs772035220 Chr.4: 55082596-55082596 on Build GRCh38

View Full Product Details