Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87337133-87337639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B11
Entrez Gene ID
51170
Gene Name
hydroxysteroid 17-beta dehydrogenase 11
Gene Aliases
17-BETA-HSD11, 17-BETA-HSDXI, 17BHSD11, DHRS8, PAN1B, RETSDR2, SDR16C2
UniGene
Hs.594923
Gene Chromosome Location
Chr.4: 87336522-87391303 on Build GRCh38
Transcripts
NM_016245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3918303 c.816C>T p.I272I Chr.4: 87337363-87337363 on Build GRCh38
1058602 c.827G>A p.R276H Chr.4: 87337352-87337352 on Build GRCh38
4126570 c.826C>T p.R276C Chr.4: 87337353-87337353 on Build GRCh38
7227220 c.852A>G p.K284K Chr.4: 87337327-87337327 on Build GRCh38
7315964 c.831C>G p.F277L Chr.4: 87337348-87337348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762262793 Chr.4: 87337282-87337282 on Build GRCh38
rs760806729 Chr.4: 87337379-87337379 on Build GRCh38
rs773226306 Chr.4: 87337377-87337377 on Build GRCh38
rs566810767 Chr.4: 87337602-87337602 on Build GRCh38
rs780868840 Chr.4: 87337348-87337348 on Build GRCh38
rs878986772 Chr.4: 87337186-87337187 on Build GRCh38
rs933087246 Chr.4: 87337444-87337444 on Build GRCh38
rs774820994 Chr.4: 87337273-87337273 on Build GRCh38
rs754786631 Chr.4: 87337315-87337315 on Build GRCh38
rs777162512 Chr.4: 87337332-87337332 on Build GRCh38
rs775777322 Chr.4: 87337248-87337248 on Build GRCh38
rs9991501 Chr.4: 87337331-87337331 on Build GRCh38
rs367692388 Chr.4: 87337228-87337228 on Build GRCh38
rs753640740 Chr.4: 87337301-87337301 on Build GRCh38
rs539591358 Chr.4: 87337575-87337575 on Build GRCh38
rs545124516 Chr.4: 87337352-87337352 on Build GRCh38
rs756831968 Chr.4: 87337323-87337323 on Build GRCh38
rs141325794 Chr.4: 87337397-87337397 on Build GRCh38
rs960084275 Chr.4: 87337306-87337306 on Build GRCh38
rs1027716634 Chr.4: 87337448-87337448 on Build GRCh38
rs993639057 Chr.4: 87337588-87337588 on Build GRCh38
rs774653796 Chr.4: 87337359-87337359 on Build GRCh38
rs568901053 Chr.4: 87337548-87337548 on Build GRCh38
rs562443434 Chr.4: 87337283-87337283 on Build GRCh38
rs766608143 Chr.4: 87337300-87337300 on Build GRCh38
rs766552535 Chr.4: 87337381-87337381 on Build GRCh38
rs73841140 Chr.4: 87337460-87337460 on Build GRCh38
rs987952024 Chr.4: 87337399-87337399 on Build GRCh38
rs776891793 Chr.4: 87337383-87337383 on Build GRCh38
rs58693841 Chr.4: 87337499-87337499 on Build GRCh38
rs200055864 Chr.4: 87337229-87337229 on Build GRCh38
rs1016507620 Chr.4: 87337217-87337217 on Build GRCh38
rs376354024 Chr.4: 87337297-87337297 on Build GRCh38
rs1038359777 Chr.4: 87337558-87337558 on Build GRCh38
rs768539748 Chr.4: 87337591-87337591 on Build GRCh38
rs148319480 Chr.4: 87337585-87337585 on Build GRCh38
rs976381600 Chr.4: 87337511-87337511 on Build GRCh38
rs368240246 Chr.4: 87337344-87337344 on Build GRCh38
rs941668292 Chr.4: 87337539-87337539 on Build GRCh38
rs148948833 Chr.4: 87337353-87337353 on Build GRCh38
rs895969623 Chr.4: 87337594-87337594 on Build GRCh38
rs749662343 Chr.4: 87337253-87337253 on Build GRCh38
rs993612177 Chr.4: 87337410-87337410 on Build GRCh38
rs567573491 Chr.4: 87337482-87337482 on Build GRCh38
rs887862102 Chr.4: 87337455-87337455 on Build GRCh38
rs878865099 Chr.4: 87337200-87337200 on Build GRCh38
rs923031091 Chr.4: 87337428-87337428 on Build GRCh38
rs975950718 Chr.4: 87337223-87337223 on Build GRCh38
rs752703504 Chr.4: 87337323-87337323 on Build GRCh38
rs372106029 Chr.4: 87337358-87337358 on Build GRCh38
rs140758997 Chr.4: 87337477-87337478 on Build GRCh38
rs888776566 Chr.4: 87337189-87337189 on Build GRCh38
rs748654789 Chr.4: 87337363-87337363 on Build GRCh38
rs544215776 Chr.4: 87337254-87337254 on Build GRCh38
rs531485022 Chr.4: 87337489-87337489 on Build GRCh38
rs772514784 Chr.4: 87337369-87337369 on Build GRCh38
rs111964422 Chr.4: 87337194-87337194 on Build GRCh38
rs1017526794 Chr.4: 87337470-87337470 on Build GRCh38
rs746516378 Chr.4: 87337338-87337338 on Build GRCh38
rs765313087 Chr.4: 87337322-87337322 on Build GRCh38
rs770560741 Chr.4: 87337231-87337231 on Build GRCh38
rs765155613 Chr.4: 87337400-87337400 on Build GRCh38
rs143569767 Chr.4: 87337433-87337433 on Build GRCh38

View Full Product Details