Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88592400-88592887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC3
Entrez Gene ID
8916
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 3
Gene Aliases
-
UniGene
Hs.35804
Gene Chromosome Location
Chr.4: 88592423-88709303 on Build GRCh38
Transcripts
NM_001271602.1 NM_001318505.1 NM_014606.2 XM_005263327.3 XM_005263329.3 XM_005263330.3 XM_005263331.1 XM_005263332.3 XM_017008806.1 XM_017008807.1

SNP Detail

SNP ID Chromosome Location
rs185525379 Chr.4: 88592606-88592606 on Build GRCh38
rs1010196762 Chr.4: 88592653-88592653 on Build GRCh38
rs757716629 Chr.4: 88592828-88592828 on Build GRCh38
rs1050984602 Chr.4: 88592585-88592585 on Build GRCh38
rs570341552 Chr.4: 88592783-88592783 on Build GRCh38
rs757972667 Chr.4: 88592454-88592454 on Build GRCh38
rs1053885873 Chr.4: 88592792-88592792 on Build GRCh38
rs989912464 Chr.4: 88592486-88592486 on Build GRCh38
rs1032463558 Chr.4: 88592626-88592626 on Build GRCh38
rs981691943 Chr.4: 88592504-88592504 on Build GRCh38
rs752318794 Chr.4: 88592452-88592452 on Build GRCh38
rs1050908685 Chr.4: 88592749-88592749 on Build GRCh38
rs1009837094 Chr.4: 88592817-88592817 on Build GRCh38
rs934611981 Chr.4: 88592758-88592758 on Build GRCh38
rs917161149 Chr.4: 88592778-88592778 on Build GRCh38
rs1057405929 Chr.4: 88592774-88592774 on Build GRCh38
rs941872940 Chr.4: 88592513-88592513 on Build GRCh38
rs1039110751 Chr.4: 88592540-88592540 on Build GRCh38
rs933425214 Chr.4: 88592493-88592493 on Build GRCh38
rs548571116 Chr.4: 88592780-88592780 on Build GRCh38
rs919052727 Chr.4: 88592746-88592746 on Build GRCh38
rs954847201 Chr.4: 88592715-88592715 on Build GRCh38
rs530320102 Chr.4: 88592608-88592608 on Build GRCh38
rs1023823275 Chr.4: 88592428-88592428 on Build GRCh38
rs998454131 Chr.4: 88592466-88592466 on Build GRCh38
rs764817700 Chr.4: 88592439-88592439 on Build GRCh38
rs552565874 Chr.4: 88592794-88592794 on Build GRCh38
rs1023147132 Chr.4: 88592785-88592785 on Build GRCh38
rs374690057 Chr.4: 88592813-88592813 on Build GRCh38
rs759214927 Chr.4: 88592427-88592427 on Build GRCh38
rs981577352 Chr.4: 88592654-88592654 on Build GRCh38
rs929256419 Chr.4: 88592756-88592756 on Build GRCh38
rs113601495 Chr.4: 88592664-88592664 on Build GRCh38
rs986135922 Chr.4: 88592721-88592721 on Build GRCh38
rs1047792092 Chr.4: 88592512-88592512 on Build GRCh38
rs897786790 Chr.4: 88592573-88592573 on Build GRCh38
rs973268030 Chr.4: 88592740-88592740 on Build GRCh38
rs1031030782 Chr.4: 88592819-88592819 on Build GRCh38
rs923232199 Chr.4: 88592492-88592492 on Build GRCh38
rs963454467 Chr.4: 88592725-88592725 on Build GRCh38
rs534833231 Chr.4: 88592821-88592821 on Build GRCh38
rs1017611167 Chr.4: 88592866-88592866 on Build GRCh38
rs910400234 Chr.4: 88592505-88592505 on Build GRCh38
rs942150114 Chr.4: 88592538-88592538 on Build GRCh38
rs370705374 Chr.4: 88592695-88592695 on Build GRCh38
rs942009108 Chr.4: 88592710-88592710 on Build GRCh38
rs553062013 Chr.4: 88592838-88592838 on Build GRCh38
rs1007605481 Chr.4: 88592847-88592847 on Build GRCh38
rs970646639 Chr.4: 88592647-88592647 on Build GRCh38
rs564154301 Chr.4: 88592470-88592470 on Build GRCh38
rs560541771 Chr.4: 88592730-88592730 on Build GRCh38
rs979554721 Chr.4: 88592444-88592444 on Build GRCh38
rs541488408 Chr.4: 88592650-88592650 on Build GRCh38
rs950970832 Chr.4: 88592665-88592665 on Build GRCh38
rs1038636687 Chr.4: 88592430-88592430 on Build GRCh38
rs751454110 Chr.4: 88592580-88592580 on Build GRCh38
rs887832076 Chr.4: 88592423-88592423 on Build GRCh38
rs866113627 Chr.4: 88592474-88592474 on Build GRCh38
rs983702485 Chr.4: 88592680-88592680 on Build GRCh38
rs1037522189 Chr.4: 88592514-88592514 on Build GRCh38
rs1016478395 Chr.4: 88592829-88592829 on Build GRCh38
rs548000045 Chr.4: 88592603-88592603 on Build GRCh38
rs929302580 Chr.4: 88592511-88592511 on Build GRCh38
rs957372390 Chr.4: 88592476-88592476 on Build GRCh38
rs986599959 Chr.4: 88592503-88592503 on Build GRCh38
rs909198745 Chr.4: 88592523-88592523 on Build GRCh38
rs777830870 Chr.4: 88592565-88592565 on Build GRCh38
rs188786017 Chr.4: 88592787-88592787 on Build GRCh38
rs928777244 Chr.4: 88592658-88592658 on Build GRCh38
rs1031284204 Chr.4: 88592475-88592475 on Build GRCh38
rs1001026265 Chr.4: 88592618-88592618 on Build GRCh38
rs1035901351 Chr.4: 88592814-88592814 on Build GRCh38
rs757302900 Chr.4: 88592712-88592712 on Build GRCh38
rs909240863 Chr.4: 88592703-88592703 on Build GRCh38
rs988195161 Chr.4: 88592480-88592480 on Build GRCh38
rs868159760 Chr.4: 88592607-88592607 on Build GRCh38
rs528242895 Chr.4: 88592600-88592600 on Build GRCh38
rs910663868 Chr.4: 88592722-88592722 on Build GRCh38
rs948411045 Chr.4: 88592779-88592779 on Build GRCh38
rs752083207 Chr.4: 88592724-88592724 on Build GRCh38

View Full Product Details