Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99948719-99949216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFZ
Entrez Gene ID
3015
Gene Name
H2A histone family member Z
Gene Aliases
H2A.Z-1, H2A.z, H2A/z, H2AZ
UniGene
Hs.119192
Gene Chromosome Location
Chr.4: 99948087-99950355 on Build GRCh38
Transcripts
NM_002106.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687955 c.202G>C p.E68Q Chr.4: 99948934-99948934 on Build GRCh38
5487542 c.230_231insGGGGTC p.L77_K78insGS Chr.4: 99948905-99948906 on Build GRCh38
1049484 c.217G>A p.A73T Chr.4: 99948919-99948919 on Build GRCh38
1049483 c.314T>G p.I105S Chr.4: 99948822-99948822 on Build GRCh38
4972649 c.264A>G p.Q88Q Chr.4: 99948872-99948872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750928017 Chr.4: 99948942-99948943 on Build GRCh38
rs143698630 Chr.4: 99948818-99948818 on Build GRCh38
rs898288232 Chr.4: 99949080-99949080 on Build GRCh38
rs966753522 Chr.4: 99948951-99948951 on Build GRCh38
rs925394484 Chr.4: 99948969-99948969 on Build GRCh38
rs1013897392 Chr.4: 99949157-99949157 on Build GRCh38
rs760924394 Chr.4: 99948875-99948875 on Build GRCh38
rs61203457 Chr.4: 99949025-99949025 on Build GRCh38
rs375051093 Chr.4: 99948935-99948935 on Build GRCh38
rs372912583 Chr.4: 99948976-99948976 on Build GRCh38
rs898648588 Chr.4: 99949066-99949066 on Build GRCh38
rs376782735 Chr.4: 99948780-99948780 on Build GRCh38
rs186120127 Chr.4: 99949168-99949168 on Build GRCh38
rs986757615 Chr.4: 99949166-99949166 on Build GRCh38
rs758223843 Chr.4: 99948966-99948966 on Build GRCh38
rs1045970701 Chr.4: 99949054-99949054 on Build GRCh38
rs933586979 Chr.4: 99949024-99949024 on Build GRCh38
rs776146590 Chr.4: 99948782-99948782 on Build GRCh38
rs752531577 Chr.4: 99948956-99948956 on Build GRCh38
rs906087762 Chr.4: 99949057-99949057 on Build GRCh38
rs959314900 Chr.4: 99948779-99948779 on Build GRCh38
rs1013525467 Chr.4: 99948791-99948791 on Build GRCh38
rs765712066 Chr.4: 99948938-99948939 on Build GRCh38
rs939708556 Chr.4: 99949071-99949072 on Build GRCh38
rs1024904709 Chr.4: 99949158-99949158 on Build GRCh38
rs565394949 Chr.4: 99949148-99949148 on Build GRCh38
rs975051796 Chr.4: 99949179-99949179 on Build GRCh38
rs770368210 Chr.4: 99948781-99948781 on Build GRCh38
rs11554386 Chr.4: 99948886-99948886 on Build GRCh38
rs925832673 Chr.4: 99949184-99949184 on Build GRCh38
rs746676079 Chr.4: 99948982-99948982 on Build GRCh38
rs762968422 Chr.4: 99949023-99949023 on Build GRCh38
rs879232278 Chr.4: 99949077-99949077 on Build GRCh38
rs569763623 Chr.4: 99949114-99949114 on Build GRCh38
rs964691313 Chr.4: 99949174-99949174 on Build GRCh38
rs114299525 Chr.4: 99949013-99949013 on Build GRCh38
rs761394746 Chr.4: 99948960-99948960 on Build GRCh38
rs750703840 Chr.4: 99948872-99948872 on Build GRCh38
rs1017138792 Chr.4: 99949059-99949059 on Build GRCh38
rs11554394 Chr.4: 99948898-99948898 on Build GRCh38
rs527891782 Chr.4: 99949152-99949153 on Build GRCh38
rs774636266 Chr.4: 99948799-99948799 on Build GRCh38
rs766374566 Chr.4: 99948892-99948892 on Build GRCh38
rs545675261 Chr.4: 99948773-99948773 on Build GRCh38
rs369350507 Chr.4: 99948812-99948812 on Build GRCh38
rs772823667 Chr.4: 99948768-99948768 on Build GRCh38
rs778892566 Chr.4: 99948955-99948955 on Build GRCh38
rs999573007 Chr.4: 99949180-99949180 on Build GRCh38
rs958191201 Chr.4: 99948978-99948978 on Build GRCh38
rs995291245 Chr.4: 99949103-99949103 on Build GRCh38
rs545426612 Chr.4: 99949027-99949028 on Build GRCh38
rs559185347 Chr.4: 99949011-99949011 on Build GRCh38
rs376045749 Chr.4: 99949076-99949076 on Build GRCh38
rs540936185 Chr.4: 99948798-99948798 on Build GRCh38
rs1002192955 Chr.4: 99949058-99949058 on Build GRCh38
rs763370339 Chr.4: 99948790-99948790 on Build GRCh38
rs753948840 Chr.4: 99948932-99948932 on Build GRCh38
rs955364625 Chr.4: 99949150-99949150 on Build GRCh38
rs1054911814 Chr.4: 99949117-99949117 on Build GRCh38

View Full Product Details