Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 153549785-153550325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0922
Entrez Gene ID
23240
Gene Name
KIAA0922
Gene Aliases
TMEM131L
UniGene
Hs.732450
Gene Chromosome Location
Chr.4: 153466346-153637608 on Build GRCh38
Transcripts
NM_001131007.1 NM_015196.3 XM_011531780.1 XM_011531781.1 XM_011531783.1 XM_017007924.1 XM_017007925.1 XM_017007926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6329008 c.240-10C>T p.? Chr.4: 153550063-153550063 on Build GRCh38
447513 c.297T>C p.D99D Chr.4: 153550130-153550130 on Build GRCh38
3825392 c.289G>T p.V97F Chr.4: 153550122-153550122 on Build GRCh38
6424974 c.307C>A p.Q103K Chr.4: 153550140-153550140 on Build GRCh38
5612175 c.255A>G p.L85L Chr.4: 153550088-153550088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763193069 Chr.4: 153550203-153550203 on Build GRCh38
rs780932081 Chr.4: 153550196-153550196 on Build GRCh38
rs1037651581 Chr.4: 153549910-153549910 on Build GRCh38
rs955670493 Chr.4: 153550051-153550051 on Build GRCh38
rs992564773 Chr.4: 153549829-153549829 on Build GRCh38
rs535065838 Chr.4: 153550023-153550023 on Build GRCh38
rs575847827 Chr.4: 153550296-153550296 on Build GRCh38
rs950222639 Chr.4: 153550021-153550021 on Build GRCh38
rs574229717 Chr.4: 153550017-153550017 on Build GRCh38
rs962614295 Chr.4: 153550150-153550150 on Build GRCh38
rs1052315954 Chr.4: 153550212-153550212 on Build GRCh38
rs201330911 Chr.4: 153550091-153550091 on Build GRCh38
rs991922763 Chr.4: 153550213-153550213 on Build GRCh38
rs572802382 Chr.4: 153549990-153549990 on Build GRCh38
rs144378127 Chr.4: 153549873-153549873 on Build GRCh38
rs565812852 Chr.4: 153549955-153549955 on Build GRCh38
rs1014426634 Chr.4: 153550053-153550053 on Build GRCh38
rs1041980697 Chr.4: 153550040-153550040 on Build GRCh38
rs183881006 Chr.4: 153550216-153550216 on Build GRCh38
rs778559058 Chr.4: 153550058-153550058 on Build GRCh38
rs754624423 Chr.4: 153550115-153550115 on Build GRCh38
rs750073446 Chr.4: 153550173-153550173 on Build GRCh38
rs779705832 Chr.4: 153550118-153550118 on Build GRCh38
rs1014520378 Chr.4: 153549869-153549869 on Build GRCh38
rs778668064 Chr.4: 153550172-153550176 on Build GRCh38
rs955355030 Chr.4: 153549977-153549977 on Build GRCh38
rs766525405 Chr.4: 153550059-153550059 on Build GRCh38
rs191709903 Chr.4: 153550047-153550047 on Build GRCh38
rs563413502 Chr.4: 153550222-153550222 on Build GRCh38
rs917091573 Chr.4: 153549832-153549832 on Build GRCh38
rs376141880 Chr.4: 153549809-153549809 on Build GRCh38
rs1032966409 Chr.4: 153549940-153549940 on Build GRCh38
rs919229340 Chr.4: 153549947-153549947 on Build GRCh38
rs982738742 Chr.4: 153549982-153549982 on Build GRCh38
rs545101449 Chr.4: 153550221-153550221 on Build GRCh38
rs749145054 Chr.4: 153550044-153550044 on Build GRCh38
rs549685868 Chr.4: 153549823-153549823 on Build GRCh38
rs943558185 Chr.4: 153550233-153550233 on Build GRCh38
rs929411097 Chr.4: 153549948-153549948 on Build GRCh38
rs945137970 Chr.4: 153550239-153550239 on Build GRCh38
rs745675183 Chr.4: 153550063-153550063 on Build GRCh38
rs1056586529 Chr.4: 153550013-153550013 on Build GRCh38
rs909830500 Chr.4: 153550129-153550129 on Build GRCh38
rs537611214 Chr.4: 153549998-153549998 on Build GRCh38
rs9996529 Chr.4: 153549961-153549961 on Build GRCh38
rs531489191 Chr.4: 153549879-153549879 on Build GRCh38
rs926288732 Chr.4: 153550200-153550200 on Build GRCh38
rs982989867 Chr.4: 153550155-153550155 on Build GRCh38
rs970332632 Chr.4: 153550088-153550088 on Build GRCh38
rs753746383 Chr.4: 153550046-153550050 on Build GRCh38
rs149562943 Chr.4: 153550054-153550055 on Build GRCh38
rs547053633 Chr.4: 153549952-153549952 on Build GRCh38
rs577573259 Chr.4: 153550198-153550198 on Build GRCh38
rs937648903 Chr.4: 153550209-153550209 on Build GRCh38
rs551451276 Chr.4: 153549965-153549965 on Build GRCh38
rs909796459 Chr.4: 153549880-153549880 on Build GRCh38
rs528914212 Chr.4: 153549890-153549890 on Build GRCh38
rs570287494 Chr.4: 153549984-153549984 on Build GRCh38
rs59743932 Chr.4: 153550009-153550009 on Build GRCh38
rs747708758 Chr.4: 153550256-153550256 on Build GRCh38
rs1007719508 Chr.4: 153549986-153549986 on Build GRCh38
rs942058675 Chr.4: 153549893-153549893 on Build GRCh38
rs926275856 Chr.4: 153550002-153550002 on Build GRCh38
rs746715082 Chr.4: 153550184-153550184 on Build GRCh38

View Full Product Details