Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 175651713-175652250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPM6A
Entrez Gene ID
2823
Gene Name
glycoprotein M6A
Gene Aliases
GPM6, M6A
UniGene
Hs.75819
Gene Chromosome Location
Chr.4: 175632937-176002691 on Build GRCh38
Transcripts
NM_001261447.1 NM_001261448.1 NM_005277.4 NM_201591.2 NM_201592.2 NR_048571.1 XM_006714183.1 XM_011531877.1 XM_017008036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208561 c.541G>T p.G181* Chr.4: 175651834-175651834 on Build GRCh38
6673327 c.522C>T p.C174C Chr.4: 175651853-175651853 on Build GRCh38
6673329 c.488G>A p.R163Q Chr.4: 175651887-175651887 on Build GRCh38
6673325 c.401C>T p.T134M Chr.4: 175651941-175651941 on Build GRCh38
6038774 c.482A>T p.N161I Chr.4: 175651860-175651860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768034858 Chr.4: 175651881-175651881 on Build GRCh38
rs776065886 Chr.4: 175651949-175651949 on Build GRCh38
rs754747152 Chr.4: 175652038-175652038 on Build GRCh38
rs556443862 Chr.4: 175652152-175652152 on Build GRCh38
rs866184504 Chr.4: 175651814-175651814 on Build GRCh38
rs760954509 Chr.4: 175651951-175651951 on Build GRCh38
rs778696357 Chr.4: 175651900-175651900 on Build GRCh38
rs756731427 Chr.4: 175651896-175651896 on Build GRCh38
rs775371356 Chr.4: 175651824-175651824 on Build GRCh38
rs375692732 Chr.4: 175651974-175651974 on Build GRCh38
rs774276270 Chr.4: 175652131-175652131 on Build GRCh38
rs1040781219 Chr.4: 175652188-175652188 on Build GRCh38
rs369330000 Chr.4: 175651888-175651888 on Build GRCh38
rs761301919 Chr.4: 175651886-175651886 on Build GRCh38
rs781003179 Chr.4: 175651929-175651929 on Build GRCh38
rs750110827 Chr.4: 175651907-175651907 on Build GRCh38
rs201531480 Chr.4: 175651822-175651822 on Build GRCh38
rs943721593 Chr.4: 175652169-175652169 on Build GRCh38
rs761349389 Chr.4: 175651966-175651966 on Build GRCh38
rs768834518 Chr.4: 175651843-175651843 on Build GRCh38
rs778489165 Chr.4: 175651901-175651901 on Build GRCh38
rs940555052 Chr.4: 175651905-175651905 on Build GRCh38
rs751308429 Chr.4: 175652024-175652024 on Build GRCh38
rs746992386 Chr.4: 175651826-175651826 on Build GRCh38
rs757003932 Chr.4: 175651796-175651796 on Build GRCh38
rs72702640 Chr.4: 175652150-175652150 on Build GRCh38
rs11545192 Chr.4: 175651922-175651922 on Build GRCh38
rs771122871 Chr.4: 175651873-175651873 on Build GRCh38
rs913364698 Chr.4: 175651737-175651737 on Build GRCh38
rs749337410 Chr.4: 175651934-175651934 on Build GRCh38
rs925535411 Chr.4: 175652077-175652077 on Build GRCh38
rs1025501758 Chr.4: 175651957-175651957 on Build GRCh38
rs989001598 Chr.4: 175652109-175652109 on Build GRCh38
rs576358857 Chr.4: 175652205-175652205 on Build GRCh38
rs764604248 Chr.4: 175651960-175651960 on Build GRCh38
rs753231161 Chr.4: 175651882-175651882 on Build GRCh38
rs779875512 Chr.4: 175651915-175651915 on Build GRCh38
rs774683670 Chr.4: 175651940-175651940 on Build GRCh38
rs762944128 Chr.4: 175651872-175651872 on Build GRCh38
rs754325730 Chr.4: 175651965-175651965 on Build GRCh38
rs770946649 Chr.4: 175651938-175651938 on Build GRCh38
rs750057267 Chr.4: 175651979-175651979 on Build GRCh38
rs746939779 Chr.4: 175651918-175651918 on Build GRCh38
rs764772109 Chr.4: 175651973-175651973 on Build GRCh38
rs553518576 Chr.4: 175651887-175651887 on Build GRCh38
rs748880422 Chr.4: 175651793-175651793 on Build GRCh38
rs762721812 Chr.4: 175652055-175652055 on Build GRCh38
rs979364909 Chr.4: 175652161-175652161 on Build GRCh38
rs770642244 Chr.4: 175652057-175652057 on Build GRCh38
rs11545193 Chr.4: 175651954-175651954 on Build GRCh38
rs956374822 Chr.4: 175652104-175652104 on Build GRCh38
rs759939268 Chr.4: 175651880-175651880 on Build GRCh38
rs151245748 Chr.4: 175651908-175651908 on Build GRCh38
rs772550704 Chr.4: 175651945-175651945 on Build GRCh38
rs758610134 Chr.4: 175652107-175652108 on Build GRCh38
rs777390141 Chr.4: 175651790-175651790 on Build GRCh38
rs910862094 Chr.4: 175652159-175652159 on Build GRCh38
rs994025519 Chr.4: 175651846-175651846 on Build GRCh38
rs778434702 Chr.4: 175651809-175651809 on Build GRCh38
rs1034611795 Chr.4: 175652091-175652091 on Build GRCh38
rs773405274 Chr.4: 175651864-175651864 on Build GRCh38
rs978263178 Chr.4: 175651996-175651996 on Build GRCh38
rs758107436 Chr.4: 175651982-175651982 on Build GRCh38
rs959647938 Chr.4: 175651923-175651923 on Build GRCh38
rs371534832 Chr.4: 175651787-175651787 on Build GRCh38
rs373061459 Chr.4: 175651941-175651941 on Build GRCh38
rs745605782 Chr.4: 175651821-175651821 on Build GRCh38
rs531535079 Chr.4: 175652226-175652226 on Build GRCh38
rs72702639 Chr.4: 175651877-175651877 on Build GRCh38
rs753324121 Chr.4: 175651889-175651889 on Build GRCh38
rs779683240 Chr.4: 175652005-175652005 on Build GRCh38
rs745357551 Chr.4: 175652120-175652120 on Build GRCh38
rs768569041 Chr.4: 175651925-175651925 on Build GRCh38

View Full Product Details