Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185159573-185160081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP97
Entrez Gene ID
57587
Gene Name
cilia and flagella associated protein 97
Gene Aliases
KIAA1430, hmw
UniGene
Hs.535734
Gene Chromosome Location
Chr.4: 185159662-185210251 on Build GRCh38
Transcripts
NM_020827.2 XM_017008483.1 XM_017008484.1 XM_017008485.1 XM_017008486.1

SNP Detail

SNP ID Chromosome Location
rs1050117795 Chr.4: 185159679-185159682 on Build GRCh38
rs912455573 Chr.4: 185160026-185160026 on Build GRCh38
rs1005449573 Chr.4: 185159977-185159977 on Build GRCh38
rs1011632621 Chr.4: 185159808-185159808 on Build GRCh38
rs941489343 Chr.4: 185159922-185159922 on Build GRCh38
rs972286479 Chr.4: 185159756-185159756 on Build GRCh38
rs527625033 Chr.4: 185159682-185159682 on Build GRCh38
rs555972353 Chr.4: 185160001-185160001 on Build GRCh38
rs560320811 Chr.4: 185159815-185159815 on Build GRCh38
rs961093032 Chr.4: 185159995-185159995 on Build GRCh38
rs939781777 Chr.4: 185159616-185159616 on Build GRCh38
rs193004718 Chr.4: 185159600-185159600 on Build GRCh38
rs1015125050 Chr.4: 185159987-185159987 on Build GRCh38
rs943859891 Chr.4: 185159751-185159751 on Build GRCh38
rs768110037 Chr.4: 185159677-185159677 on Build GRCh38
rs532788257 Chr.4: 185159791-185159791 on Build GRCh38
rs189085054 Chr.4: 185160002-185160002 on Build GRCh38
rs369004814 Chr.4: 185160027-185160027 on Build GRCh38
rs542075080 Chr.4: 185159621-185159621 on Build GRCh38
rs140156146 Chr.4: 185160019-185160019 on Build GRCh38
rs973716791 Chr.4: 185159617-185159617 on Build GRCh38
rs569446501 Chr.4: 185159851-185159851 on Build GRCh38
rs546562827 Chr.4: 185159677-185159677 on Build GRCh38
rs999291463 Chr.4: 185160021-185160021 on Build GRCh38
rs765655881 Chr.4: 185159603-185159603 on Build GRCh38
rs551892159 Chr.4: 185159839-185159839 on Build GRCh38
rs1033080776 Chr.4: 185160036-185160036 on Build GRCh38
rs184497239 Chr.4: 185159758-185159758 on Build GRCh38
rs558167504 Chr.4: 185160020-185160020 on Build GRCh38
rs1048814804 Chr.4: 185159840-185159840 on Build GRCh38
rs891860623 Chr.4: 185159806-185159806 on Build GRCh38
rs908982782 Chr.4: 185159916-185159916 on Build GRCh38
rs570381085 Chr.4: 185159783-185159783 on Build GRCh38
rs199576227 Chr.4: 185159802-185159802 on Build GRCh38
rs755595955 Chr.4: 185159940-185159940 on Build GRCh38
rs143135869 Chr.4: 185159776-185159776 on Build GRCh38
rs957491214 Chr.4: 185160044-185160044 on Build GRCh38
rs532170433 Chr.4: 185159834-185159834 on Build GRCh38
rs953849231 Chr.4: 185160018-185160018 on Build GRCh38
rs971321434 Chr.4: 185159718-185159718 on Build GRCh38
rs573759447 Chr.4: 185159729-185159729 on Build GRCh38
rs151170734 Chr.4: 185159952-185159952 on Build GRCh38
rs753306716 Chr.4: 185159630-185159630 on Build GRCh38
rs866264512 Chr.4: 185159973-185159973 on Build GRCh38

View Full Product Details