Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 2750883-2751414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRX2
Entrez Gene ID
153572
Gene Name
iroquois homeobox 2
Gene Aliases
IRXA2
UniGene
Hs.282089
Gene Chromosome Location
Chr.5: 2707930-2751655 on Build GRCh38
Transcripts
NM_001134222.1 NM_033267.4 XM_011513979.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5413615 c.142_143insC p.Y48fs*142 Chr.5: 2751271-2751272 on Build GRCh38
6512794 c.41C>T p.S14L Chr.5: 2751373-2751373 on Build GRCh38
6512793 c.215C>T p.A72V Chr.5: 2751199-2751199 on Build GRCh38
6687500 c.47C>T p.A16V Chr.5: 2751367-2751367 on Build GRCh38
6687499 c.11C>A p.P4Q Chr.5: 2751403-2751403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888617936 Chr.5: 2750981-2750981 on Build GRCh38
rs1012604488 Chr.5: 2751182-2751187 on Build GRCh38
rs978849415 Chr.5: 2750948-2750948 on Build GRCh38
rs189486179 Chr.5: 2750923-2750923 on Build GRCh38
rs750317029 Chr.5: 2751348-2751348 on Build GRCh38
rs865774137 Chr.5: 2750963-2750963 on Build GRCh38
rs896878236 Chr.5: 2751169-2751169 on Build GRCh38
rs959317841 Chr.5: 2751067-2751067 on Build GRCh38
rs924633324 Chr.5: 2750954-2750954 on Build GRCh38
rs1050381239 Chr.5: 2750978-2750978 on Build GRCh38
rs1003126402 Chr.5: 2751044-2751044 on Build GRCh38
rs990751729 Chr.5: 2751072-2751072 on Build GRCh38
rs765814750 Chr.5: 2751360-2751360 on Build GRCh38
rs968302962 Chr.5: 2751196-2751196 on Build GRCh38
rs146008500 Chr.5: 2751016-2751016 on Build GRCh38
rs956027086 Chr.5: 2751132-2751132 on Build GRCh38
rs961016962 Chr.5: 2750926-2750927 on Build GRCh38
rs751083463 Chr.5: 2751372-2751372 on Build GRCh38
rs947499877 Chr.5: 2750946-2750946 on Build GRCh38
rs746409440 Chr.5: 2751183-2751183 on Build GRCh38
rs572713224 Chr.5: 2750942-2750942 on Build GRCh38
rs976715302 Chr.5: 2751134-2751134 on Build GRCh38
rs896844361 Chr.5: 2751029-2751029 on Build GRCh38
rs993010707 Chr.5: 2751079-2751079 on Build GRCh38
rs1005742675 Chr.5: 2750995-2750995 on Build GRCh38
rs924663768 Chr.5: 2751127-2751127 on Build GRCh38
rs986136145 Chr.5: 2751295-2751295 on Build GRCh38
rs574905642 Chr.5: 2751051-2751051 on Build GRCh38
rs751032410 Chr.5: 2751182-2751184 on Build GRCh38
rs775726682 Chr.5: 2751249-2751249 on Build GRCh38
rs1041336027 Chr.5: 2750966-2750966 on Build GRCh38
rs755344405 Chr.5: 2750915-2750915 on Build GRCh38
rs999324649 Chr.5: 2751233-2751233 on Build GRCh38
rs903042197 Chr.5: 2751280-2751281 on Build GRCh38
rs576005663 Chr.5: 2751246-2751246 on Build GRCh38
rs934617425 Chr.5: 2750962-2750962 on Build GRCh38
rs982007570 Chr.5: 2750931-2750931 on Build GRCh38
rs765133403 Chr.5: 2751339-2751339 on Build GRCh38
rs1037192052 Chr.5: 2751159-2751159 on Build GRCh38
rs760744147 Chr.5: 2751285-2751285 on Build GRCh38
rs539397952 Chr.5: 2750914-2750914 on Build GRCh38
rs776280500 Chr.5: 2751327-2751327 on Build GRCh38
rs976829815 Chr.5: 2751264-2751264 on Build GRCh38
rs758350668 Chr.5: 2751350-2751350 on Build GRCh38
rs922620568 Chr.5: 2751269-2751269 on Build GRCh38
rs932935457 Chr.5: 2751144-2751144 on Build GRCh38
rs901385856 Chr.5: 2750970-2750970 on Build GRCh38
rs11539256 Chr.5: 2751297-2751297 on Build GRCh38
rs1049998207 Chr.5: 2751148-2751148 on Build GRCh38
rs546809247 Chr.5: 2751028-2751029 on Build GRCh38
rs959203704 Chr.5: 2750937-2750937 on Build GRCh38
rs775047470 Chr.5: 2751117-2751117 on Build GRCh38
rs1003241482 Chr.5: 2751181-2751182 on Build GRCh38
rs541482658 Chr.5: 2750975-2750975 on Build GRCh38
rs967893098 Chr.5: 2751080-2751080 on Build GRCh38
rs922589519 Chr.5: 2751140-2751140 on Build GRCh38
rs377527492 Chr.5: 2751078-2751078 on Build GRCh38
rs991029094 Chr.5: 2750938-2750938 on Build GRCh38
rs564628487 Chr.5: 2751334-2751334 on Build GRCh38
rs541842586 Chr.5: 2751053-2751053 on Build GRCh38
rs910054919 Chr.5: 2751158-2751158 on Build GRCh38
rs754581300 Chr.5: 2751381-2751381 on Build GRCh38
rs779588233 Chr.5: 2751182-2751190 on Build GRCh38
rs1034877120 Chr.5: 2750933-2750933 on Build GRCh38
rs964616354 Chr.5: 2750913-2750913 on Build GRCh38

View Full Product Details