Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 45255955-45256495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN1
Entrez Gene ID
348980
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Gene Aliases
BCNG-1, BCNG1, EIEE24, HAC-2
UniGene
Hs.353176
Gene Chromosome Location
Chr.5: 45254950-45696118 on Build GRCh38
Transcripts
NM_021072.3

SNP Detail

SNP ID Chromosome Location
rs1033096072 Chr.5: 45256296-45256296 on Build GRCh38
rs1046775108 Chr.5: 45256344-45256344 on Build GRCh38
rs34239756 Chr.5: 45256373-45256374 on Build GRCh38
rs186412020 Chr.5: 45256145-45256145 on Build GRCh38
rs1050823208 Chr.5: 45256268-45256268 on Build GRCh38
rs545966543 Chr.5: 45256087-45256087 on Build GRCh38
rs1003751948 Chr.5: 45256247-45256247 on Build GRCh38
rs375811705 Chr.5: 45256146-45256146 on Build GRCh38
rs991367351 Chr.5: 45256018-45256018 on Build GRCh38
rs80097230 Chr.5: 45256391-45256391 on Build GRCh38
rs971243707 Chr.5: 45256389-45256389 on Build GRCh38
rs562120390 Chr.5: 45256323-45256323 on Build GRCh38
rs369410542 Chr.5: 45256303-45256303 on Build GRCh38
rs543560738 Chr.5: 45256075-45256076 on Build GRCh38
rs372808808 Chr.5: 45255991-45255991 on Build GRCh38
rs905338255 Chr.5: 45256065-45256065 on Build GRCh38
rs1004390724 Chr.5: 45256286-45256286 on Build GRCh38
rs760375460 Chr.5: 45256413-45256413 on Build GRCh38
rs962781465 Chr.5: 45256131-45256131 on Build GRCh38
rs777878090 Chr.5: 45256241-45256241 on Build GRCh38
rs562530658 Chr.5: 45256101-45256101 on Build GRCh38
rs998572393 Chr.5: 45256288-45256288 on Build GRCh38
rs542381904 Chr.5: 45256260-45256260 on Build GRCh38
rs975370286 Chr.5: 45256350-45256350 on Build GRCh38
rs1012854898 Chr.5: 45256302-45256302 on Build GRCh38
rs1024239321 Chr.5: 45256038-45256038 on Build GRCh38
rs112382387 Chr.5: 45256175-45256175 on Build GRCh38
rs774366496 Chr.5: 45256028-45256028 on Build GRCh38
rs1054121474 Chr.5: 45255990-45255990 on Build GRCh38
rs757458816 Chr.5: 45256450-45256450 on Build GRCh38
rs951713519 Chr.5: 45256093-45256093 on Build GRCh38
rs957030052 Chr.5: 45256300-45256300 on Build GRCh38
rs962707912 Chr.5: 45256341-45256341 on Build GRCh38
rs1028441415 Chr.5: 45256419-45256419 on Build GRCh38
rs79443250 Chr.5: 45256392-45256392 on Build GRCh38
rs975445282 Chr.5: 45256139-45256139 on Build GRCh38
rs34921695 Chr.5: 45256391-45256391 on Build GRCh38
rs1054130868 Chr.5: 45256172-45256172 on Build GRCh38
rs867609193 Chr.5: 45256168-45256168 on Build GRCh38
rs376997531 Chr.5: 45256372-45256373 on Build GRCh38
rs111958996 Chr.5: 45256253-45256253 on Build GRCh38
rs181296325 Chr.5: 45256073-45256073 on Build GRCh38
rs906935224 Chr.5: 45256242-45256242 on Build GRCh38
rs941609774 Chr.5: 45256201-45256203 on Build GRCh38
rs981261210 Chr.5: 45256393-45256393 on Build GRCh38
rs934525879 Chr.5: 45256171-45256171 on Build GRCh38
rs1016908244 Chr.5: 45256312-45256312 on Build GRCh38
rs532035519 Chr.5: 45256374-45256375 on Build GRCh38
rs1016853752 Chr.5: 45256103-45256103 on Build GRCh38
rs1012638947 Chr.5: 45256032-45256032 on Build GRCh38
rs866595686 Chr.5: 45256315-45256315 on Build GRCh38
rs1035147010 Chr.5: 45256428-45256428 on Build GRCh38
rs921329778 Chr.5: 45256142-45256142 on Build GRCh38
rs933736192 Chr.5: 45256166-45256166 on Build GRCh38
rs553837878 Chr.5: 45256043-45256043 on Build GRCh38
rs887365993 Chr.5: 45256272-45256272 on Build GRCh38
rs1044177522 Chr.5: 45256228-45256228 on Build GRCh38
rs892780085 Chr.5: 45256174-45256174 on Build GRCh38

View Full Product Details