Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 45260902-45261406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN1
Entrez Gene ID
348980
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Gene Aliases
BCNG-1, BCNG1, EIEE24, HAC-2
UniGene
Hs.353176
Gene Chromosome Location
Chr.5: 45254950-45696118 on Build GRCh38
Transcripts
NM_021072.3

SNP Detail

SNP ID Chromosome Location
rs888324806 Chr.5: 45261354-45261354 on Build GRCh38
rs1005600066 Chr.5: 45261134-45261134 on Build GRCh38
rs916709682 Chr.5: 45261295-45261295 on Build GRCh38
rs146206633 Chr.5: 45261351-45261352 on Build GRCh38
rs191985582 Chr.5: 45261372-45261372 on Build GRCh38
rs748522033 Chr.5: 45261074-45261074 on Build GRCh38
rs547335427 Chr.5: 45260944-45260944 on Build GRCh38
rs906494004 Chr.5: 45261037-45261037 on Build GRCh38
rs149744675 Chr.5: 45261221-45261221 on Build GRCh38
rs555001055 Chr.5: 45261117-45261117 on Build GRCh38
rs537999572 Chr.5: 45261002-45261002 on Build GRCh38
rs182394918 Chr.5: 45261302-45261302 on Build GRCh38
rs989848307 Chr.5: 45261292-45261292 on Build GRCh38
rs947893130 Chr.5: 45261019-45261019 on Build GRCh38
rs757378689 Chr.5: 45261243-45261243 on Build GRCh38
rs1042413174 Chr.5: 45261080-45261080 on Build GRCh38
rs768421883 Chr.5: 45260931-45260931 on Build GRCh38
rs759676407 Chr.5: 45261374-45261374 on Build GRCh38
rs923345026 Chr.5: 45261020-45261020 on Build GRCh38
rs1046212691 Chr.5: 45261027-45261027 on Build GRCh38
rs559025160 Chr.5: 45260996-45260996 on Build GRCh38
rs539315058 Chr.5: 45260974-45260974 on Build GRCh38
rs772498478 Chr.5: 45261212-45261212 on Build GRCh38
rs981938747 Chr.5: 45261308-45261308 on Build GRCh38
rs572836605 Chr.5: 45261279-45261279 on Build GRCh38
rs996382913 Chr.5: 45261226-45261226 on Build GRCh38
rs929785998 Chr.5: 45261347-45261347 on Build GRCh38
rs989593649 Chr.5: 45261005-45261005 on Build GRCh38
rs916618561 Chr.5: 45261008-45261008 on Build GRCh38
rs529049340 Chr.5: 45261079-45261079 on Build GRCh38
rs942511541 Chr.5: 45260979-45260979 on Build GRCh38
rs569224195 Chr.5: 45260997-45260997 on Build GRCh38
rs139660444 Chr.5: 45260962-45260962 on Build GRCh38
rs187397058 Chr.5: 45261353-45261353 on Build GRCh38
rs929880939 Chr.5: 45261077-45261077 on Build GRCh38
rs191386456 Chr.5: 45261153-45261153 on Build GRCh38
rs552869843 Chr.5: 45261244-45261244 on Build GRCh38
rs936646328 Chr.5: 45260998-45260998 on Build GRCh38
rs1030493277 Chr.5: 45261233-45261233 on Build GRCh38
rs952359341 Chr.5: 45261350-45261350 on Build GRCh38
rs547294512 Chr.5: 45261081-45261081 on Build GRCh38
rs1031642960 Chr.5: 45261274-45261274 on Build GRCh38
rs1018029224 Chr.5: 45261168-45261168 on Build GRCh38
rs899538476 Chr.5: 45261225-45261225 on Build GRCh38
rs927781825 Chr.5: 45261309-45261309 on Build GRCh38
rs760761636 Chr.5: 45261297-45261297 on Build GRCh38
rs530478432 Chr.5: 45261370-45261370 on Build GRCh38

View Full Product Details