Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 51382885-51383386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISL1
Entrez Gene ID
3670
Gene Name
ISL LIM homeobox 1
Gene Aliases
ISLET1, Isl-1
UniGene
Hs.505
Gene Chromosome Location
Chr.5: 51383124-51394730 on Build GRCh38
Transcripts
NM_002202.2 NR_046243.1

Gene Symbol
LOC642366
Entrez Gene ID
642366
Gene Name
uncharacterized LOC642366
Gene Aliases
-
UniGene
Hs.544139
Gene Chromosome Location
Chr.5: 51372736-51383332 on Build GRCh38
Transcripts
NM_002202.2 NR_046243.1

SNP Detail

SNP ID Chromosome Location
rs909283194 Chr.5: 51382918-51382918 on Build GRCh38
rs1052076311 Chr.5: 51383019-51383019 on Build GRCh38
rs1035886396 Chr.5: 51383234-51383234 on Build GRCh38
rs1057481552 Chr.5: 51383294-51383294 on Build GRCh38
rs751548119 Chr.5: 51383179-51383179 on Build GRCh38
rs905988258 Chr.5: 51383084-51383084 on Build GRCh38
rs1037933025 Chr.5: 51383320-51383320 on Build GRCh38
rs3762977 Chr.5: 51383180-51383180 on Build GRCh38
rs974758162 Chr.5: 51383254-51383254 on Build GRCh38
rs915177829 Chr.5: 51383272-51383272 on Build GRCh38
rs1003441672 Chr.5: 51383229-51383229 on Build GRCh38
rs761906656 Chr.5: 51383258-51383258 on Build GRCh38
rs1029461006 Chr.5: 51383255-51383255 on Build GRCh38
rs906257598 Chr.5: 51383289-51383289 on Build GRCh38
rs902444619 Chr.5: 51383024-51383024 on Build GRCh38
rs899426402 Chr.5: 51383345-51383345 on Build GRCh38
rs1038883448 Chr.5: 51382952-51382952 on Build GRCh38
rs71618117 Chr.5: 51383190-51383190 on Build GRCh38
rs535478822 Chr.5: 51382977-51382977 on Build GRCh38
rs531848833 Chr.5: 51383250-51383250 on Build GRCh38
rs770978830 Chr.5: 51382939-51382940 on Build GRCh38
rs951025430 Chr.5: 51383126-51383131 on Build GRCh38
rs996164231 Chr.5: 51383352-51383352 on Build GRCh38
rs749522827 Chr.5: 51383091-51383091 on Build GRCh38
rs756386972 Chr.5: 51383228-51383228 on Build GRCh38
rs550926254 Chr.5: 51382978-51382978 on Build GRCh38
rs954844452 Chr.5: 51383257-51383257 on Build GRCh38
rs542209929 Chr.5: 51383175-51383175 on Build GRCh38
rs974789531 Chr.5: 51383148-51383148 on Build GRCh38
rs922041159 Chr.5: 51383156-51383156 on Build GRCh38
rs765255469 Chr.5: 51383057-51383057 on Build GRCh38
rs576234312 Chr.5: 51382920-51382920 on Build GRCh38
rs527789216 Chr.5: 51383280-51383280 on Build GRCh38
rs570852043 Chr.5: 51383350-51383350 on Build GRCh38
rs963581379 Chr.5: 51383143-51383143 on Build GRCh38
rs933123895 Chr.5: 51383173-51383173 on Build GRCh38
rs561698186 Chr.5: 51383271-51383271 on Build GRCh38
rs935271310 Chr.5: 51383182-51383182 on Build GRCh38
rs539646165 Chr.5: 51383357-51383357 on Build GRCh38
rs886572343 Chr.5: 51383309-51383309 on Build GRCh38
rs935175624 Chr.5: 51383262-51383262 on Build GRCh38
rs1053380187 Chr.5: 51383187-51383187 on Build GRCh38
rs1011987509 Chr.5: 51383074-51383074 on Build GRCh38
rs1053411738 Chr.5: 51383035-51383035 on Build GRCh38
rs377679399 Chr.5: 51383247-51383247 on Build GRCh38
rs1016532854 Chr.5: 51383253-51383253 on Build GRCh38
rs374921619 Chr.5: 51383237-51383237 on Build GRCh38
rs1016312907 Chr.5: 51383142-51383142 on Build GRCh38
rs947728678 Chr.5: 51383198-51383198 on Build GRCh38
rs933174916 Chr.5: 51383005-51383005 on Build GRCh38
rs572194082 Chr.5: 51383027-51383027 on Build GRCh38
rs759365410 Chr.5: 51382949-51382949 on Build GRCh38
rs541081886 Chr.5: 51383046-51383046 on Build GRCh38
rs750642961 Chr.5: 51383204-51383204 on Build GRCh38
rs1003062099 Chr.5: 51383107-51383107 on Build GRCh38
rs983433098 Chr.5: 51383140-51383140 on Build GRCh38
rs1044770966 Chr.5: 51383221-51383221 on Build GRCh38
rs577409885 Chr.5: 51383117-51383117 on Build GRCh38
rs540370407 Chr.5: 51383267-51383267 on Build GRCh38
rs886688613 Chr.5: 51383236-51383236 on Build GRCh38
rs547910932 Chr.5: 51383316-51383316 on Build GRCh38
rs1023351090 Chr.5: 51383082-51383082 on Build GRCh38
rs947696160 Chr.5: 51383276-51383276 on Build GRCh38
rs954953693 Chr.5: 51383353-51383353 on Build GRCh38
rs942116825 Chr.5: 51382946-51382946 on Build GRCh38
rs939154555 Chr.5: 51383292-51383292 on Build GRCh38
rs776558382 Chr.5: 51383353-51383353 on Build GRCh38
rs560531941 Chr.5: 51383113-51383113 on Build GRCh38

View Full Product Details