Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55031273-55031773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMK
Entrez Gene ID
3003
Gene Name
granzyme K
Gene Aliases
TRYP2
UniGene
Hs.277937
Gene Chromosome Location
Chr.5: 55024279-55034132 on Build GRCh38
Transcripts
NM_002104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6677325 c.594C>A p.V198V Chr.5: 55031594-55031594 on Build GRCh38
329390 c.624T>C p.D208D Chr.5: 55031624-55031624 on Build GRCh38
6569709 c.563A>C p.N188T Chr.5: 55031563-55031563 on Build GRCh38
4554445 c.621G>A p.K207K Chr.5: 55031621-55031621 on Build GRCh38
6677328 c.375C>T p.A125A Chr.5: 55031375-55031375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773433624 Chr.5: 55031354-55031354 on Build GRCh38
rs35259610 Chr.5: 55031441-55031441 on Build GRCh38
rs765024687 Chr.5: 55031640-55031640 on Build GRCh38
rs192602074 Chr.5: 55031403-55031403 on Build GRCh38
rs867639749 Chr.5: 55031565-55031565 on Build GRCh38
rs754488159 Chr.5: 55031476-55031476 on Build GRCh38
rs750021891 Chr.5: 55031391-55031391 on Build GRCh38
rs751541408 Chr.5: 55031568-55031568 on Build GRCh38
rs760813589 Chr.5: 55031355-55031355 on Build GRCh38
rs775052768 Chr.5: 55031465-55031476 on Build GRCh38
rs762532246 Chr.5: 55031707-55031709 on Build GRCh38
rs987783257 Chr.5: 55031324-55031324 on Build GRCh38
rs192691619 Chr.5: 55031725-55031725 on Build GRCh38
rs915952083 Chr.5: 55031659-55031659 on Build GRCh38
rs377736354 Chr.5: 55031376-55031376 on Build GRCh38
rs775119162 Chr.5: 55031642-55031642 on Build GRCh38
rs576026064 Chr.5: 55031358-55031358 on Build GRCh38
rs372389313 Chr.5: 55031538-55031538 on Build GRCh38
rs139113584 Chr.5: 55031435-55031435 on Build GRCh38
rs770055265 Chr.5: 55031598-55031598 on Build GRCh38
rs764476549 Chr.5: 55031390-55031390 on Build GRCh38
rs920356531 Chr.5: 55031394-55031394 on Build GRCh38
rs765776190 Chr.5: 55031375-55031375 on Build GRCh38
rs778595106 Chr.5: 55031484-55031484 on Build GRCh38
rs554814374 Chr.5: 55031472-55031472 on Build GRCh38
rs751390705 Chr.5: 55031668-55031668 on Build GRCh38
rs781226539 Chr.5: 55031504-55031504 on Build GRCh38
rs748116368 Chr.5: 55031410-55031410 on Build GRCh38
rs78739734 Chr.5: 55031564-55031564 on Build GRCh38
rs752332855 Chr.5: 55031532-55031532 on Build GRCh38
rs760367862 Chr.5: 55031467-55031467 on Build GRCh38
rs780301056 Chr.5: 55031682-55031682 on Build GRCh38
rs369110428 Chr.5: 55031523-55031523 on Build GRCh38
rs749407552 Chr.5: 55031606-55031606 on Build GRCh38
rs143795810 Chr.5: 55031411-55031411 on Build GRCh38
rs561131107 Chr.5: 55031392-55031392 on Build GRCh38
rs867228118 Chr.5: 55031482-55031482 on Build GRCh38
rs566625175 Chr.5: 55031581-55031581 on Build GRCh38
rs866260127 Chr.5: 55031566-55031566 on Build GRCh38
rs781500556 Chr.5: 55031365-55031365 on Build GRCh38
rs759664624 Chr.5: 55031448-55031448 on Build GRCh38
rs774279232 Chr.5: 55031329-55031329 on Build GRCh38
rs372763068 Chr.5: 55031505-55031505 on Build GRCh38
rs144778115 Chr.5: 55031506-55031506 on Build GRCh38
rs763694403 Chr.5: 55031667-55031667 on Build GRCh38
rs759077808 Chr.5: 55031346-55031347 on Build GRCh38
rs752613681 Chr.5: 55031490-55031490 on Build GRCh38
rs765985227 Chr.5: 55031471-55031471 on Build GRCh38
rs781450102 Chr.5: 55031416-55031416 on Build GRCh38
rs771531426 Chr.5: 55031459-55031459 on Build GRCh38
rs145011379 Chr.5: 55031313-55031313 on Build GRCh38
rs764828378 Chr.5: 55031557-55031557 on Build GRCh38
rs766175872 Chr.5: 55031335-55031335 on Build GRCh38
rs772400267 Chr.5: 55031353-55031353 on Build GRCh38
rs149102030 Chr.5: 55031462-55031462 on Build GRCh38
rs747995734 Chr.5: 55031337-55031337 on Build GRCh38
rs372502807 Chr.5: 55031615-55031615 on Build GRCh38
rs140185452 Chr.5: 55031620-55031620 on Build GRCh38
rs970527121 Chr.5: 55031687-55031687 on Build GRCh38
rs759258226 Chr.5: 55031635-55031635 on Build GRCh38
rs1010753520 Chr.5: 55031303-55031303 on Build GRCh38
rs978895453 Chr.5: 55031712-55031712 on Build GRCh38
rs200532050 Chr.5: 55031600-55031600 on Build GRCh38
rs750280617 Chr.5: 55031676-55031676 on Build GRCh38
rs4865597 Chr.5: 55031689-55031689 on Build GRCh38
rs1046012948 Chr.5: 55031437-55031437 on Build GRCh38
rs368111434 Chr.5: 55031308-55031308 on Build GRCh38
rs753500430 Chr.5: 55031533-55031533 on Build GRCh38
rs776632763 Chr.5: 55031360-55031360 on Build GRCh38
rs762295925 Chr.5: 55031466-55031467 on Build GRCh38
rs758898648 Chr.5: 55031409-55031409 on Build GRCh38
rs777620371 Chr.5: 55031497-55031497 on Build GRCh38
rs747244335 Chr.5: 55031440-55031440 on Build GRCh38
rs760032485 Chr.5: 55031371-55031371 on Build GRCh38
rs187556392 Chr.5: 55031398-55031398 on Build GRCh38
rs1033098531 Chr.5: 55031300-55031300 on Build GRCh38
rs919231178 Chr.5: 55031296-55031296 on Build GRCh38
rs763434219 Chr.5: 55031518-55031518 on Build GRCh38
rs142007081 Chr.5: 55031567-55031567 on Build GRCh38
rs538081918 Chr.5: 55031643-55031643 on Build GRCh38
rs368180397 Chr.5: 55031576-55031576 on Build GRCh38
rs146138819 Chr.5: 55031589-55031589 on Build GRCh38
rs549132428 Chr.5: 55031333-55031333 on Build GRCh38
rs750796943 Chr.5: 55031443-55031443 on Build GRCh38
rs775557536 Chr.5: 55031511-55031511 on Build GRCh38
rs555947639 Chr.5: 55031677-55031677 on Build GRCh38
rs771165949 Chr.5: 55031503-55031503 on Build GRCh38
rs545000046 Chr.5: 55031306-55031306 on Build GRCh38
rs751925124 Chr.5: 55031465-55031465 on Build GRCh38
rs145184473 Chr.5: 55031301-55031301 on Build GRCh38
rs186746029 Chr.5: 55031608-55031608 on Build GRCh38
rs757235663 Chr.5: 55031669-55031669 on Build GRCh38
rs573855509 Chr.5: 55031612-55031612 on Build GRCh38
rs764459042 Chr.5: 55031463-55031463 on Build GRCh38
rs146397309 Chr.5: 55031423-55031423 on Build GRCh38
rs370792575 Chr.5: 55031495-55031495 on Build GRCh38
rs746820075 Chr.5: 55031502-55031502 on Build GRCh38
rs376887235 Chr.5: 55031522-55031522 on Build GRCh38
rs769889388 Chr.5: 55031451-55031451 on Build GRCh38
rs376294307 Chr.5: 55031446-55031446 on Build GRCh38
rs951856522 Chr.5: 55031715-55031715 on Build GRCh38
rs370635228 Chr.5: 55031405-55031405 on Build GRCh38
rs775939517 Chr.5: 55031460-55031460 on Build GRCh38
rs781502723 Chr.5: 55031594-55031594 on Build GRCh38
rs758715971 Chr.5: 55031383-55031383 on Build GRCh38

View Full Product Details