Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 90558693-90559223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRV1
Entrez Gene ID
84059
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
UniGene
Hs.591777
Gene Chromosome Location
Chr.5: 90558681-91164226 on Build GRCh38
Transcripts
NM_032119.3 NR_003149.1 XM_017009963.1 XM_017009964.1 XM_017009965.1 XM_017009966.1 XM_017009967.1 XM_017009968.1 XM_017009969.1 XM_017009970.1 XM_017009971.1 XM_017009974.1

SNP Detail

SNP ID Chromosome Location
rs116110048 Chr.5: 90558849-90558849 on Build GRCh38
rs35794858 Chr.5: 90559036-90559037 on Build GRCh38
rs561663289 Chr.5: 90559122-90559124 on Build GRCh38
rs556739703 Chr.5: 90558901-90558901 on Build GRCh38
rs973537840 Chr.5: 90558910-90558910 on Build GRCh38
rs954760195 Chr.5: 90559107-90559107 on Build GRCh38
rs763582206 Chr.5: 90558893-90558893 on Build GRCh38
rs774010727 Chr.5: 90558897-90558897 on Build GRCh38
rs154567 Chr.5: 90558796-90558796 on Build GRCh38
rs192362302 Chr.5: 90558852-90558852 on Build GRCh38
rs1010785089 Chr.5: 90559018-90559018 on Build GRCh38
rs1005271445 Chr.5: 90559122-90559122 on Build GRCh38
rs371383993 Chr.5: 90558856-90558856 on Build GRCh38
rs556073416 Chr.5: 90559056-90559056 on Build GRCh38
rs969507178 Chr.5: 90559176-90559176 on Build GRCh38
rs889969165 Chr.5: 90558843-90558843 on Build GRCh38
rs779879190 Chr.5: 90558842-90558842 on Build GRCh38
rs963954233 Chr.5: 90558834-90558834 on Build GRCh38
rs752199647 Chr.5: 90558800-90558800 on Build GRCh38
rs924941030 Chr.5: 90559062-90559062 on Build GRCh38
rs977833240 Chr.5: 90559027-90559027 on Build GRCh38
rs984658381 Chr.5: 90558749-90558749 on Build GRCh38
rs916751525 Chr.5: 90559175-90559175 on Build GRCh38
rs951941826 Chr.5: 90558725-90558725 on Build GRCh38
rs564682921 Chr.5: 90559155-90559155 on Build GRCh38
rs899627732 Chr.5: 90559071-90559071 on Build GRCh38
rs727504831 Chr.5: 90558916-90558916 on Build GRCh38
rs771283759 Chr.5: 90558783-90558783 on Build GRCh38
rs777776792 Chr.5: 90558965-90558965 on Build GRCh38
rs576014032 Chr.5: 90559090-90559090 on Build GRCh38
rs936316203 Chr.5: 90559065-90559065 on Build GRCh38
rs528592353 Chr.5: 90559150-90559150 on Build GRCh38
rs368604803 Chr.5: 90558926-90558926 on Build GRCh38
rs998120425 Chr.5: 90559072-90559072 on Build GRCh38
rs751250505 Chr.5: 90558827-90558827 on Build GRCh38
rs544818133 Chr.5: 90558787-90558787 on Build GRCh38
rs528051508 Chr.5: 90558832-90558832 on Build GRCh38
rs959154664 Chr.5: 90558765-90558765 on Build GRCh38
rs548497029 Chr.5: 90558835-90558835 on Build GRCh38
rs750189983 Chr.5: 90558831-90558831 on Build GRCh38
rs746237291 Chr.5: 90558871-90558871 on Build GRCh38
rs966853597 Chr.5: 90558986-90558986 on Build GRCh38
rs367934526 Chr.5: 90558866-90558866 on Build GRCh38
rs893654315 Chr.5: 90558809-90558809 on Build GRCh38
rs778984818 Chr.5: 90558755-90558755 on Build GRCh38
rs773403691 Chr.5: 90558771-90558771 on Build GRCh38
rs753717540 Chr.5: 90558958-90558958 on Build GRCh38
rs552599194 Chr.5: 90558729-90558729 on Build GRCh38
rs73176098 Chr.5: 90558750-90558750 on Build GRCh38
rs749480523 Chr.5: 90559044-90559044 on Build GRCh38
rs375309830 Chr.5: 90558900-90558900 on Build GRCh38
rs200137338 Chr.5: 90558948-90558948 on Build GRCh38
rs372874315 Chr.5: 90558934-90558934 on Build GRCh38
rs776937064 Chr.5: 90558868-90558868 on Build GRCh38
rs947761338 Chr.5: 90558737-90558737 on Build GRCh38
rs945085670 Chr.5: 90558808-90558808 on Build GRCh38
rs150816712 Chr.5: 90558829-90558829 on Build GRCh38
rs727504484 Chr.5: 90558885-90558885 on Build GRCh38
rs760931458 Chr.5: 90558775-90558775 on Build GRCh38
rs7706392 Chr.5: 90558973-90558973 on Build GRCh38
rs973426725 Chr.5: 90559156-90559156 on Build GRCh38
rs7706355 Chr.5: 90558798-90558798 on Build GRCh38
rs954849459 Chr.5: 90558853-90558853 on Build GRCh38
rs1029556456 Chr.5: 90559097-90559097 on Build GRCh38
rs754970339 Chr.5: 90558960-90558960 on Build GRCh38
rs748209771 Chr.5: 90558764-90558764 on Build GRCh38
rs41311623 Chr.5: 90558979-90558979 on Build GRCh38
rs770240032 Chr.5: 90558875-90558875 on Build GRCh38
rs551948481 Chr.5: 90558823-90558823 on Build GRCh38
rs763683206 Chr.5: 90558894-90558894 on Build GRCh38
rs776515209 Chr.5: 90559061-90559061 on Build GRCh38
rs570271767 Chr.5: 90558908-90558908 on Build GRCh38
rs767310903 Chr.5: 90558923-90558923 on Build GRCh38
rs927671934 Chr.5: 90558959-90558959 on Build GRCh38
rs772290118 Chr.5: 90558769-90558769 on Build GRCh38
rs111857611 Chr.5: 90558881-90558881 on Build GRCh38
rs963784682 Chr.5: 90559133-90559133 on Build GRCh38
rs893529649 Chr.5: 90558997-90558997 on Build GRCh38
rs1041608290 Chr.5: 90559162-90559162 on Build GRCh38
rs747223342 Chr.5: 90558855-90558855 on Build GRCh38
rs544881512 Chr.5: 90559130-90559130 on Build GRCh38
rs762345137 Chr.5: 90558808-90558808 on Build GRCh38

View Full Product Details