Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 90619083-90619583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRV1
Entrez Gene ID
84059
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
UniGene
Hs.591777
Gene Chromosome Location
Chr.5: 90558681-91164226 on Build GRCh38
Transcripts
NM_032119.3 NR_003149.1 XM_017009963.1 XM_017009964.1 XM_017009965.1 XM_017009966.1 XM_017009967.1 XM_017009968.1 XM_017009969.1 XM_017009970.1 XM_017009971.1 XM_017009974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
261167 c.381T>C p.L127L Chr.5: 90619109-90619109 on Build GRCh38
6251251 c.403G>T p.V135L Chr.5: 90619131-90619131 on Build GRCh38
6047372 c.382G>T p.G128* Chr.5: 90619110-90619110 on Build GRCh38
3920445 c.443C>T p.S148L Chr.5: 90619171-90619171 on Build GRCh38
5868928 c.401C>T p.T134I Chr.5: 90619129-90619129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183238323 Chr.5: 90619228-90619228 on Build GRCh38
rs539914117 Chr.5: 90619230-90619230 on Build GRCh38
rs868740040 Chr.5: 90619239-90619239 on Build GRCh38
rs373664192 Chr.5: 90619130-90619130 on Build GRCh38
rs933732970 Chr.5: 90619394-90619394 on Build GRCh38
rs777818974 Chr.5: 90619149-90619149 on Build GRCh38
rs751649763 Chr.5: 90619196-90619198 on Build GRCh38
rs768784430 Chr.5: 90619189-90619189 on Build GRCh38
rs989735864 Chr.5: 90619345-90619345 on Build GRCh38
rs907293345 Chr.5: 90619344-90619344 on Build GRCh38
rs970793757 Chr.5: 90619252-90619252 on Build GRCh38
rs775440063 Chr.5: 90619177-90619177 on Build GRCh38
rs727504665 Chr.5: 90619153-90619153 on Build GRCh38
rs900890586 Chr.5: 90619393-90619393 on Build GRCh38
rs574668847 Chr.5: 90619257-90619257 on Build GRCh38
rs16868854 Chr.5: 90619204-90619204 on Build GRCh38
rs1031680812 Chr.5: 90619452-90619452 on Build GRCh38
rs747004600 Chr.5: 90619151-90619151 on Build GRCh38
rs190443389 Chr.5: 90619521-90619521 on Build GRCh38
rs982204915 Chr.5: 90619313-90619313 on Build GRCh38
rs915502806 Chr.5: 90619374-90619374 on Build GRCh38
rs763180112 Chr.5: 90619188-90619188 on Build GRCh38
rs565096556 Chr.5: 90619547-90619547 on Build GRCh38
rs778709054 Chr.5: 90619111-90619111 on Build GRCh38
rs573447853 Chr.5: 90619328-90619328 on Build GRCh38
rs531887122 Chr.5: 90619436-90619436 on Build GRCh38
rs769994130 Chr.5: 90619170-90619170 on Build GRCh38
rs942359467 Chr.5: 90619377-90619377 on Build GRCh38
rs1002839679 Chr.5: 90619387-90619387 on Build GRCh38
rs761236403 Chr.5: 90619226-90619226 on Build GRCh38
rs924110622 Chr.5: 90619337-90619337 on Build GRCh38
rs1022323452 Chr.5: 90619510-90619510 on Build GRCh38
rs1012517937 Chr.5: 90619260-90619260 on Build GRCh38
rs1039403486 Chr.5: 90619378-90619378 on Build GRCh38
rs886931998 Chr.5: 90619474-90619474 on Build GRCh38
rs185748582 Chr.5: 90619336-90619336 on Build GRCh38
rs1017292494 Chr.5: 90619213-90619213 on Build GRCh38
rs777138250 Chr.5: 90619542-90619542 on Build GRCh38
rs562775324 Chr.5: 90619375-90619375 on Build GRCh38
rs1016825367 Chr.5: 90619518-90619520 on Build GRCh38
rs191815005 Chr.5: 90619211-90619211 on Build GRCh38
rs923018892 Chr.5: 90619545-90619545 on Build GRCh38
rs990393762 Chr.5: 90619475-90619475 on Build GRCh38
rs575602255 Chr.5: 90619137-90619137 on Build GRCh38
rs774739780 Chr.5: 90619190-90619190 on Build GRCh38
rs956064709 Chr.5: 90619465-90619465 on Build GRCh38
rs762135855 Chr.5: 90619197-90619197 on Build GRCh38
rs41311333 Chr.5: 90619108-90619108 on Build GRCh38
rs999555008 Chr.5: 90619460-90619472 on Build GRCh38
rs1023026258 Chr.5: 90619311-90619311 on Build GRCh38
rs951231140 Chr.5: 90619209-90619209 on Build GRCh38

View Full Product Details