Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 94152381-94152928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0825
Entrez Gene ID
285600
Gene Name
KIAA0825
Gene Aliases
C5orf36
UniGene
Hs.425123
Gene Chromosome Location
Chr.5: 94150851-94618604 on Build GRCh38
Transcripts
NM_001145678.1 XM_006714588.2

SNP Detail

SNP ID Chromosome Location
rs978266725 Chr.5: 94152765-94152765 on Build GRCh38
rs993946726 Chr.5: 94152540-94152540 on Build GRCh38
rs979588491 Chr.5: 94152680-94152680 on Build GRCh38
rs776007077 Chr.5: 94152537-94152537 on Build GRCh38
rs561081580 Chr.5: 94152484-94152484 on Build GRCh38
rs55755253 Chr.5: 94152861-94152861 on Build GRCh38
rs868118378 Chr.5: 94152855-94152855 on Build GRCh38
rs550530841 Chr.5: 94152881-94152901 on Build GRCh38
rs921433963 Chr.5: 94152694-94152694 on Build GRCh38
rs890808917 Chr.5: 94152513-94152513 on Build GRCh38
rs549936222 Chr.5: 94152679-94152679 on Build GRCh38
rs56060568 Chr.5: 94152897-94152897 on Build GRCh38
rs968613944 Chr.5: 94152677-94152677 on Build GRCh38
rs1022812863 Chr.5: 94152553-94152553 on Build GRCh38
rs6892655 Chr.5: 94152903-94152903 on Build GRCh38
rs915321734 Chr.5: 94152705-94152705 on Build GRCh38
rs995389000 Chr.5: 94152548-94152548 on Build GRCh38
rs939921550 Chr.5: 94152515-94152515 on Build GRCh38
rs868039400 Chr.5: 94152870-94152870 on Build GRCh38
rs56394625 Chr.5: 94152864-94152864 on Build GRCh38
rs1050677824 Chr.5: 94152498-94152498 on Build GRCh38
rs74424283 Chr.5: 94152757-94152757 on Build GRCh38
rs55690460 Chr.5: 94152899-94152903 on Build GRCh38
rs768099582 Chr.5: 94152638-94152638 on Build GRCh38
rs974866067 Chr.5: 94152681-94152681 on Build GRCh38
rs946929032 Chr.5: 94152405-94152405 on Build GRCh38
rs10530351 Chr.5: 94152856-94152857 on Build GRCh38
rs979587933 Chr.5: 94152408-94152408 on Build GRCh38
rs1036976206 Chr.5: 94152531-94152531 on Build GRCh38
rs529741431 Chr.5: 94152507-94152507 on Build GRCh38
rs10055994 Chr.5: 94152720-94152720 on Build GRCh38
rs181938570 Chr.5: 94152736-94152736 on Build GRCh38
rs565600769 Chr.5: 94152715-94152715 on Build GRCh38
rs866107778 Chr.5: 94152868-94152868 on Build GRCh38
rs552071863 Chr.5: 94152658-94152658 on Build GRCh38
rs894902165 Chr.5: 94152428-94152428 on Build GRCh38
rs567789264 Chr.5: 94152753-94152753 on Build GRCh38
rs28460476 Chr.5: 94152856-94152856 on Build GRCh38
rs72771613 Chr.5: 94152797-94152797 on Build GRCh38
rs147680136 Chr.5: 94152654-94152654 on Build GRCh38
rs1011912257 Chr.5: 94152446-94152446 on Build GRCh38
rs56294349 Chr.5: 94152859-94152859 on Build GRCh38
rs541591862 Chr.5: 94152412-94152412 on Build GRCh38
rs1002307883 Chr.5: 94152586-94152586 on Build GRCh38
rs540112403 Chr.5: 94152614-94152614 on Build GRCh38
rs905239672 Chr.5: 94152554-94152554 on Build GRCh38
rs145346072 Chr.5: 94152639-94152639 on Build GRCh38

View Full Product Details