Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135895283-135895814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL9
Entrez Gene ID
3578
Gene Name
interleukin 9
Gene Aliases
HP40, IL-9, P40
UniGene
Hs.960
Gene Chromosome Location
Chr.5: 135892246-135895827 on Build GRCh38
Transcripts
NM_000590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685461 c.150+9delA p.? Chr.5: 135895546-135895546 on Build GRCh38
6685460 c.115-10_115-7delTCTA p.? Chr.5: 135895597-135895600 on Build GRCh38
4397349 c.44C>T p.S15F Chr.5: 135895773-135895773 on Build GRCh38
6349013 c.144T>A p.S48R Chr.5: 135895561-135895561 on Build GRCh38
3975169 c.75G>A p.A25A Chr.5: 135895742-135895742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776009290 Chr.5: 135895410-135895410 on Build GRCh38
rs367906598 Chr.5: 135895629-135895629 on Build GRCh38
rs752469605 Chr.5: 135895633-135895633 on Build GRCh38
rs775539518 Chr.5: 135895633-135895633 on Build GRCh38
rs774133953 Chr.5: 135895771-135895771 on Build GRCh38
rs55692658 Chr.5: 135895540-135895540 on Build GRCh38
rs543055457 Chr.5: 135895369-135895369 on Build GRCh38
rs768479610 Chr.5: 135895770-135895770 on Build GRCh38
rs773103703 Chr.5: 135895434-135895434 on Build GRCh38
rs755424202 Chr.5: 135895599-135895599 on Build GRCh38
rs868627777 Chr.5: 135895780-135895780 on Build GRCh38
rs772132419 Chr.5: 135895527-135895527 on Build GRCh38
rs141211399 Chr.5: 135895554-135895554 on Build GRCh38
rs754661939 Chr.5: 135895485-135895485 on Build GRCh38
rs765180351 Chr.5: 135895462-135895462 on Build GRCh38
rs765698493 Chr.5: 135895689-135895689 on Build GRCh38
rs1001083456 Chr.5: 135895619-135895619 on Build GRCh38
rs2069875 Chr.5: 135895388-135895388 on Build GRCh38
rs780519648 Chr.5: 135895499-135895499 on Build GRCh38
rs779186550 Chr.5: 135895510-135895510 on Build GRCh38
rs760158998 Chr.5: 135895776-135895776 on Build GRCh38
rs370719672 Chr.5: 135895592-135895592 on Build GRCh38
rs746542871 Chr.5: 135895394-135895394 on Build GRCh38
rs2066758 Chr.5: 135895506-135895506 on Build GRCh38
rs763624650 Chr.5: 135895585-135895585 on Build GRCh38
rs113288599 Chr.5: 135895536-135895536 on Build GRCh38
rs749883097 Chr.5: 135895683-135895683 on Build GRCh38
rs748418233 Chr.5: 135895601-135895601 on Build GRCh38
rs759613732 Chr.5: 135895544-135895544 on Build GRCh38
rs770151653 Chr.5: 135895402-135895402 on Build GRCh38
rs757930597 Chr.5: 135895464-135895464 on Build GRCh38
rs748554426 Chr.5: 135895525-135895525 on Build GRCh38
rs368855253 Chr.5: 135895432-135895432 on Build GRCh38
rs1055168966 Chr.5: 135895377-135895377 on Build GRCh38
rs770041351 Chr.5: 135895757-135895757 on Build GRCh38
rs767173791 Chr.5: 135895678-135895678 on Build GRCh38
rs764010324 Chr.5: 135895657-135895657 on Build GRCh38
rs771129561 Chr.5: 135895625-135895625 on Build GRCh38
rs564739256 Chr.5: 135895466-135895466 on Build GRCh38
rs769060925 Chr.5: 135895424-135895424 on Build GRCh38
rs2066760 Chr.5: 135895458-135895458 on Build GRCh38
rs760637041 Chr.5: 135895531-135895531 on Build GRCh38
rs530922688 Chr.5: 135895353-135895353 on Build GRCh38
rs923834612 Chr.5: 135895749-135895749 on Build GRCh38
rs779411427 Chr.5: 135895600-135895600 on Build GRCh38
rs775274460 Chr.5: 135895759-135895759 on Build GRCh38
rs749944547 Chr.5: 135895597-135895597 on Build GRCh38
rs765049151 Chr.5: 135895559-135895569 on Build GRCh38
rs908954426 Chr.5: 135895344-135895344 on Build GRCh38
rs949285839 Chr.5: 135895419-135895419 on Build GRCh38
rs942538803 Chr.5: 135895343-135895343 on Build GRCh38
rs765270121 Chr.5: 135895547-135895547 on Build GRCh38
rs774545436 Chr.5: 135895430-135895430 on Build GRCh38
rs565684626 Chr.5: 135895590-135895590 on Build GRCh38
rs370164592 Chr.5: 135895630-135895630 on Build GRCh38
rs986903538 Chr.5: 135895679-135895679 on Build GRCh38
rs868460439 Chr.5: 135895548-135895548 on Build GRCh38
rs762635010 Chr.5: 135895654-135895654 on Build GRCh38
rs201454539 Chr.5: 135895693-135895693 on Build GRCh38
rs758137112 Chr.5: 135895703-135895716 on Build GRCh38
rs776784578 Chr.5: 135895626-135895626 on Build GRCh38
rs777714256 Chr.5: 135895755-135895755 on Build GRCh38
rs777834314 Chr.5: 135895611-135895611 on Build GRCh38
rs747339286 Chr.5: 135895740-135895740 on Build GRCh38
rs762839156 Chr.5: 135895560-135895560 on Build GRCh38
rs975248428 Chr.5: 135895754-135895754 on Build GRCh38
rs747312051 Chr.5: 135895623-135895623 on Build GRCh38
rs776694647 Chr.5: 135895541-135895541 on Build GRCh38
rs569600530 Chr.5: 135895772-135895772 on Build GRCh38
rs147808011 Chr.5: 135895470-135895470 on Build GRCh38
rs1042325178 Chr.5: 135895421-135895421 on Build GRCh38
rs373713337 Chr.5: 135895700-135895700 on Build GRCh38
rs748036907 Chr.5: 135895750-135895750 on Build GRCh38
rs754796542 Chr.5: 135895457-135895457 on Build GRCh38
rs377764975 Chr.5: 135895484-135895484 on Build GRCh38
rs145258862 Chr.5: 135895577-135895577 on Build GRCh38
rs749624019 Chr.5: 135895411-135895411 on Build GRCh38
rs753863759 Chr.5: 135895442-135895442 on Build GRCh38
rs772895208 Chr.5: 135895773-135895773 on Build GRCh38
rs749855054 Chr.5: 135895502-135895502 on Build GRCh38
rs749311227 Chr.5: 135895765-135895765 on Build GRCh38
rs755648595 Chr.5: 135895688-135895688 on Build GRCh38
rs373038396 Chr.5: 135895436-135895436 on Build GRCh38
rs780750822 Chr.5: 135895593-135895593 on Build GRCh38
rs1000686176 Chr.5: 135895439-135895439 on Build GRCh38
rs368804746 Chr.5: 135895742-135895742 on Build GRCh38
rs193148109 Chr.5: 135895607-135895607 on Build GRCh38
rs373246078 Chr.5: 135895530-135895530 on Build GRCh38
rs753445242 Chr.5: 135895397-135895397 on Build GRCh38
rs758674734 Chr.5: 135895727-135895727 on Build GRCh38
rs563683408 Chr.5: 135895352-135895352 on Build GRCh38
rs949073096 Chr.5: 135895692-135895692 on Build GRCh38
rs751170097 Chr.5: 135895659-135895659 on Build GRCh38
rs964156835 Chr.5: 135895720-135895720 on Build GRCh38
rs778265553 Chr.5: 135895728-135895728 on Build GRCh38
rs937709007 Chr.5: 135895347-135895347 on Build GRCh38
rs761465688 Chr.5: 135895662-135895662 on Build GRCh38
rs766184152 Chr.5: 135895440-135895440 on Build GRCh38

View Full Product Details