Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 137753968-137754474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPA0
Entrez Gene ID
10949
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Gene Aliases
HNRPA0
UniGene
Hs.645902
Gene Chromosome Location
Chr.5: 137751384-137754350 on Build GRCh38
Transcripts
NM_006805.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587998 c.22A>G p.K8E Chr.5: 137754045-137754045 on Build GRCh38
5525056 c.66C>T p.G22G Chr.5: 137754001-137754001 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886751428 Chr.5: 137754239-137754239 on Build GRCh38
rs757908158 Chr.5: 137754002-137754002 on Build GRCh38
rs761908057 Chr.5: 137754350-137754350 on Build GRCh38
rs933580982 Chr.5: 137754231-137754231 on Build GRCh38
rs989055168 Chr.5: 137754397-137754397 on Build GRCh38
rs997054881 Chr.5: 137754314-137754315 on Build GRCh38
rs907456860 Chr.5: 137754108-137754108 on Build GRCh38
rs1051226447 Chr.5: 137754232-137754232 on Build GRCh38
rs765071214 Chr.5: 137754095-137754095 on Build GRCh38
rs938644789 Chr.5: 137754178-137754178 on Build GRCh38
rs557925987 Chr.5: 137754325-137754325 on Build GRCh38
rs939786157 Chr.5: 137754243-137754243 on Build GRCh38
rs1021680316 Chr.5: 137754331-137754331 on Build GRCh38
rs373962496 Chr.5: 137754226-137754226 on Build GRCh38
rs772080943 Chr.5: 137754068-137754068 on Build GRCh38
rs759271902 Chr.5: 137754087-137754087 on Build GRCh38
rs982690621 Chr.5: 137754199-137754199 on Build GRCh38
rs900602789 Chr.5: 137754313-137754313 on Build GRCh38
rs955148589 Chr.5: 137754343-137754343 on Build GRCh38
rs766411737 Chr.5: 137754405-137754405 on Build GRCh38
rs770404007 Chr.5: 137754013-137754013 on Build GRCh38
rs908224007 Chr.5: 137754361-137754361 on Build GRCh38
rs948482812 Chr.5: 137754393-137754393 on Build GRCh38
rs975350256 Chr.5: 137754339-137754339 on Build GRCh38
rs891804871 Chr.5: 137754413-137754413 on Build GRCh38
rs975026242 Chr.5: 137754204-137754204 on Build GRCh38
rs540025785 Chr.5: 137754320-137754320 on Build GRCh38
rs751288737 Chr.5: 137754326-137754327 on Build GRCh38
rs187197502 Chr.5: 137754198-137754198 on Build GRCh38
rs914758822 Chr.5: 137754194-137754194 on Build GRCh38
rs1053123278 Chr.5: 137754141-137754141 on Build GRCh38
rs907013867 Chr.5: 137754401-137754401 on Build GRCh38
rs965187534 Chr.5: 137754434-137754434 on Build GRCh38
rs971333483 Chr.5: 137754097-137754097 on Build GRCh38
rs535450874 Chr.5: 137754183-137754183 on Build GRCh38
rs1038049965 Chr.5: 137754255-137754255 on Build GRCh38
rs199553659 Chr.5: 137754011-137754011 on Build GRCh38
rs999240554 Chr.5: 137754113-137754113 on Build GRCh38
rs772861651 Chr.5: 137754450-137754450 on Build GRCh38
rs566240432 Chr.5: 137754003-137754003 on Build GRCh38
rs915968020 Chr.5: 137754253-137754253 on Build GRCh38
rs955997508 Chr.5: 137754314-137754314 on Build GRCh38
rs201904905 Chr.5: 137753995-137753995 on Build GRCh38
rs776417931 Chr.5: 137754086-137754086 on Build GRCh38
rs370608061 Chr.5: 137754028-137754028 on Build GRCh38
rs900528656 Chr.5: 137754428-137754428 on Build GRCh38
rs370328533 Chr.5: 137754082-137754082 on Build GRCh38
rs773315447 Chr.5: 137754073-137754073 on Build GRCh38
rs1044646238 Chr.5: 137754395-137754395 on Build GRCh38
rs1029969858 Chr.5: 137754184-137754184 on Build GRCh38
rs893171813 Chr.5: 137754143-137754143 on Build GRCh38
rs1046372719 Chr.5: 137754090-137754090 on Build GRCh38
rs890975990 Chr.5: 137754315-137754317 on Build GRCh38
rs928853722 Chr.5: 137754260-137754260 on Build GRCh38
rs1021628942 Chr.5: 137754431-137754431 on Build GRCh38
rs947345648 Chr.5: 137754402-137754402 on Build GRCh38
rs762929999 Chr.5: 137754266-137754266 on Build GRCh38
rs948862990 Chr.5: 137754236-137754236 on Build GRCh38
rs962390859 Chr.5: 137754221-137754221 on Build GRCh38
rs1052992190 Chr.5: 137754412-137754412 on Build GRCh38
rs956134489 Chr.5: 137754156-137754156 on Build GRCh38
rs189655296 Chr.5: 137754268-137754268 on Build GRCh38
rs1002777137 Chr.5: 137754306-137754306 on Build GRCh38
rs947403906 Chr.5: 137754282-137754282 on Build GRCh38
rs1031697414 Chr.5: 137754308-137754308 on Build GRCh38
rs866567566 Chr.5: 137754347-137754347 on Build GRCh38
rs981653311 Chr.5: 137754242-137754242 on Build GRCh38
rs115184876 Chr.5: 137754265-137754265 on Build GRCh38
rs374604487 Chr.5: 137754089-137754089 on Build GRCh38
rs1011623898 Chr.5: 137754427-137754427 on Build GRCh38
rs757424005 Chr.5: 137754313-137754314 on Build GRCh38
rs201588081 Chr.5: 137754098-137754098 on Build GRCh38
rs777333191 Chr.5: 137754010-137754010 on Build GRCh38
rs907141179 Chr.5: 137754280-137754280 on Build GRCh38
rs368179259 Chr.5: 137754348-137754348 on Build GRCh38
rs879452751 Chr.5: 137754135-137754135 on Build GRCh38
rs780589922 Chr.5: 137754016-137754016 on Build GRCh38
rs1031661448 Chr.5: 137754152-137754152 on Build GRCh38
rs182466584 Chr.5: 137754385-137754385 on Build GRCh38
rs751170494 Chr.5: 137754410-137754410 on Build GRCh38
rs773308311 Chr.5: 137754218-137754218 on Build GRCh38
rs746746294 Chr.5: 137754040-137754040 on Build GRCh38
rs1015496772 Chr.5: 137754333-137754333 on Build GRCh38
rs752269159 Chr.5: 137754001-137754001 on Build GRCh38
rs933400520 Chr.5: 137754444-137754444 on Build GRCh38
rs764771431 Chr.5: 137754354-137754354 on Build GRCh38
rs769011730 Chr.5: 137754034-137754034 on Build GRCh38
rs534204658 Chr.5: 137754365-137754365 on Build GRCh38
rs1037996008 Chr.5: 137754367-137754367 on Build GRCh38
rs923444744 Chr.5: 137754340-137754340 on Build GRCh38
rs1017742168 Chr.5: 137754158-137754158 on Build GRCh38
rs758037608 Chr.5: 137754104-137754104 on Build GRCh38
rs760625108 Chr.5: 137754078-137754078 on Build GRCh38
rs142227807 Chr.5: 137754037-137754037 on Build GRCh38
rs568423134 Chr.5: 137754176-137754176 on Build GRCh38
rs768542554 Chr.5: 137754042-137754042 on Build GRCh38
rs1039098194 Chr.5: 137754301-137754301 on Build GRCh38
rs573597137 Chr.5: 137754419-137754419 on Build GRCh38
rs964789185 Chr.5: 137754337-137754337 on Build GRCh38

View Full Product Details