Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140647507-140648011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IK
Entrez Gene ID
3550
Gene Name
IK cytokine, down-regulator of HLA II
Gene Aliases
CSA2, RED, RER
UniGene
Hs.421245
Gene Chromosome Location
Chr.5: 140647799-140662480 on Build GRCh38
Transcripts
NM_006083.3 NR_037428.1 NM_001185012.1 NM_002488.4 NR_033697.1

Gene Symbol
MIR3655
Entrez Gene ID
100500820
Gene Name
microRNA 3655
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.5: 140647844-140647926 on Build GRCh38
Transcripts
NM_006083.3 NR_037428.1 NM_001185012.1 NM_002488.4 NR_033697.1

Gene Symbol
NDUFA2
Entrez Gene ID
4695
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Gene Aliases
B8, CD14, CIB8
UniGene
Hs.75914
Gene Chromosome Location
Chr.5: 140645363-140647785 on Build GRCh38
Transcripts
NM_006083.3 NR_037428.1 NM_001185012.1 NM_002488.4 NR_033697.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1061731 c.8C>T p.A3V Chr.5: 140647576-140647576 on Build GRCh38
3850687 c.63C>T p.I21I Chr.5: 140647521-140647521 on Build GRCh38
5000544 c.5C>T p.P2L Chr.5: 140647913-140647913 on Build GRCh38
6233399 c.10C>A p.R4R Chr.5: 140647918-140647918 on Build GRCh38
1579450 c.74A>T p.Q25L Chr.5: 140647510-140647510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773844456 Chr.5: 140647541-140647541 on Build GRCh38
rs372239752 Chr.5: 140647600-140647600 on Build GRCh38
rs770245226 Chr.5: 140647594-140647594 on Build GRCh38
rs374534339 Chr.5: 140647899-140647899 on Build GRCh38
rs940767479 Chr.5: 140647886-140647886 on Build GRCh38
rs761616160 Chr.5: 140647778-140647778 on Build GRCh38
rs779534000 Chr.5: 140647835-140647835 on Build GRCh38
rs775426500 Chr.5: 140647957-140647957 on Build GRCh38
rs759580754 Chr.5: 140647801-140647801 on Build GRCh38
rs755127827 Chr.5: 140647639-140647639 on Build GRCh38
rs770192248 Chr.5: 140647648-140647648 on Build GRCh38
rs748188529 Chr.5: 140647930-140647930 on Build GRCh38
rs370015250 Chr.5: 140647574-140647574 on Build GRCh38
rs1022558681 Chr.5: 140647552-140647552 on Build GRCh38
rs748517973 Chr.5: 140647590-140647590 on Build GRCh38
rs747265872 Chr.5: 140647855-140647855 on Build GRCh38
rs781422758 Chr.5: 140647642-140647642 on Build GRCh38
rs747859371 Chr.5: 140647822-140647822 on Build GRCh38
rs535080968 Chr.5: 140647875-140647875 on Build GRCh38
rs776923081 Chr.5: 140647589-140647589 on Build GRCh38
rs756466349 Chr.5: 140647741-140647741 on Build GRCh38
rs746448356 Chr.5: 140647836-140647836 on Build GRCh38
rs752956448 Chr.5: 140647740-140647740 on Build GRCh38
rs763847679 Chr.5: 140647625-140647625 on Build GRCh38
rs370832670 Chr.5: 140647947-140647947 on Build GRCh38
rs760643148 Chr.5: 140647961-140647961 on Build GRCh38
rs748460855 Chr.5: 140647646-140647646 on Build GRCh38
rs778058720 Chr.5: 140647828-140647828 on Build GRCh38
rs956375600 Chr.5: 140647578-140647578 on Build GRCh38
rs368962559 Chr.5: 140647838-140647838 on Build GRCh38
rs369495222 Chr.5: 140647626-140647626 on Build GRCh38
rs201961300 Chr.5: 140647891-140647891 on Build GRCh38
rs755697876 Chr.5: 140647914-140647914 on Build GRCh38
rs892339213 Chr.5: 140647950-140647950 on Build GRCh38
rs759062130 Chr.5: 140647606-140647606 on Build GRCh38
rs761673298 Chr.5: 140647710-140647710 on Build GRCh38
rs371585072 Chr.5: 140647887-140647887 on Build GRCh38
rs777258491 Chr.5: 140647920-140647920 on Build GRCh38
rs760458290 Chr.5: 140647623-140647623 on Build GRCh38
rs769500961 Chr.5: 140647791-140647791 on Build GRCh38
rs947633020 Chr.5: 140647609-140647609 on Build GRCh38
rs775477746 Chr.5: 140647874-140647874 on Build GRCh38
rs766346615 Chr.5: 140647730-140647730 on Build GRCh38
rs553111700 Chr.5: 140647890-140647890 on Build GRCh38
rs752340193 Chr.5: 140647545-140647545 on Build GRCh38
rs143857164 Chr.5: 140647633-140647633 on Build GRCh38
rs780739269 Chr.5: 140647905-140647905 on Build GRCh38
rs377685148 Chr.5: 140647937-140647937 on Build GRCh38
rs747127939 Chr.5: 140647636-140647636 on Build GRCh38
rs752899359 Chr.5: 140647814-140647814 on Build GRCh38
rs967712893 Chr.5: 140647808-140647808 on Build GRCh38
rs746497677 Chr.5: 140647756-140647756 on Build GRCh38
rs768376691 Chr.5: 140647704-140647704 on Build GRCh38
rs35672074 Chr.5: 140647544-140647544 on Build GRCh38
rs779028375 Chr.5: 140647565-140647565 on Build GRCh38
rs763621450 Chr.5: 140647610-140647610 on Build GRCh38
rs768325530 Chr.5: 140647770-140647770 on Build GRCh38
rs765747363 Chr.5: 140647539-140647539 on Build GRCh38
rs1003805017 Chr.5: 140647733-140647733 on Build GRCh38
rs769452041 Chr.5: 140647563-140647565 on Build GRCh38
rs1036761169 Chr.5: 140647687-140647687 on Build GRCh38
rs766177915 Chr.5: 140647796-140647796 on Build GRCh38
rs940665723 Chr.5: 140647652-140647652 on Build GRCh38
rs913453092 Chr.5: 140647596-140647596 on Build GRCh38
rs765011689 Chr.5: 140647712-140647712 on Build GRCh38
rs755070637 Chr.5: 140647735-140647735 on Build GRCh38
rs886060016 Chr.5: 140647634-140647634 on Build GRCh38
rs751600651 Chr.5: 140647732-140647732 on Build GRCh38
rs905111941 Chr.5: 140647924-140647924 on Build GRCh38
rs751299721 Chr.5: 140647604-140647604 on Build GRCh38
rs757145018 Chr.5: 140647627-140647627 on Build GRCh38
rs760264090 Chr.5: 140647697-140647697 on Build GRCh38
rs773736551 Chr.5: 140647654-140647654 on Build GRCh38
rs377007416 Chr.5: 140647871-140647871 on Build GRCh38
rs375301523 Chr.5: 140647978-140647978 on Build GRCh38
rs754133084 Chr.5: 140647878-140647878 on Build GRCh38
rs185280379 Chr.5: 140647831-140647831 on Build GRCh38
rs747563860 Chr.5: 140647910-140647910 on Build GRCh38
rs376581584 Chr.5: 140647616-140647616 on Build GRCh38
rs1044206055 Chr.5: 140647635-140647635 on Build GRCh38
rs780294618 Chr.5: 140647581-140647581 on Build GRCh38
rs886460495 Chr.5: 140647661-140647661 on Build GRCh38
rs996387742 Chr.5: 140647721-140647721 on Build GRCh38
rs374432948 Chr.5: 140647624-140647624 on Build GRCh38
rs139482464 Chr.5: 140647863-140647863 on Build GRCh38
rs867446887 Chr.5: 140647781-140647781 on Build GRCh38
rs779878149 Chr.5: 140647664-140647702 on Build GRCh38
rs747263398 Chr.5: 140647584-140647584 on Build GRCh38
rs767649892 Chr.5: 140647805-140647805 on Build GRCh38
rs146400503 Chr.5: 140647850-140647850 on Build GRCh38
rs771273606 Chr.5: 140647753-140647753 on Build GRCh38
rs992864035 Chr.5: 140647832-140647832 on Build GRCh38
rs111226907 Chr.5: 140647839-140647839 on Build GRCh38
rs776474256 Chr.5: 140647709-140647709 on Build GRCh38
rs778593 Chr.5: 140647631-140647631 on Build GRCh38
rs775201684 Chr.5: 140647622-140647622 on Build GRCh38
rs988215320 Chr.5: 140647588-140647588 on Build GRCh38
rs1022465563 Chr.5: 140647734-140647734 on Build GRCh38
rs774801934 Chr.5: 140647682-140647682 on Build GRCh38
rs770798087 Chr.5: 140647861-140647861 on Build GRCh38
rs773615023 Chr.5: 140647595-140647595 on Build GRCh38
rs774071371 Chr.5: 140647931-140647931 on Build GRCh38
rs751545213 Chr.5: 140647800-140647800 on Build GRCh38
rs764303843 Chr.5: 140647971-140647971 on Build GRCh38
rs778054089 Chr.5: 140647752-140647752 on Build GRCh38
rs749013145 Chr.5: 140647857-140647857 on Build GRCh38
rs778594 Chr.5: 140647719-140647719 on Build GRCh38
rs900623800 Chr.5: 140647959-140647959 on Build GRCh38
rs757200194 Chr.5: 140647554-140647554 on Build GRCh38
rs759527321 Chr.5: 140647866-140647866 on Build GRCh38
rs539275827 Chr.5: 140647898-140647898 on Build GRCh38
rs987943600 Chr.5: 140647758-140647758 on Build GRCh38
rs530439525 Chr.5: 140647772-140647772 on Build GRCh38
rs765201873 Chr.5: 140647628-140647628 on Build GRCh38
rs755904736 Chr.5: 140647548-140647548 on Build GRCh38
rs747666614 Chr.5: 140647843-140647843 on Build GRCh38
rs745915609 Chr.5: 140647567-140647567 on Build GRCh38
rs746278030 Chr.5: 140647795-140647795 on Build GRCh38
rs886060017 Chr.5: 140647728-140647728 on Build GRCh38
rs113236159 Chr.5: 140647751-140647751 on Build GRCh38
rs200203867 Chr.5: 140647689-140647689 on Build GRCh38
rs546286895 Chr.5: 140647681-140647681 on Build GRCh38
rs772818096 Chr.5: 140647793-140647793 on Build GRCh38
rs768635352 Chr.5: 140647959-140647967 on Build GRCh38
rs999798095 Chr.5: 140647754-140647754 on Build GRCh38
rs756228978 Chr.5: 140647820-140647820 on Build GRCh38
rs535439456 Chr.5: 140647984-140647984 on Build GRCh38
rs749824414 Chr.5: 140647664-140647664 on Build GRCh38
rs749036471 Chr.5: 140647926-140647926 on Build GRCh38
rs375905956 Chr.5: 140647553-140647553 on Build GRCh38

View Full Product Details