Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140657437-140657899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IK
Entrez Gene ID
3550
Gene Name
IK cytokine, down-regulator of HLA II
Gene Aliases
CSA2, RED, RER
UniGene
Hs.421245
Gene Chromosome Location
Chr.5: 140647799-140662480 on Build GRCh38
Transcripts
NM_006083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5867442 c.875C>T p.T292I Chr.5: 140657630-140657630 on Build GRCh38
1186849 c.887A>G p.K296R Chr.5: 140657639-140657639 on Build GRCh38
5219037 c.864G>T p.L288L Chr.5: 140657619-140657619 on Build GRCh38
3393176 c.815T>G p.L272R Chr.5: 140657567-140657567 on Build GRCh38
5867441 c.878C>T p.T293I Chr.5: 140657630-140657630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007724119 Chr.5: 140657494-140657494 on Build GRCh38
rs764840216 Chr.5: 140657613-140657613 on Build GRCh38
rs911263007 Chr.5: 140657838-140657838 on Build GRCh38
rs375224727 Chr.5: 140657566-140657566 on Build GRCh38
rs781303354 Chr.5: 140657674-140657674 on Build GRCh38
rs377508460 Chr.5: 140657723-140657723 on Build GRCh38
rs538253015 Chr.5: 140657726-140657726 on Build GRCh38
rs191528016 Chr.5: 140657870-140657870 on Build GRCh38
rs918054035 Chr.5: 140657708-140657708 on Build GRCh38
rs745985445 Chr.5: 140657746-140657746 on Build GRCh38
rs754983526 Chr.5: 140657535-140657535 on Build GRCh38
rs754930426 Chr.5: 140657672-140657672 on Build GRCh38
rs748271757 Chr.5: 140657543-140657543 on Build GRCh38
rs771263392 Chr.5: 140657556-140657556 on Build GRCh38
rs531516469 Chr.5: 140657685-140657685 on Build GRCh38
rs6888409 Chr.5: 140657875-140657875 on Build GRCh38
rs901040530 Chr.5: 140657505-140657505 on Build GRCh38
rs937995304 Chr.5: 140657760-140657760 on Build GRCh38
rs746261321 Chr.5: 140657689-140657689 on Build GRCh38
rs978559642 Chr.5: 140657794-140657794 on Build GRCh38
rs761334132 Chr.5: 140657568-140657568 on Build GRCh38
rs145110375 Chr.5: 140657704-140657704 on Build GRCh38
rs750112203 Chr.5: 140657631-140657631 on Build GRCh38
rs202075814 Chr.5: 140657517-140657517 on Build GRCh38
rs758198998 Chr.5: 140657632-140657632 on Build GRCh38
rs1040894153 Chr.5: 140657466-140657466 on Build GRCh38
rs371984526 Chr.5: 140657552-140657552 on Build GRCh38
rs770095389 Chr.5: 140657544-140657544 on Build GRCh38
rs774905279 Chr.5: 140657560-140657560 on Build GRCh38
rs73271555 Chr.5: 140657864-140657864 on Build GRCh38
rs749631304 Chr.5: 140657555-140657555 on Build GRCh38
rs376819795 Chr.5: 140657530-140657530 on Build GRCh38
rs371474850 Chr.5: 140657601-140657601 on Build GRCh38
rs772313333 Chr.5: 140657858-140657858 on Build GRCh38
rs555342709 Chr.5: 140657768-140657768 on Build GRCh38
rs924064056 Chr.5: 140657798-140657798 on Build GRCh38
rs1053767254 Chr.5: 140657822-140657822 on Build GRCh38
rs549217846 Chr.5: 140657565-140657565 on Build GRCh38
rs149950597 Chr.5: 140657681-140657681 on Build GRCh38
rs936778828 Chr.5: 140657804-140657804 on Build GRCh38
rs778011691 Chr.5: 140657644-140657644 on Build GRCh38
rs781358456 Chr.5: 140657537-140657537 on Build GRCh38
rs776019260 Chr.5: 140657705-140657705 on Build GRCh38
rs761309890 Chr.5: 140657716-140657716 on Build GRCh38
rs778596 Chr.5: 140657687-140657687 on Build GRCh38
rs761859061 Chr.5: 140657874-140657874 on Build GRCh38
rs758169749 Chr.5: 140657504-140657504 on Build GRCh38
rs771196772 Chr.5: 140657688-140657688 on Build GRCh38
rs553313510 Chr.5: 140657766-140657766 on Build GRCh38
rs751377977 Chr.5: 140657654-140657654 on Build GRCh38

View Full Product Details