Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 145866410-145866917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRXCR2
Entrez Gene ID
643226
Gene Name
glutaredoxin and cysteine rich domain containing 2
Gene Aliases
DFNB101
UniGene
Hs.570895
Gene Chromosome Location
Chr.5: 145858538-145931673 on Build GRCh38
Transcripts
NM_001080516.1 XM_017009708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
275396 c.397C>T p.R133* Chr.5: 145866668-145866668 on Build GRCh38
4453957 c.376A>C p.T126P Chr.5: 145866689-145866689 on Build GRCh38
4159597 c.479A>C p.K160T Chr.5: 145866586-145866586 on Build GRCh38
4141529 c.537C>T p.S179S Chr.5: 145866528-145866528 on Build GRCh38
3852396 c.419A>G p.D140G Chr.5: 145866646-145866646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887955648 Chr.5: 145866854-145866854 on Build GRCh38
rs747417642 Chr.5: 145866710-145866710 on Build GRCh38
rs757534438 Chr.5: 145866529-145866529 on Build GRCh38
rs761197305 Chr.5: 145866616-145866616 on Build GRCh38
rs374891281 Chr.5: 145866723-145866723 on Build GRCh38
rs374097883 Chr.5: 145866455-145866455 on Build GRCh38
rs368569020 Chr.5: 145866497-145866497 on Build GRCh38
rs770597524 Chr.5: 145866574-145866574 on Build GRCh38
rs539502019 Chr.5: 145866742-145866742 on Build GRCh38
rs775921491 Chr.5: 145866612-145866612 on Build GRCh38
rs756150239 Chr.5: 145866661-145866661 on Build GRCh38
rs761305184 Chr.5: 145866508-145866508 on Build GRCh38
rs1041061849 Chr.5: 145866594-145866594 on Build GRCh38
rs765480071 Chr.5: 145866644-145866644 on Build GRCh38
rs754314197 Chr.5: 145866716-145866716 on Build GRCh38
rs756734443 Chr.5: 145866767-145866767 on Build GRCh38
rs767999715 Chr.5: 145866512-145866512 on Build GRCh38
rs201457352 Chr.5: 145866748-145866748 on Build GRCh38
rs368301231 Chr.5: 145866456-145866456 on Build GRCh38
rs930767233 Chr.5: 145866541-145866541 on Build GRCh38
rs750478745 Chr.5: 145866846-145866846 on Build GRCh38
rs779925305 Chr.5: 145866472-145866472 on Build GRCh38
rs752630207 Chr.5: 145866660-145866660 on Build GRCh38
rs771151044 Chr.5: 145866506-145866506 on Build GRCh38
rs952440970 Chr.5: 145866785-145866785 on Build GRCh38
rs570479885 Chr.5: 145866722-145866722 on Build GRCh38
rs1049686035 Chr.5: 145866848-145866848 on Build GRCh38
rs755627545 Chr.5: 145866559-145866559 on Build GRCh38
rs368175587 Chr.5: 145866463-145866463 on Build GRCh38
rs753377119 Chr.5: 145866667-145866667 on Build GRCh38
rs750674789 Chr.5: 145866649-145866649 on Build GRCh38
rs772431701 Chr.5: 145866600-145866600 on Build GRCh38
rs776738076 Chr.5: 145866637-145866637 on Build GRCh38
rs370725478 Chr.5: 145866453-145866453 on Build GRCh38
rs372401844 Chr.5: 145866511-145866511 on Build GRCh38
rs940917921 Chr.5: 145866864-145866864 on Build GRCh38
rs748981520 Chr.5: 145866503-145866503 on Build GRCh38
rs767433552 Chr.5: 145866659-145866659 on Build GRCh38
rs772381799 Chr.5: 145866690-145866690 on Build GRCh38
rs765385394 Chr.5: 145866789-145866789 on Build GRCh38
rs200657078 Chr.5: 145866471-145866471 on Build GRCh38
rs745536409 Chr.5: 145866681-145866681 on Build GRCh38
rs114615413 Chr.5: 145866555-145866555 on Build GRCh38
rs528834976 Chr.5: 145866517-145866517 on Build GRCh38
rs80137097 Chr.5: 145866772-145866772 on Build GRCh38
rs2569006 Chr.5: 145866522-145866522 on Build GRCh38
rs368999309 Chr.5: 145866752-145866752 on Build GRCh38
rs767378623 Chr.5: 145866746-145866746 on Build GRCh38
rs777060343 Chr.5: 145866715-145866715 on Build GRCh38
rs762039896 Chr.5: 145866732-145866732 on Build GRCh38
rs764090284 Chr.5: 145866751-145866751 on Build GRCh38
rs372492111 Chr.5: 145866768-145866768 on Build GRCh38
rs982566857 Chr.5: 145866786-145866786 on Build GRCh38
rs756784401 Chr.5: 145866668-145866668 on Build GRCh38
rs765415094 Chr.5: 145866531-145866531 on Build GRCh38
rs751345210 Chr.5: 145866537-145866537 on Build GRCh38
rs953157430 Chr.5: 145866650-145866650 on Build GRCh38
rs989213073 Chr.5: 145866459-145866459 on Build GRCh38
rs759337723 Chr.5: 145866658-145866658 on Build GRCh38
rs566124751 Chr.5: 145866795-145866795 on Build GRCh38
rs375844725 Chr.5: 145866633-145866633 on Build GRCh38
rs149954479 Chr.5: 145866525-145866525 on Build GRCh38
rs759306920 Chr.5: 145866600-145866601 on Build GRCh38
rs748851479 Chr.5: 145866567-145866567 on Build GRCh38
rs769764331 Chr.5: 145866721-145866721 on Build GRCh38
rs372393420 Chr.5: 145866464-145866464 on Build GRCh38
rs546249529 Chr.5: 145866495-145866495 on Build GRCh38
rs183701679 Chr.5: 145866475-145866475 on Build GRCh38
rs200072851 Chr.5: 145866745-145866745 on Build GRCh38
rs144587935 Chr.5: 145866712-145866712 on Build GRCh38
rs748100037 Chr.5: 145866557-145866557 on Build GRCh38
rs979234447 Chr.5: 145866664-145866664 on Build GRCh38
rs144617184 Chr.5: 145866639-145866639 on Build GRCh38
rs867529802 Chr.5: 145866620-145866620 on Build GRCh38
rs777426348 Chr.5: 145866676-145866676 on Build GRCh38
rs776292091 Chr.5: 145866542-145866542 on Build GRCh38
rs559768717 Chr.5: 145866496-145866496 on Build GRCh38
rs780429994 Chr.5: 145866698-145866698 on Build GRCh38
rs551121997 Chr.5: 145866680-145866680 on Build GRCh38
rs777621955 Chr.5: 145866666-145866666 on Build GRCh38
rs754874283 Chr.5: 145866554-145866554 on Build GRCh38
rs140713353 Chr.5: 145866588-145866588 on Build GRCh38
rs760684583 Chr.5: 145866518-145866518 on Build GRCh38
rs754911421 Chr.5: 145866485-145866485 on Build GRCh38
rs908083649 Chr.5: 145866565-145866565 on Build GRCh38
rs760605874 Chr.5: 145866750-145866750 on Build GRCh38
rs777445349 Chr.5: 145866563-145866563 on Build GRCh38
rs778489752 Chr.5: 145866585-145866585 on Build GRCh38

View Full Product Details