Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150895809-150896310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRGM
Entrez Gene ID
345611
Gene Name
immunity related GTPase M
Gene Aliases
IFI1, IRGM1, LRG-47, LRG47
UniGene
Hs.519680
Gene Chromosome Location
Chr.5: 150846523-150902402 on Build GRCh38
Transcripts
XM_011537641.2 NM_001172831.1 NM_001172832.1 NM_052860.2 XM_011537702.2

Gene Symbol
ZNF300
Entrez Gene ID
91975
Gene Name
zinc finger protein 300
Gene Aliases
-
UniGene
Hs.134885
Gene Chromosome Location
Chr.5: 150892958-150904983 on Build GRCh38
Transcripts
XM_011537641.2 NM_001172831.1 NM_001172832.1 NM_052860.2 XM_011537702.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3612604 c.1188G>A p.L396L Chr.5: 150896051-150896051 on Build GRCh38
4783441 c.1193A>C p.E398A Chr.5: 150896094-150896094 on Build GRCh38
5356680 c.1185A>C p.K395N Chr.5: 150896102-150896102 on Build GRCh38
1435248 c.1145A>C p.E382A Chr.5: 150896094-150896094 on Build GRCh38
6102810 c.1097A>T p.E366V Chr.5: 150896190-150896190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779775626 Chr.5: 150895850-150895850 on Build GRCh38
rs755650345 Chr.5: 150896228-150896228 on Build GRCh38
rs867406447 Chr.5: 150896170-150896170 on Build GRCh38
rs575599619 Chr.5: 150896079-150896079 on Build GRCh38
rs774340985 Chr.5: 150895944-150895944 on Build GRCh38
rs145326125 Chr.5: 150896042-150896042 on Build GRCh38
rs974592826 Chr.5: 150895851-150895851 on Build GRCh38
rs533898079 Chr.5: 150896201-150896201 on Build GRCh38
rs866815097 Chr.5: 150896121-150896121 on Build GRCh38
rs761818091 Chr.5: 150896189-150896189 on Build GRCh38
rs777193595 Chr.5: 150895849-150895850 on Build GRCh38
rs912873167 Chr.5: 150896161-150896161 on Build GRCh38
rs760627803 Chr.5: 150895952-150895952 on Build GRCh38
rs1044887445 Chr.5: 150896084-150896085 on Build GRCh38
rs866762535 Chr.5: 150896275-150896275 on Build GRCh38
rs371710157 Chr.5: 150896243-150896243 on Build GRCh38
rs776151574 Chr.5: 150896114-150896125 on Build GRCh38
rs763897048 Chr.5: 150895959-150895959 on Build GRCh38
rs761906020 Chr.5: 150896084-150896084 on Build GRCh38
rs1988688 Chr.5: 150896231-150896231 on Build GRCh38
rs191242853 Chr.5: 150895983-150895983 on Build GRCh38
rs754389152 Chr.5: 150896108-150896108 on Build GRCh38
rs752255527 Chr.5: 150896226-150896226 on Build GRCh38
rs866561860 Chr.5: 150896119-150896119 on Build GRCh38
rs922742238 Chr.5: 150896006-150896006 on Build GRCh38
rs763168249 Chr.5: 150895907-150895907 on Build GRCh38
rs924839781 Chr.5: 150896019-150896019 on Build GRCh38
rs761175973 Chr.5: 150896185-150896186 on Build GRCh38
rs141369580 Chr.5: 150896016-150896016 on Build GRCh38
rs779596151 Chr.5: 150896002-150896002 on Build GRCh38
rs771638678 Chr.5: 150895848-150895848 on Build GRCh38
rs769894357 Chr.5: 150896190-150896190 on Build GRCh38
rs765124998 Chr.5: 150895981-150895981 on Build GRCh38
rs17800771 Chr.5: 150896225-150896225 on Build GRCh38
rs371764879 Chr.5: 150896076-150896076 on Build GRCh38
rs778945218 Chr.5: 150896158-150896158 on Build GRCh38
rs751110636 Chr.5: 150896219-150896219 on Build GRCh38
rs894553548 Chr.5: 150896048-150896048 on Build GRCh38
rs769095101 Chr.5: 150895864-150895864 on Build GRCh38
rs376308554 Chr.5: 150895872-150895872 on Build GRCh38
rs369365423 Chr.5: 150896183-150896183 on Build GRCh38
rs148013564 Chr.5: 150896165-150896165 on Build GRCh38
rs748426240 Chr.5: 150895875-150895875 on Build GRCh38
rs566492867 Chr.5: 150895966-150895966 on Build GRCh38
rs977584194 Chr.5: 150896259-150896259 on Build GRCh38
rs765911691 Chr.5: 150896099-150896099 on Build GRCh38
rs774664117 Chr.5: 150896050-150896050 on Build GRCh38
rs756673248 Chr.5: 150896116-150896116 on Build GRCh38
rs199921485 Chr.5: 150896215-150896215 on Build GRCh38
rs770167284 Chr.5: 150895889-150895889 on Build GRCh38
rs1003311736 Chr.5: 150896111-150896111 on Build GRCh38
rs760116918 Chr.5: 150895996-150895997 on Build GRCh38
rs557353484 Chr.5: 150896053-150896053 on Build GRCh38
rs372607730 Chr.5: 150895845-150895845 on Build GRCh38
rs765108536 Chr.5: 150896085-150896085 on Build GRCh38
rs868415176 Chr.5: 150896253-150896253 on Build GRCh38
rs987549582 Chr.5: 150896037-150896037 on Build GRCh38
rs746103177 Chr.5: 150896164-150896164 on Build GRCh38
rs754351765 Chr.5: 150896255-150896255 on Build GRCh38
rs770077751 Chr.5: 150896029-150896029 on Build GRCh38
rs759303549 Chr.5: 150896221-150896221 on Build GRCh38
rs780758598 Chr.5: 150896112-150896112 on Build GRCh38
rs757057143 Chr.5: 150895971-150895971 on Build GRCh38
rs141611335 Chr.5: 150896202-150896202 on Build GRCh38
rs561426657 Chr.5: 150896140-150896140 on Build GRCh38
rs754611688 Chr.5: 150896132-150896132 on Build GRCh38
rs751251277 Chr.5: 150896107-150896107 on Build GRCh38
rs921715774 Chr.5: 150895986-150895986 on Build GRCh38
rs542704110 Chr.5: 150896080-150896080 on Build GRCh38
rs558818519 Chr.5: 150896003-150896003 on Build GRCh38
rs765768381 Chr.5: 150896101-150896102 on Build GRCh38
rs758181226 Chr.5: 150895991-150895991 on Build GRCh38
rs773366649 Chr.5: 150895905-150895905 on Build GRCh38
rs554652530 Chr.5: 150895916-150895916 on Build GRCh38
rs773634665 Chr.5: 150896030-150896030 on Build GRCh38
rs865897313 Chr.5: 150896257-150896257 on Build GRCh38
rs866964851 Chr.5: 150896218-150896218 on Build GRCh38
rs778078278 Chr.5: 150896127-150896127 on Build GRCh38
rs747540382 Chr.5: 150895854-150895854 on Build GRCh38
rs748650915 Chr.5: 150896114-150896114 on Build GRCh38
rs76474888 Chr.5: 150896015-150896015 on Build GRCh38
rs749612793 Chr.5: 150896137-150896137 on Build GRCh38
rs374986497 Chr.5: 150896090-150896090 on Build GRCh38
rs767082659 Chr.5: 150896212-150896213 on Build GRCh38
rs748667386 Chr.5: 150896024-150896024 on Build GRCh38
rs867532860 Chr.5: 150896203-150896203 on Build GRCh38

View Full Product Details