Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 153490434-153490929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIA1
Entrez Gene ID
2890
Gene Name
glutamate ionotropic receptor AMPA type subunit 1
Gene Aliases
GLUH1, GLUR1, GLURA, GluA1, HBGR1
UniGene
Hs.519693
Gene Chromosome Location
Chr.5: 153489615-153813873 on Build GRCh38
Transcripts
NM_000827.3 NM_001114183.1 NM_001258019.1 NM_001258020.1 NR_047578.1 XM_011537635.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6102744 c.22T>C p.F8L Chr.5: 153490910-153490910 on Build GRCh38
1496079 c.31G>T p.G11C Chr.5: 153490919-153490919 on Build GRCh38
1496078 c.31G>T p.G11C Chr.5: 153490919-153490919 on Build GRCh38
7156662 c.24C>T p.F8F Chr.5: 153490912-153490912 on Build GRCh38
6102745 c.22T>C p.F8L Chr.5: 153490910-153490910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144275743 Chr.5: 153490807-153490807 on Build GRCh38
rs879872646 Chr.5: 153490627-153490627 on Build GRCh38
rs897579909 Chr.5: 153490711-153490711 on Build GRCh38
rs771331045 Chr.5: 153490878-153490878 on Build GRCh38
rs559598738 Chr.5: 153490759-153490759 on Build GRCh38
rs772798293 Chr.5: 153490501-153490501 on Build GRCh38
rs949944083 Chr.5: 153490650-153490650 on Build GRCh38
rs768184274 Chr.5: 153490488-153490488 on Build GRCh38
rs911372828 Chr.5: 153490468-153490468 on Build GRCh38
rs777672321 Chr.5: 153490906-153490908 on Build GRCh38
rs944644873 Chr.5: 153490571-153490571 on Build GRCh38
rs540864196 Chr.5: 153490698-153490698 on Build GRCh38
rs755737996 Chr.5: 153490863-153490863 on Build GRCh38
rs894282856 Chr.5: 153490643-153490643 on Build GRCh38
rs1004846432 Chr.5: 153490724-153490724 on Build GRCh38
rs937750781 Chr.5: 153490462-153490462 on Build GRCh38
rs946458496 Chr.5: 153490827-153490827 on Build GRCh38
rs1055668273 Chr.5: 153490630-153490630 on Build GRCh38
rs746152276 Chr.5: 153490885-153490885 on Build GRCh38
rs754678234 Chr.5: 153490857-153490857 on Build GRCh38
rs376550685 Chr.5: 153490734-153490737 on Build GRCh38
rs766061038 Chr.5: 153490763-153490763 on Build GRCh38
rs755941162 Chr.5: 153490875-153490875 on Build GRCh38
rs750176124 Chr.5: 153490851-153490851 on Build GRCh38
rs1041693847 Chr.5: 153490618-153490618 on Build GRCh38
rs879132249 Chr.5: 153490832-153490832 on Build GRCh38
rs887867994 Chr.5: 153490722-153490722 on Build GRCh38
rs543665008 Chr.5: 153490510-153490510 on Build GRCh38
rs530412703 Chr.5: 153490871-153490871 on Build GRCh38
rs777390402 Chr.5: 153490870-153490870 on Build GRCh38
rs780947693 Chr.5: 153490861-153490861 on Build GRCh38
rs1027712516 Chr.5: 153490686-153490686 on Build GRCh38
rs958843138 Chr.5: 153490787-153490787 on Build GRCh38
rs772311830 Chr.5: 153490901-153490901 on Build GRCh38
rs529215830 Chr.5: 153490674-153490674 on Build GRCh38
rs200412983 Chr.5: 153490856-153490856 on Build GRCh38
rs61999280 Chr.5: 153490852-153490852 on Build GRCh38
rs761677542 Chr.5: 153490678-153490678 on Build GRCh38
rs984410600 Chr.5: 153490463-153490463 on Build GRCh38
rs1015378311 Chr.5: 153490731-153490734 on Build GRCh38
rs887464841 Chr.5: 153490782-153490782 on Build GRCh38
rs981690147 Chr.5: 153490751-153490751 on Build GRCh38
rs902387274 Chr.5: 153490525-153490525 on Build GRCh38
rs760292926 Chr.5: 153490547-153490547 on Build GRCh38
rs976832894 Chr.5: 153490486-153490486 on Build GRCh38
rs1045448311 Chr.5: 153490664-153490664 on Build GRCh38
rs182546001 Chr.5: 153490672-153490672 on Build GRCh38
rs964219574 Chr.5: 153490812-153490812 on Build GRCh38
rs964588566 Chr.5: 153490469-153490469 on Build GRCh38
rs1040223697 Chr.5: 153490598-153490598 on Build GRCh38
rs900363824 Chr.5: 153490625-153490625 on Build GRCh38
rs766252626 Chr.5: 153490847-153490847 on Build GRCh38
rs990289481 Chr.5: 153490792-153490792 on Build GRCh38
rs758261854 Chr.5: 153490844-153490844 on Build GRCh38
rs1034463345 Chr.5: 153490762-153490762 on Build GRCh38
rs993292689 Chr.5: 153490737-153490737 on Build GRCh38
rs1005792337 Chr.5: 153490801-153490801 on Build GRCh38
rs946029288 Chr.5: 153490607-153490607 on Build GRCh38
rs774793095 Chr.5: 153490880-153490880 on Build GRCh38
rs759102190 Chr.5: 153490479-153490480 on Build GRCh38
rs935903445 Chr.5: 153490628-153490628 on Build GRCh38
rs944592329 Chr.5: 153490858-153490858 on Build GRCh38

View Full Product Details