Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 153490804-153491315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIA1
Entrez Gene ID
2890
Gene Name
glutamate ionotropic receptor AMPA type subunit 1
Gene Aliases
GLUH1, GLUR1, GLURA, GluA1, HBGR1
UniGene
Hs.519693
Gene Chromosome Location
Chr.5: 153489615-153813873 on Build GRCh38
Transcripts
NM_000827.3 NM_001114183.1 NM_001258019.1 NM_001258020.1 NM_001258023.1 NR_047578.1 XM_011537635.2 XM_017009393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6102744 c.22T>C p.F8L Chr.5: 153490910-153490910 on Build GRCh38
1496079 c.31G>T p.G11C Chr.5: 153490919-153490919 on Build GRCh38
1696602 c.64C>T p.P22S Chr.5: 153490952-153490952 on Build GRCh38
1496078 c.31G>T p.G11C Chr.5: 153490919-153490919 on Build GRCh38
7156662 c.24C>T p.F8F Chr.5: 153490912-153490912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925943465 Chr.5: 153491036-153491036 on Build GRCh38
rs545259186 Chr.5: 153491218-153491218 on Build GRCh38
rs530680159 Chr.5: 153491235-153491235 on Build GRCh38
rs543429678 Chr.5: 153491088-153491088 on Build GRCh38
rs1007250309 Chr.5: 153491130-153491130 on Build GRCh38
rs767402812 Chr.5: 153490990-153490990 on Build GRCh38
rs771331045 Chr.5: 153490878-153490878 on Build GRCh38
rs201847050 Chr.5: 153490970-153490970 on Build GRCh38
rs574412596 Chr.5: 153491245-153491246 on Build GRCh38
rs777401453 Chr.5: 153491009-153491009 on Build GRCh38
rs957330026 Chr.5: 153491236-153491236 on Build GRCh38
rs576163964 Chr.5: 153491064-153491064 on Build GRCh38
rs1018488281 Chr.5: 153491142-153491142 on Build GRCh38
rs563353757 Chr.5: 153491229-153491229 on Build GRCh38
rs1002848968 Chr.5: 153491057-153491057 on Build GRCh38
rs201065021 Chr.5: 153491003-153491003 on Build GRCh38
rs1034533547 Chr.5: 153491070-153491070 on Build GRCh38
rs573842460 Chr.5: 153491101-153491101 on Build GRCh38
rs184978422 Chr.5: 153491291-153491291 on Build GRCh38
rs555777805 Chr.5: 153491089-153491089 on Build GRCh38
rs777672321 Chr.5: 153490906-153490908 on Build GRCh38
rs878861356 Chr.5: 153491033-153491033 on Build GRCh38
rs991459779 Chr.5: 153491242-153491242 on Build GRCh38
rs748690192 Chr.5: 153491016-153491016 on Build GRCh38
rs978273713 Chr.5: 153491162-153491163 on Build GRCh38
rs755737996 Chr.5: 153490863-153490863 on Build GRCh38
rs745912471 Chr.5: 153491275-153491275 on Build GRCh38
rs776558930 Chr.5: 153490941-153490941 on Build GRCh38
rs765079299 Chr.5: 153491113-153491113 on Build GRCh38
rs539516010 Chr.5: 153491030-153491030 on Build GRCh38
rs946458496 Chr.5: 153490827-153490827 on Build GRCh38
rs753505123 Chr.5: 153491197-153491197 on Build GRCh38
rs746152276 Chr.5: 153490885-153490885 on Build GRCh38
rs754678234 Chr.5: 153490857-153490857 on Build GRCh38
rs557843190 Chr.5: 153491037-153491037 on Build GRCh38
rs150205263 Chr.5: 153490969-153490969 on Build GRCh38
rs148776379 Chr.5: 153490963-153490963 on Build GRCh38
rs757788047 Chr.5: 153491018-153491018 on Build GRCh38
rs755941162 Chr.5: 153490875-153490875 on Build GRCh38
rs921859652 Chr.5: 153490931-153490931 on Build GRCh38
rs879132249 Chr.5: 153490832-153490832 on Build GRCh38
rs750176124 Chr.5: 153490851-153490851 on Build GRCh38
rs887412611 Chr.5: 153491126-153491126 on Build GRCh38
rs991343822 Chr.5: 153491048-153491048 on Build GRCh38
rs530412703 Chr.5: 153490871-153490871 on Build GRCh38
rs777390402 Chr.5: 153490870-153490870 on Build GRCh38
rs775737542 Chr.5: 153490918-153490918 on Build GRCh38
rs145860630 Chr.5: 153490952-153490952 on Build GRCh38
rs967601998 Chr.5: 153491211-153491211 on Build GRCh38
rs780947693 Chr.5: 153490861-153490861 on Build GRCh38
rs1022248100 Chr.5: 153491104-153491104 on Build GRCh38
rs900091279 Chr.5: 153491155-153491155 on Build GRCh38
rs150905426 Chr.5: 153491007-153491007 on Build GRCh38
rs147571145 Chr.5: 153491190-153491190 on Build GRCh38
rs772311830 Chr.5: 153490901-153490901 on Build GRCh38
rs752475276 Chr.5: 153491120-153491120 on Build GRCh38
rs397780450 Chr.5: 153491255-153491256 on Build GRCh38
rs200412983 Chr.5: 153490856-153490856 on Build GRCh38
rs61999280 Chr.5: 153490852-153490852 on Build GRCh38
rs923611536 Chr.5: 153491228-153491228 on Build GRCh38
rs145649709 Chr.5: 153491159-153491159 on Build GRCh38
rs929019581 Chr.5: 153491114-153491114 on Build GRCh38
rs373129424 Chr.5: 153490988-153490988 on Build GRCh38
rs949883292 Chr.5: 153491095-153491095 on Build GRCh38
rs897694207 Chr.5: 153491111-153491111 on Build GRCh38
rs760854220 Chr.5: 153490933-153490933 on Build GRCh38
rs758309986 Chr.5: 153491174-153491174 on Build GRCh38
rs998487388 Chr.5: 153491180-153491180 on Build GRCh38
rs533177457 Chr.5: 153491195-153491195 on Build GRCh38
rs766252626 Chr.5: 153490847-153490847 on Build GRCh38
rs371551878 Chr.5: 153491054-153491054 on Build GRCh38
rs565934520 Chr.5: 153491026-153491026 on Build GRCh38
rs758261854 Chr.5: 153490844-153490844 on Build GRCh38
rs369322935 Chr.5: 153490938-153490938 on Build GRCh38
rs747137692 Chr.5: 153491011-153491013 on Build GRCh38
rs34449755 Chr.5: 153491246-153491247 on Build GRCh38
rs1029712267 Chr.5: 153490937-153490937 on Build GRCh38
rs770962751 Chr.5: 153491019-153491020 on Build GRCh38
rs915950530 Chr.5: 153491269-153491269 on Build GRCh38
rs936017438 Chr.5: 153491038-153491038 on Build GRCh38
rs774793095 Chr.5: 153490880-153490880 on Build GRCh38
rs3749655 Chr.5: 153490980-153490980 on Build GRCh38
rs944592329 Chr.5: 153490858-153490858 on Build GRCh38

View Full Product Details