Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157180814-157181319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITK
Entrez Gene ID
3702
Gene Name
IL2 inducible T-cell kinase
Gene Aliases
EMT, LPFS1, LYK, PSCTK2
UniGene
Hs.558348
Gene Chromosome Location
Chr.5: 157180896-157255185 on Build GRCh38
Transcripts
NM_005546.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564061 c.54_55GA>AT p.R19* Chr.5: 157181031-157181032 on Build GRCh38
317493 c.86G>T p.R29L Chr.5: 157181063-157181063 on Build GRCh38
3246264 c.130C>T p.R44C Chr.5: 157181107-157181107 on Build GRCh38
94534 c.121T>A p.F41I Chr.5: 157181098-157181098 on Build GRCh38
3853114 c.61A>G p.T21A Chr.5: 157181038-157181038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146496148 Chr.5: 157181064-157181064 on Build GRCh38
rs367920133 Chr.5: 157180998-157180998 on Build GRCh38
rs761792747 Chr.5: 157181134-157181134 on Build GRCh38
rs1040832025 Chr.5: 157180897-157180897 on Build GRCh38
rs1005569262 Chr.5: 157181116-157181116 on Build GRCh38
rs776401801 Chr.5: 157181158-157181159 on Build GRCh38
rs200603754 Chr.5: 157181128-157181128 on Build GRCh38
rs765948222 Chr.5: 157181010-157181010 on Build GRCh38
rs757943266 Chr.5: 157181250-157181250 on Build GRCh38
rs199521747 Chr.5: 157181146-157181146 on Build GRCh38
rs568635034 Chr.5: 157180971-157180971 on Build GRCh38
rs755319823 Chr.5: 157180934-157180935 on Build GRCh38
rs766419332 Chr.5: 157181132-157181133 on Build GRCh38
rs397514261 Chr.5: 157181063-157181063 on Build GRCh38
rs140064341 Chr.5: 157181092-157181092 on Build GRCh38
rs773347606 Chr.5: 157180947-157180947 on Build GRCh38
rs924720745 Chr.5: 157180862-157180862 on Build GRCh38
rs531949883 Chr.5: 157180918-157180918 on Build GRCh38
rs776585958 Chr.5: 157180984-157180984 on Build GRCh38
rs111423533 Chr.5: 157181047-157181047 on Build GRCh38
rs117806853 Chr.5: 157180883-157180883 on Build GRCh38
rs751083005 Chr.5: 157181013-157181013 on Build GRCh38
rs201892355 Chr.5: 157181108-157181108 on Build GRCh38
rs546548192 Chr.5: 157180890-157180890 on Build GRCh38
rs900615034 Chr.5: 157180898-157180898 on Build GRCh38
rs779466042 Chr.5: 157181004-157181004 on Build GRCh38
rs781686293 Chr.5: 157181026-157181026 on Build GRCh38
rs959120596 Chr.5: 157181189-157181189 on Build GRCh38
rs762676771 Chr.5: 157181145-157181145 on Build GRCh38
rs923105557 Chr.5: 157180922-157180922 on Build GRCh38
rs1031603146 Chr.5: 157181265-157181265 on Build GRCh38
rs754433426 Chr.5: 157181022-157181022 on Build GRCh38
rs539417591 Chr.5: 157181151-157181151 on Build GRCh38
rs749506217 Chr.5: 157180949-157180949 on Build GRCh38
rs778189459 Chr.5: 157181083-157181083 on Build GRCh38
rs759626126 Chr.5: 157180974-157180974 on Build GRCh38
rs113811363 Chr.5: 157180904-157180904 on Build GRCh38
rs202178950 Chr.5: 157181143-157181143 on Build GRCh38
rs148134638 Chr.5: 157180946-157180946 on Build GRCh38
rs769943503 Chr.5: 157180845-157180845 on Build GRCh38
rs145768533 Chr.5: 157180861-157180861 on Build GRCh38
rs868709834 Chr.5: 157180999-157180999 on Build GRCh38
rs756436987 Chr.5: 157180851-157180851 on Build GRCh38
rs765005531 Chr.5: 157181136-157181136 on Build GRCh38
rs750096142 Chr.5: 157180992-157180992 on Build GRCh38
rs761547914 Chr.5: 157180986-157180986 on Build GRCh38
rs774659403 Chr.5: 157181107-157181107 on Build GRCh38
rs978443660 Chr.5: 157181233-157181233 on Build GRCh38
rs749541467 Chr.5: 157181091-157181091 on Build GRCh38
rs756484580 Chr.5: 157181067-157181067 on Build GRCh38
rs1022179608 Chr.5: 157181218-157181218 on Build GRCh38
rs1013496341 Chr.5: 157181195-157181195 on Build GRCh38
rs764145653 Chr.5: 157180983-157180983 on Build GRCh38
rs201513154 Chr.5: 157181062-157181062 on Build GRCh38
rs765056835 Chr.5: 157180989-157180989 on Build GRCh38
rs967975462 Chr.5: 157181223-157181223 on Build GRCh38
rs140725323 Chr.5: 157180839-157180839 on Build GRCh38
rs1013131703 Chr.5: 157180842-157180842 on Build GRCh38
rs909127502 Chr.5: 157181049-157181049 on Build GRCh38
rs771193249 Chr.5: 157180968-157180968 on Build GRCh38
rs758061743 Chr.5: 157181005-157181005 on Build GRCh38
rs409602 Chr.5: 157181273-157181273 on Build GRCh38
rs540329798 Chr.5: 157180843-157180843 on Build GRCh38
rs990646050 Chr.5: 157181194-157181194 on Build GRCh38

View Full Product Details