Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157240082-157240519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITK
Entrez Gene ID
3702
Gene Name
IL2 inducible T-cell kinase
Gene Aliases
EMT, LPFS1, LYK, PSCTK2
UniGene
Hs.558348
Gene Chromosome Location
Chr.5: 157180896-157255185 on Build GRCh38
Transcripts
NM_005546.3 XM_017009443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1435541 c.941A>T p.D314V Chr.5: 157240151-157240151 on Build GRCh38
5591401 c.952C>T p.L318F Chr.5: 157240162-157240162 on Build GRCh38
6688554 c.923C>T p.A308V Chr.5: 157240133-157240133 on Build GRCh38
4606109 c.910C>G p.R304G Chr.5: 157240120-157240120 on Build GRCh38
6280203 c.937T>A p.F313I Chr.5: 157240147-157240147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183753479 Chr.5: 157240131-157240131 on Build GRCh38
rs78723238 Chr.5: 157240250-157240250 on Build GRCh38
rs372201861 Chr.5: 157240137-157240137 on Build GRCh38
rs30119 Chr.5: 157240436-157240436 on Build GRCh38
rs1048991414 Chr.5: 157240430-157240430 on Build GRCh38
rs749826776 Chr.5: 157240214-157240214 on Build GRCh38
rs879199229 Chr.5: 157240108-157240108 on Build GRCh38
rs751812748 Chr.5: 157240164-157240164 on Build GRCh38
rs1002068488 Chr.5: 157240438-157240438 on Build GRCh38
rs886425802 Chr.5: 157240347-157240347 on Build GRCh38
rs753127825 Chr.5: 157240192-157240192 on Build GRCh38
rs957728410 Chr.5: 157240226-157240226 on Build GRCh38
rs752946188 Chr.5: 157240331-157240331 on Build GRCh38
rs886381051 Chr.5: 157240434-157240434 on Build GRCh38
rs778347135 Chr.5: 157240207-157240207 on Build GRCh38
rs138078237 Chr.5: 157240150-157240150 on Build GRCh38
rs994602522 Chr.5: 157240422-157240422 on Build GRCh38
rs930570146 Chr.5: 157240326-157240326 on Build GRCh38
rs763088444 Chr.5: 157240157-157240157 on Build GRCh38
rs532438850 Chr.5: 157240298-157240298 on Build GRCh38
rs990746194 Chr.5: 157240481-157240481 on Build GRCh38
rs768847270 Chr.5: 157240210-157240210 on Build GRCh38
rs144715799 Chr.5: 157240392-157240392 on Build GRCh38
rs550882682 Chr.5: 157240473-157240473 on Build GRCh38
rs950157818 Chr.5: 157240366-157240366 on Build GRCh38
rs867792646 Chr.5: 157240447-157240447 on Build GRCh38
rs755099007 Chr.5: 157240229-157240229 on Build GRCh38
rs893410373 Chr.5: 157240466-157240466 on Build GRCh38
rs773986316 Chr.5: 157240125-157240125 on Build GRCh38
rs199589124 Chr.5: 157240227-157240227 on Build GRCh38
rs6860090 Chr.5: 157240270-157240270 on Build GRCh38
rs756521301 Chr.5: 157240203-157240203 on Build GRCh38
rs770504605 Chr.5: 157240123-157240123 on Build GRCh38
rs368648728 Chr.5: 157240221-157240221 on Build GRCh38
rs756744263 Chr.5: 157240217-157240217 on Build GRCh38
rs748842677 Chr.5: 157240121-157240121 on Build GRCh38
rs760485497 Chr.5: 157240140-157240140 on Build GRCh38
rs776620764 Chr.5: 157240235-157240235 on Build GRCh38
rs761893928 Chr.5: 157240236-157240236 on Build GRCh38
rs762956142 Chr.5: 157240131-157240131 on Build GRCh38
rs766625478 Chr.5: 157240162-157240162 on Build GRCh38
rs911745703 Chr.5: 157240254-157240254 on Build GRCh38
rs966342708 Chr.5: 157240493-157240493 on Build GRCh38
rs968068546 Chr.5: 157240491-157240491 on Build GRCh38
rs186976396 Chr.5: 157240497-157240497 on Build GRCh38
rs1024427618 Chr.5: 157240442-157240442 on Build GRCh38
rs571566681 Chr.5: 157240397-157240397 on Build GRCh38
rs1001290633 Chr.5: 157240467-157240467 on Build GRCh38
rs920548484 Chr.5: 157240317-157240317 on Build GRCh38
rs771829181 Chr.5: 157240132-157240132 on Build GRCh38
rs778440201 Chr.5: 157240218-157240218 on Build GRCh38
rs567512946 Chr.5: 157240474-157240474 on Build GRCh38
rs767955456 Chr.5: 157240188-157240188 on Build GRCh38
rs1001809076 Chr.5: 157240156-157240156 on Build GRCh38
rs755292379 Chr.5: 157240184-157240184 on Build GRCh38
rs1020656557 Chr.5: 157240484-157240484 on Build GRCh38
rs771739318 Chr.5: 157240222-157240222 on Build GRCh38
rs974690974 Chr.5: 157240294-157240294 on Build GRCh38
rs369067925 Chr.5: 157240228-157240228 on Build GRCh38
rs764110831 Chr.5: 157240145-157240145 on Build GRCh38

View Full Product Details