Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163473976-163474480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMMR
Entrez Gene ID
3161
Gene Name
hyaluronan mediated motility receptor
Gene Aliases
CD168, IHABP, RHAMM
UniGene
Hs.740467
Gene Chromosome Location
Chr.5: 163460511-163491945 on Build GRCh38
Transcripts
NM_001142556.1 NM_001142557.1 NM_012484.2 NM_012485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6586321 c.904G>T p.D302Y Chr.5: 163474059-163474059 on Build GRCh38
482470 c.993A>G p.E331E Chr.5: 163474148-163474148 on Build GRCh38
4417057 c.960C>G p.N320K Chr.5: 163474115-163474115 on Build GRCh38
5591277 c.990G>A p.Q330Q Chr.5: 163474145-163474145 on Build GRCh38
3248040 c.939T>C p.N313N Chr.5: 163474094-163474094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774707555 Chr.5: 163474207-163474207 on Build GRCh38
rs544846255 Chr.5: 163474366-163474366 on Build GRCh38
rs950877583 Chr.5: 163474225-163474225 on Build GRCh38
rs1000946454 Chr.5: 163474137-163474137 on Build GRCh38
rs917792757 Chr.5: 163474386-163474386 on Build GRCh38
rs766905156 Chr.5: 163474222-163474222 on Build GRCh38
rs146089957 Chr.5: 163474066-163474066 on Build GRCh38
rs773884555 Chr.5: 163474083-163474083 on Build GRCh38
rs533687708 Chr.5: 163474325-163474325 on Build GRCh38
rs770301976 Chr.5: 163474170-163474170 on Build GRCh38
rs549343728 Chr.5: 163474101-163474101 on Build GRCh38
rs2303077 Chr.5: 163474115-163474115 on Build GRCh38
rs1040774755 Chr.5: 163474142-163474142 on Build GRCh38
rs956867582 Chr.5: 163474371-163474371 on Build GRCh38
rs567581726 Chr.5: 163474109-163474109 on Build GRCh38
rs763883043 Chr.5: 163474248-163474248 on Build GRCh38
rs757514146 Chr.5: 163474111-163474111 on Build GRCh38
rs373512642 Chr.5: 163474125-163474125 on Build GRCh38
rs751042026 Chr.5: 163474098-163474098 on Build GRCh38
rs572992389 Chr.5: 163474288-163474288 on Build GRCh38
rs1025517007 Chr.5: 163474260-163474260 on Build GRCh38
rs181404631 Chr.5: 163474346-163474346 on Build GRCh38
rs879907375 Chr.5: 163474445-163474445 on Build GRCh38
rs749813056 Chr.5: 163474202-163474202 on Build GRCh38
rs980524132 Chr.5: 163474328-163474328 on Build GRCh38
rs771628469 Chr.5: 163474206-163474206 on Build GRCh38
rs114418785 Chr.5: 163474033-163474033 on Build GRCh38
rs1024082596 Chr.5: 163474333-163474333 on Build GRCh38
rs143570587 Chr.5: 163474196-163474196 on Build GRCh38
rs777735803 Chr.5: 163474011-163474011 on Build GRCh38
rs2303078 Chr.5: 163474150-163474150 on Build GRCh38
rs74933333 Chr.5: 163474197-163474197 on Build GRCh38
rs772162647 Chr.5: 163474047-163474047 on Build GRCh38
rs935460473 Chr.5: 163474030-163474030 on Build GRCh38
rs776708341 Chr.5: 163474061-163474061 on Build GRCh38
rs776499506 Chr.5: 163474147-163474147 on Build GRCh38
rs72808221 Chr.5: 163474268-163474268 on Build GRCh38
rs189760157 Chr.5: 163474005-163474005 on Build GRCh38
rs147062489 Chr.5: 163474391-163474391 on Build GRCh38
rs762290867 Chr.5: 163474063-163474063 on Build GRCh38
rs550643129 Chr.5: 163474149-163474149 on Build GRCh38
rs915410068 Chr.5: 163474435-163474435 on Build GRCh38
rs759133666 Chr.5: 163474208-163474208 on Build GRCh38
rs969400327 Chr.5: 163474362-163474362 on Build GRCh38
rs141692321 Chr.5: 163474365-163474365 on Build GRCh38
rs770748815 Chr.5: 163474145-163474145 on Build GRCh38
rs768994182 Chr.5: 163474060-163474060 on Build GRCh38
rs376582749 Chr.5: 163474048-163474048 on Build GRCh38
rs746725934 Chr.5: 163474130-163474179 on Build GRCh38
rs928144138 Chr.5: 163474028-163474028 on Build GRCh38
rs775238452 Chr.5: 163474232-163474232 on Build GRCh38
rs753788370 Chr.5: 163474121-163474121 on Build GRCh38
rs184988348 Chr.5: 163474446-163474446 on Build GRCh38
rs980086073 Chr.5: 163474010-163474010 on Build GRCh38
rs557859410 Chr.5: 163474274-163474274 on Build GRCh38
rs374170075 Chr.5: 163474153-163474153 on Build GRCh38
rs778818119 Chr.5: 163474143-163474143 on Build GRCh38
rs982207825 Chr.5: 163474016-163474016 on Build GRCh38
rs760189290 Chr.5: 163474240-163474240 on Build GRCh38
rs371249746 Chr.5: 163474213-163474213 on Build GRCh38
rs111891464 Chr.5: 163474331-163474331 on Build GRCh38
rs763502082 Chr.5: 163474087-163474087 on Build GRCh38
rs888400987 Chr.5: 163474265-163474265 on Build GRCh38
rs373048592 Chr.5: 163474261-163474261 on Build GRCh38
rs765996849 Chr.5: 163474088-163474088 on Build GRCh38
rs758964372 Chr.5: 163474099-163474099 on Build GRCh38
rs988270213 Chr.5: 163474378-163474378 on Build GRCh38
rs879068707 Chr.5: 163474136-163474136 on Build GRCh38

View Full Product Details