Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25725929-25726431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2AA
Entrez Gene ID
221613
Gene Name
histone cluster 1, H2aa
Gene Aliases
H2AA, H2AFR, TH2A, bA317E16.2
UniGene
Hs.406739
Gene Chromosome Location
Chr.6: 25726063-25726562 on Build GRCh38
Transcripts
NM_170745.3 XM_017011201.1

Gene Symbol
SLC17A1
Entrez Gene ID
6568
Gene Name
solute carrier family 17 member 1
Gene Aliases
NAPI-1, NPT-1, NPT1
UniGene
Hs.205816
Gene Chromosome Location
Chr.6: 25723744-25832108 on Build GRCh38
Transcripts
NM_170745.3 XM_017011201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3921177 c.209C>G p.A70G Chr.6: 25726319-25726319 on Build GRCh38
6453118 c.274G>A p.E92K Chr.6: 25726254-25726254 on Build GRCh38
6500493 c.169G>T p.E57* Chr.6: 25726359-25726359 on Build GRCh38
4393818 c.113G>A p.G38E Chr.6: 25726415-25726415 on Build GRCh38
6680213 c.98G>A p.R33H Chr.6: 25726430-25726430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151084411 Chr.6: 25726324-25726324 on Build GRCh38
rs148750226 Chr.6: 25726203-25726203 on Build GRCh38
rs755399560 Chr.6: 25726056-25726056 on Build GRCh38
rs952111605 Chr.6: 25726092-25726092 on Build GRCh38
rs772978548 Chr.6: 25726314-25726314 on Build GRCh38
rs551081966 Chr.6: 25726243-25726243 on Build GRCh38
rs746365522 Chr.6: 25726287-25726287 on Build GRCh38
rs575327686 Chr.6: 25725984-25725984 on Build GRCh38
rs148811117 Chr.6: 25726112-25726112 on Build GRCh38
rs748966391 Chr.6: 25726167-25726169 on Build GRCh38
rs771578160 Chr.6: 25726313-25726313 on Build GRCh38
rs770350629 Chr.6: 25726289-25726289 on Build GRCh38
rs574206474 Chr.6: 25726023-25726023 on Build GRCh38
rs763406620 Chr.6: 25726312-25726312 on Build GRCh38
rs368937249 Chr.6: 25725988-25725988 on Build GRCh38
rs367687104 Chr.6: 25726387-25726387 on Build GRCh38
rs775650743 Chr.6: 25726297-25726297 on Build GRCh38
rs142933340 Chr.6: 25726146-25726146 on Build GRCh38
rs761026578 Chr.6: 25726156-25726156 on Build GRCh38
rs767204976 Chr.6: 25726235-25726235 on Build GRCh38
rs548892947 Chr.6: 25725960-25725960 on Build GRCh38
rs758505437 Chr.6: 25726276-25726276 on Build GRCh38
rs748954973 Chr.6: 25726043-25726044 on Build GRCh38
rs774506049 Chr.6: 25726098-25726098 on Build GRCh38
rs749044206 Chr.6: 25726195-25726195 on Build GRCh38
rs757976662 Chr.6: 25726113-25726113 on Build GRCh38
rs574118105 Chr.6: 25726329-25726329 on Build GRCh38
rs745870694 Chr.6: 25726081-25726081 on Build GRCh38
rs764669263 Chr.6: 25726106-25726107 on Build GRCh38
rs753820689 Chr.6: 25726252-25726252 on Build GRCh38
rs765247096 Chr.6: 25726332-25726332 on Build GRCh38
rs751373475 Chr.6: 25726402-25726402 on Build GRCh38
rs780635723 Chr.6: 25726374-25726374 on Build GRCh38
rs376093687 Chr.6: 25726139-25726139 on Build GRCh38
rs760286610 Chr.6: 25726238-25726238 on Build GRCh38
rs200653429 Chr.6: 25726063-25726063 on Build GRCh38
rs6940348 Chr.6: 25726115-25726115 on Build GRCh38
rs932188509 Chr.6: 25726075-25726075 on Build GRCh38
rs540559740 Chr.6: 25726029-25726029 on Build GRCh38
rs370836712 Chr.6: 25726183-25726183 on Build GRCh38
rs143443930 Chr.6: 25726278-25726278 on Build GRCh38
rs747849538 Chr.6: 25726348-25726349 on Build GRCh38
rs780173095 Chr.6: 25726188-25726188 on Build GRCh38
rs144269597 Chr.6: 25726186-25726186 on Build GRCh38
rs946262481 Chr.6: 25726399-25726399 on Build GRCh38
rs778804735 Chr.6: 25726299-25726299 on Build GRCh38
rs9467583 Chr.6: 25726393-25726393 on Build GRCh38
rs146789964 Chr.6: 25726266-25726266 on Build GRCh38
rs865997326 Chr.6: 25726364-25726365 on Build GRCh38
rs180715162 Chr.6: 25726061-25726061 on Build GRCh38
rs200865884 Chr.6: 25726391-25726391 on Build GRCh38
rs750864746 Chr.6: 25726110-25726110 on Build GRCh38
rs1053042964 Chr.6: 25726378-25726378 on Build GRCh38
rs773104554 Chr.6: 25726359-25726360 on Build GRCh38
rs1002664843 Chr.6: 25726026-25726026 on Build GRCh38
rs199912105 Chr.6: 25726330-25726330 on Build GRCh38
rs150250677 Chr.6: 25726381-25726381 on Build GRCh38
rs76591917 Chr.6: 25726339-25726339 on Build GRCh38
rs776238056 Chr.6: 25726390-25726390 on Build GRCh38
rs112943240 Chr.6: 25726101-25726102 on Build GRCh38
rs745792343 Chr.6: 25726131-25726131 on Build GRCh38
rs147403459 Chr.6: 25726159-25726159 on Build GRCh38
rs752537189 Chr.6: 25726273-25726273 on Build GRCh38
rs16890971 Chr.6: 25725959-25725959 on Build GRCh38
rs767683699 Chr.6: 25726108-25726108 on Build GRCh38
rs983853765 Chr.6: 25726095-25726095 on Build GRCh38
rs769015649 Chr.6: 25726089-25726089 on Build GRCh38
rs893435193 Chr.6: 25726047-25726047 on Build GRCh38
rs755024291 Chr.6: 25726259-25726259 on Build GRCh38
rs560105380 Chr.6: 25726059-25726059 on Build GRCh38
rs749990653 Chr.6: 25726126-25726127 on Build GRCh38
rs187505332 Chr.6: 25726318-25726318 on Build GRCh38
rs762868475 Chr.6: 25726400-25726400 on Build GRCh38
rs112598837 Chr.6: 25726379-25726379 on Build GRCh38
rs144559425 Chr.6: 25726282-25726282 on Build GRCh38
rs536726454 Chr.6: 25726285-25726285 on Build GRCh38
rs952133011 Chr.6: 25725987-25725987 on Build GRCh38
rs781194803 Chr.6: 25726367-25726367 on Build GRCh38
rs565643275 Chr.6: 25726225-25726225 on Build GRCh38
rs1042905450 Chr.6: 25726336-25726336 on Build GRCh38
rs751905438 Chr.6: 25726036-25726036 on Build GRCh38
rs368562425 Chr.6: 25726153-25726153 on Build GRCh38
rs761819890 Chr.6: 25726103-25726103 on Build GRCh38
rs367946056 Chr.6: 25726231-25726231 on Build GRCh38
rs990364637 Chr.6: 25726161-25726161 on Build GRCh38
rs775985152 Chr.6: 25726230-25726230 on Build GRCh38
rs747121209 Chr.6: 25726221-25726221 on Build GRCh38
rs537191502 Chr.6: 25725999-25725999 on Build GRCh38
rs748439979 Chr.6: 25726152-25726152 on Build GRCh38
rs200598078 Chr.6: 25726229-25726229 on Build GRCh38
rs199747810 Chr.6: 25726095-25726102 on Build GRCh38
rs768242433 Chr.6: 25726084-25726085 on Build GRCh38
rs529086208 Chr.6: 25726167-25726167 on Build GRCh38
rs765641279 Chr.6: 25726171-25726171 on Build GRCh38
rs558676738 Chr.6: 25726407-25726407 on Build GRCh38
rs752963573 Chr.6: 25726086-25726088 on Build GRCh38
rs902243915 Chr.6: 25726060-25726060 on Build GRCh38
rs185440568 Chr.6: 25726068-25726068 on Build GRCh38
rs199954745 Chr.6: 25726217-25726217 on Build GRCh38
rs77698027 Chr.6: 25726046-25726046 on Build GRCh38
rs760346071 Chr.6: 25726386-25726386 on Build GRCh38
rs763938944 Chr.6: 25726401-25726401 on Build GRCh38
rs775464003 Chr.6: 25726396-25726396 on Build GRCh38
rs751778560 Chr.6: 25726351-25726351 on Build GRCh38
rs201764642 Chr.6: 25726101-25726101 on Build GRCh38
rs1036015351 Chr.6: 25726011-25726011 on Build GRCh38
rs370462147 Chr.6: 25726136-25726136 on Build GRCh38
rs747160353 Chr.6: 25726379-25726379 on Build GRCh38
rs760079232 Chr.6: 25726227-25726227 on Build GRCh38
rs754524268 Chr.6: 25726241-25726243 on Build GRCh38
rs375336327 Chr.6: 25726370-25726370 on Build GRCh38
rs764125264 Chr.6: 25725956-25725956 on Build GRCh38
rs749102345 Chr.6: 25726138-25726138 on Build GRCh38
rs900310623 Chr.6: 25726264-25726264 on Build GRCh38
rs573588303 Chr.6: 25726364-25726364 on Build GRCh38
rs778835363 Chr.6: 25726204-25726204 on Build GRCh38
rs759104722 Chr.6: 25726392-25726392 on Build GRCh38
rs779913025 Chr.6: 25726164-25726165 on Build GRCh38
rs937545177 Chr.6: 25726041-25726041 on Build GRCh38
rs9379785 Chr.6: 25725985-25725985 on Build GRCh38
rs372043302 Chr.6: 25726237-25726237 on Build GRCh38
rs756702597 Chr.6: 25726125-25726125 on Build GRCh38
rs907942124 Chr.6: 25725994-25725994 on Build GRCh38
rs769448281 Chr.6: 25726083-25726083 on Build GRCh38
rs78532821 Chr.6: 25725986-25725986 on Build GRCh38
rs775495831 Chr.6: 25726087-25726087 on Build GRCh38
rs201841051 Chr.6: 25726127-25726127 on Build GRCh38
rs923395168 Chr.6: 25726074-25726074 on Build GRCh38
rs768939863 Chr.6: 25726184-25726190 on Build GRCh38
rs775281918 Chr.6: 25726174-25726174 on Build GRCh38
rs765256961 Chr.6: 25725976-25725976 on Build GRCh38
rs375561515 Chr.6: 25726088-25726088 on Build GRCh38
rs372965867 Chr.6: 25726119-25726119 on Build GRCh38
rs191563173 Chr.6: 25726100-25726100 on Build GRCh38
rs953617398 Chr.6: 25726240-25726240 on Build GRCh38
rs143540809 Chr.6: 25726218-25726218 on Build GRCh38
rs773398885 Chr.6: 25726154-25726154 on Build GRCh38
rs777472670 Chr.6: 25726349-25726349 on Build GRCh38
rs766927556 Chr.6: 25726182-25726182 on Build GRCh38
rs950285359 Chr.6: 25726010-25726010 on Build GRCh38
rs1031050142 Chr.6: 25726065-25726065 on Build GRCh38
rs113723230 Chr.6: 25726210-25726210 on Build GRCh38
rs771375608 Chr.6: 25726222-25726222 on Build GRCh38
rs756179010 Chr.6: 25726133-25726133 on Build GRCh38
rs774912477 Chr.6: 25726385-25726385 on Build GRCh38
rs191966362 Chr.6: 25726333-25726333 on Build GRCh38
rs150006012 Chr.6: 25726179-25726179 on Build GRCh38
rs755678708 Chr.6: 25726151-25726152 on Build GRCh38
rs557271934 Chr.6: 25726015-25726015 on Build GRCh38
rs1018889380 Chr.6: 25725969-25725969 on Build GRCh38
rs761965424 Chr.6: 25725964-25725964 on Build GRCh38
rs1057254652 Chr.6: 25726293-25726293 on Build GRCh38
rs573611231 Chr.6: 25726022-25726022 on Build GRCh38
rs565319041 Chr.6: 25725970-25725970 on Build GRCh38
rs766871210 Chr.6: 25726157-25726157 on Build GRCh38
rs376411797 Chr.6: 25726126-25726126 on Build GRCh38
rs772096226 Chr.6: 25726357-25726358 on Build GRCh38
rs1009145881 Chr.6: 25725966-25725966 on Build GRCh38
rs749809393 Chr.6: 25726380-25726380 on Build GRCh38

View Full Product Details