Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39335416-39335921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
UniGene
Hs.588202
Gene Chromosome Location
Chr.6: 39335100-39725405 on Build GRCh38
Transcripts
NM_001289020.1 NM_001289021.1 NM_001289024.1 NM_145027.4

SNP Detail

SNP ID Chromosome Location
rs557259004 Chr.6: 39335482-39335482 on Build GRCh38
rs561957643 Chr.6: 39335803-39335803 on Build GRCh38
rs985805701 Chr.6: 39335830-39335830 on Build GRCh38
rs956681517 Chr.6: 39335465-39335465 on Build GRCh38
rs953060835 Chr.6: 39335513-39335513 on Build GRCh38
rs956713982 Chr.6: 39335467-39335467 on Build GRCh38
rs16891944 Chr.6: 39335506-39335506 on Build GRCh38
rs906441816 Chr.6: 39335444-39335444 on Build GRCh38
rs1045031453 Chr.6: 39335650-39335650 on Build GRCh38
rs1003392156 Chr.6: 39335697-39335697 on Build GRCh38
rs956482009 Chr.6: 39335834-39335834 on Build GRCh38
rs1013779256 Chr.6: 39335842-39335842 on Build GRCh38
rs139304973 Chr.6: 39335564-39335564 on Build GRCh38
rs142173734 Chr.6: 39335577-39335577 on Build GRCh38
rs1007067844 Chr.6: 39335718-39335718 on Build GRCh38
rs1051202478 Chr.6: 39335618-39335618 on Build GRCh38
rs965620827 Chr.6: 39335894-39335894 on Build GRCh38
rs1030806306 Chr.6: 39335457-39335457 on Build GRCh38
rs1031256796 Chr.6: 39335721-39335721 on Build GRCh38
rs989207509 Chr.6: 39335503-39335503 on Build GRCh38
rs977341615 Chr.6: 39335585-39335585 on Build GRCh38
rs764917673 Chr.6: 39335617-39335617 on Build GRCh38
rs200765599 Chr.6: 39335566-39335566 on Build GRCh38
rs892266952 Chr.6: 39335723-39335723 on Build GRCh38
rs917796232 Chr.6: 39335497-39335497 on Build GRCh38
rs1048506695 Chr.6: 39335614-39335614 on Build GRCh38
rs991999462 Chr.6: 39335472-39335472 on Build GRCh38
rs948080539 Chr.6: 39335619-39335619 on Build GRCh38
rs944240880 Chr.6: 39335534-39335534 on Build GRCh38
rs9462531 Chr.6: 39335818-39335818 on Build GRCh38
rs918723940 Chr.6: 39335592-39335592 on Build GRCh38
rs190137715 Chr.6: 39335451-39335451 on Build GRCh38
rs927248483 Chr.6: 39335536-39335536 on Build GRCh38
rs117842239 Chr.6: 39335867-39335867 on Build GRCh38
rs906474475 Chr.6: 39335656-39335656 on Build GRCh38
rs1011151132 Chr.6: 39335786-39335786 on Build GRCh38
rs758340470 Chr.6: 39335518-39335518 on Build GRCh38
rs147813134 Chr.6: 39335593-39335593 on Build GRCh38
rs564418655 Chr.6: 39335864-39335864 on Build GRCh38
rs115025619 Chr.6: 39335531-39335531 on Build GRCh38
rs530126032 Chr.6: 39335475-39335475 on Build GRCh38
rs541863984 Chr.6: 39335725-39335725 on Build GRCh38
rs903491658 Chr.6: 39335751-39335751 on Build GRCh38
rs891317663 Chr.6: 39335671-39335671 on Build GRCh38
rs550887864 Chr.6: 39335859-39335859 on Build GRCh38

View Full Product Details