Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39345460-39345871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
UniGene
Hs.588202
Gene Chromosome Location
Chr.6: 39335100-39725405 on Build GRCh38
Transcripts
NM_001289020.1 NM_001289021.1 NM_001289024.1 NM_145027.4 XM_005248904.4 XM_011514357.2 XM_011514358.2 XM_011514359.2 XM_017010427.1 XM_017010428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3830455 c.2305A>C p.T769P Chr.6: 39345716-39345716 on Build GRCh38
4971304 c.2250A>C p.K750N Chr.6: 39345771-39345771 on Build GRCh38
3830456 c.2137A>C p.T713P Chr.6: 39345716-39345716 on Build GRCh38
3777643 c.645G>C p.Q215H Chr.6: 39345729-39345729 on Build GRCh38
5512259 c.610C>T p.P204S Chr.6: 39345764-39345764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752309377 Chr.6: 39345722-39345722 on Build GRCh38
rs3832430 Chr.6: 39345843-39345844 on Build GRCh38
rs112535273 Chr.6: 39345621-39345621 on Build GRCh38
rs111485681 Chr.6: 39345575-39345575 on Build GRCh38
rs370908730 Chr.6: 39345652-39345652 on Build GRCh38
rs976689507 Chr.6: 39345709-39345709 on Build GRCh38
rs1006321209 Chr.6: 39345623-39345623 on Build GRCh38
rs771911037 Chr.6: 39345663-39345663 on Build GRCh38
rs776439085 Chr.6: 39345825-39345825 on Build GRCh38
rs761536674 Chr.6: 39345657-39345657 on Build GRCh38
rs1050671338 Chr.6: 39345760-39345760 on Build GRCh38
rs773039483 Chr.6: 39345785-39345785 on Build GRCh38
rs538642663 Chr.6: 39345580-39345580 on Build GRCh38
rs746829504 Chr.6: 39345788-39345788 on Build GRCh38
rs759031192 Chr.6: 39345826-39345826 on Build GRCh38
rs935736484 Chr.6: 39345625-39345625 on Build GRCh38
rs756636672 Chr.6: 39345759-39345759 on Build GRCh38
rs146459902 Chr.6: 39345547-39345547 on Build GRCh38
rs770863389 Chr.6: 39345819-39345819 on Build GRCh38
rs145557784 Chr.6: 39345751-39345751 on Build GRCh38
rs762739361 Chr.6: 39345833-39345833 on Build GRCh38
rs775280979 Chr.6: 39345831-39345831 on Build GRCh38
rs761021163 Chr.6: 39345790-39345790 on Build GRCh38
rs932319150 Chr.6: 39345710-39345710 on Build GRCh38
rs80139026 Chr.6: 39345605-39345605 on Build GRCh38
rs768879836 Chr.6: 39345653-39345653 on Build GRCh38
rs771822990 Chr.6: 39345777-39345777 on Build GRCh38
rs1041913071 Chr.6: 39345487-39345487 on Build GRCh38
rs200351411 Chr.6: 39345767-39345767 on Build GRCh38
rs914253655 Chr.6: 39345791-39345791 on Build GRCh38
rs774438800 Chr.6: 39345656-39345656 on Build GRCh38
rs776159047 Chr.6: 39345719-39345719 on Build GRCh38
rs749321005 Chr.6: 39345766-39345766 on Build GRCh38
rs780453398 Chr.6: 39345764-39345764 on Build GRCh38
rs953631476 Chr.6: 39345731-39345731 on Build GRCh38
rs750933157 Chr.6: 39345754-39345754 on Build GRCh38
rs1015484575 Chr.6: 39345648-39345648 on Build GRCh38
rs938556669 Chr.6: 39345526-39345526 on Build GRCh38
rs991427880 Chr.6: 39345560-39345560 on Build GRCh38
rs748434321 Chr.6: 39345774-39345774 on Build GRCh38
rs377710231 Chr.6: 39345799-39345802 on Build GRCh38
rs375569188 Chr.6: 39345720-39345720 on Build GRCh38
rs772157647 Chr.6: 39345608-39345608 on Build GRCh38
rs763857506 Chr.6: 39345841-39345841 on Build GRCh38
rs773129001 Chr.6: 39345667-39345667 on Build GRCh38
rs779180218 Chr.6: 39345772-39345772 on Build GRCh38
rs944880180 Chr.6: 39345775-39345775 on Build GRCh38
rs143977642 Chr.6: 39345516-39345516 on Build GRCh38
rs760645523 Chr.6: 39345695-39345695 on Build GRCh38
rs369828675 Chr.6: 39345685-39345686 on Build GRCh38
rs771823834 Chr.6: 39345683-39345685 on Build GRCh38
rs764878858 Chr.6: 39345721-39345721 on Build GRCh38
rs765853513 Chr.6: 39345712-39345712 on Build GRCh38
rs764714668 Chr.6: 39345827-39345827 on Build GRCh38
rs995263864 Chr.6: 39345498-39345498 on Build GRCh38
rs34799567 Chr.6: 39345842-39345843 on Build GRCh38
rs1024225681 Chr.6: 39345561-39345561 on Build GRCh38

View Full Product Details