Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153757481-153757979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS3
Entrez Gene ID
65123
Gene Name
integrator complex subunit 3
Gene Aliases
C1orf193, C1orf60, INT3, SOSS-A, SOSSA
UniGene
Hs.438723
Gene Chromosome Location
Chr.1: 153728050-153774808 on Build GRCh38
Transcripts
NM_001324475.1 NM_023015.4 XM_005245459.2 XM_005245461.2 XM_006711490.3 XM_011509906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3965841 c.464A>G p.N155S Chr.1: 153757696-153757696 on Build GRCh38
5353174 c.525G>A p.T175T Chr.1: 153757757-153757757 on Build GRCh38
4576288 c.1011G>A p.L337L Chr.1: 153757625-153757625 on Build GRCh38
3965840 c.1082A>G p.N361S Chr.1: 153757696-153757696 on Build GRCh38
3801948 c.441C>T p.I147I Chr.1: 153757673-153757673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762466150 Chr.1: 153757543-153757543 on Build GRCh38
rs145073466 Chr.1: 153757833-153757833 on Build GRCh38
rs771794875 Chr.1: 153757646-153757646 on Build GRCh38
rs751734929 Chr.1: 153757568-153757568 on Build GRCh38
rs949633704 Chr.1: 153757878-153757878 on Build GRCh38
rs908163798 Chr.1: 153757908-153757908 on Build GRCh38
rs898279684 Chr.1: 153757639-153757639 on Build GRCh38
rs73013763 Chr.1: 153757855-153757855 on Build GRCh38
rs746652077 Chr.1: 153757657-153757657 on Build GRCh38
rs777134962 Chr.1: 153757532-153757532 on Build GRCh38
rs766667220 Chr.1: 153757754-153757754 on Build GRCh38
rs762390639 Chr.1: 153757685-153757685 on Build GRCh38
rs758859747 Chr.1: 153757560-153757560 on Build GRCh38
rs544097109 Chr.1: 153757866-153757866 on Build GRCh38
rs374781507 Chr.1: 153757529-153757529 on Build GRCh38
rs372005326 Chr.1: 153757799-153757799 on Build GRCh38
rs187788086 Chr.1: 153757783-153757783 on Build GRCh38
rs778781548 Chr.1: 153757793-153757793 on Build GRCh38
rs765668270 Chr.1: 153757548-153757548 on Build GRCh38
rs902170764 Chr.1: 153757688-153757688 on Build GRCh38
rs201591643 Chr.1: 153757522-153757522 on Build GRCh38
rs1003925044 Chr.1: 153757557-153757557 on Build GRCh38
rs147588695 Chr.1: 153757885-153757885 on Build GRCh38
rs768197035 Chr.1: 153757679-153757679 on Build GRCh38
rs961344391 Chr.1: 153757929-153757929 on Build GRCh38
rs755029012 Chr.1: 153757586-153757586 on Build GRCh38
rs746312226 Chr.1: 153757716-153757716 on Build GRCh38
rs574326869 Chr.1: 153757936-153757936 on Build GRCh38
rs376736703 Chr.1: 153757625-153757625 on Build GRCh38
rs749221912 Chr.1: 153757615-153757615 on Build GRCh38
rs143916084 Chr.1: 153757748-153757748 on Build GRCh38
rs1046572467 Chr.1: 153757518-153757518 on Build GRCh38
rs192474414 Chr.1: 153757847-153757847 on Build GRCh38
rs966521119 Chr.1: 153757800-153757800 on Build GRCh38
rs916597950 Chr.1: 153757877-153757877 on Build GRCh38
rs201573287 Chr.1: 153757562-153757562 on Build GRCh38
rs775136705 Chr.1: 153757650-153757650 on Build GRCh38
rs995568333 Chr.1: 153757704-153757704 on Build GRCh38
rs763513343 Chr.1: 153757525-153757525 on Build GRCh38
rs573308422 Chr.1: 153757857-153757857 on Build GRCh38
rs757224349 Chr.1: 153757619-153757619 on Build GRCh38
rs369022403 Chr.1: 153757556-153757556 on Build GRCh38
rs369761936 Chr.1: 153757813-153757813 on Build GRCh38
rs745612147 Chr.1: 153757630-153757630 on Build GRCh38
rs745806032 Chr.1: 153757796-153757796 on Build GRCh38
rs751793716 Chr.1: 153757757-153757757 on Build GRCh38
rs373302294 Chr.1: 153757769-153757769 on Build GRCh38
rs776323925 Chr.1: 153757680-153757680 on Build GRCh38
rs760188202 Chr.1: 153757523-153757523 on Build GRCh38
rs756207152 Chr.1: 153757792-153757792 on Build GRCh38
rs773890778 Chr.1: 153757731-153757731 on Build GRCh38
rs949678616 Chr.1: 153757506-153757506 on Build GRCh38
rs767601630 Chr.1: 153757782-153757782 on Build GRCh38

View Full Product Details