Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52186276-52186793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17A
Entrez Gene ID
3605
Gene Name
interleukin 17A
Gene Aliases
CTLA-8, CTLA8, IL-17, IL-17A, IL17
UniGene
Hs.41724
Gene Chromosome Location
Chr.6: 52186387-52190638 on Build GRCh38
Transcripts
NM_002190.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1697403 c.7C>T p.P3S Chr.6: 52186438-52186438 on Build GRCh38
3628474 c.11G>A p.G4E Chr.6: 52186442-52186442 on Build GRCh38
4524942 c.12G>A p.G4G Chr.6: 52186443-52186443 on Build GRCh38
3413939 c.27+7C>T p.? Chr.6: 52186465-52186465 on Build GRCh38
4474188 c.18C>T p.T6T Chr.6: 52186449-52186449 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs532385992 Chr.6: 52186400-52186400 on Build GRCh38
rs369837714 Chr.6: 52186486-52186486 on Build GRCh38
rs781518025 Chr.6: 52186487-52186487 on Build GRCh38
rs751747356 Chr.6: 52186407-52186407 on Build GRCh38
rs992261560 Chr.6: 52186708-52186708 on Build GRCh38
rs376028065 Chr.6: 52186373-52186373 on Build GRCh38
rs541791747 Chr.6: 52186638-52186638 on Build GRCh38
rs901508394 Chr.6: 52186714-52186714 on Build GRCh38
rs894856419 Chr.6: 52186364-52186364 on Build GRCh38
rs780956602 Chr.6: 52186726-52186726 on Build GRCh38
rs769110906 Chr.6: 52186384-52186384 on Build GRCh38
rs375071669 Chr.6: 52186402-52186402 on Build GRCh38
rs879145627 Chr.6: 52186303-52186303 on Build GRCh38
rs927559114 Chr.6: 52186611-52186611 on Build GRCh38
rs762811673 Chr.6: 52186306-52186306 on Build GRCh38
rs757834044 Chr.6: 52186425-52186427 on Build GRCh38
rs917239174 Chr.6: 52186533-52186533 on Build GRCh38
rs771480019 Chr.6: 52186411-52186411 on Build GRCh38
rs939041484 Chr.6: 52186703-52186703 on Build GRCh38
rs556315679 Chr.6: 52186538-52186538 on Build GRCh38
rs546760729 Chr.6: 52186320-52186320 on Build GRCh38
rs961739686 Chr.6: 52186519-52186519 on Build GRCh38
rs778494591 Chr.6: 52186501-52186501 on Build GRCh38
rs760224158 Chr.6: 52186418-52186418 on Build GRCh38
rs73439726 Chr.6: 52186674-52186674 on Build GRCh38
rs199815459 Chr.6: 52186440-52186440 on Build GRCh38
rs529868643 Chr.6: 52186315-52186315 on Build GRCh38
rs554295086 Chr.6: 52186514-52186514 on Build GRCh38
rs759094625 Chr.6: 52186408-52186408 on Build GRCh38
rs754412506 Chr.6: 52186547-52186547 on Build GRCh38
rs3819025 Chr.6: 52186476-52186476 on Build GRCh38
rs891988041 Chr.6: 52186649-52186649 on Build GRCh38
rs751844725 Chr.6: 52186479-52186479 on Build GRCh38
rs17879568 Chr.6: 52186359-52186359 on Build GRCh38
rs950289198 Chr.6: 52186316-52186316 on Build GRCh38
rs781660057 Chr.6: 52186392-52186392 on Build GRCh38
rs557854899 Chr.6: 52186305-52186305 on Build GRCh38
rs8193037 Chr.6: 52186311-52186311 on Build GRCh38
rs761536762 Chr.6: 52186435-52186435 on Build GRCh38
rs568834044 Chr.6: 52186431-52186431 on Build GRCh38
rs983037484 Chr.6: 52186618-52186618 on Build GRCh38
rs577513673 Chr.6: 52186520-52186520 on Build GRCh38
rs763918114 Chr.6: 52186430-52186430 on Build GRCh38
rs775271736 Chr.6: 52186417-52186417 on Build GRCh38
rs749745973 Chr.6: 52186503-52186503 on Build GRCh38
rs773802580 Chr.6: 52186404-52186404 on Build GRCh38
rs758559245 Chr.6: 52186475-52186475 on Build GRCh38
rs891552708 Chr.6: 52186387-52186387 on Build GRCh38
rs891146499 Chr.6: 52186459-52186459 on Build GRCh38
rs8193038 Chr.6: 52186584-52186584 on Build GRCh38
rs972870076 Chr.6: 52186526-52186526 on Build GRCh38
rs752836946 Chr.6: 52186453-52186453 on Build GRCh38
rs140425841 Chr.6: 52186500-52186500 on Build GRCh38
rs916229019 Chr.6: 52186768-52186768 on Build GRCh38
rs751572347 Chr.6: 52186617-52186617 on Build GRCh38
rs181786431 Chr.6: 52186554-52186554 on Build GRCh38
rs765164206 Chr.6: 52186452-52186452 on Build GRCh38
rs746287561 Chr.6: 52186594-52186594 on Build GRCh38
rs950092755 Chr.6: 52186536-52186536 on Build GRCh38
rs940006147 Chr.6: 52186310-52186310 on Build GRCh38
rs947815261 Chr.6: 52186375-52186375 on Build GRCh38
rs201292455 Chr.6: 52186365-52186365 on Build GRCh38
rs748539355 Chr.6: 52186490-52186490 on Build GRCh38
rs938974430 Chr.6: 52186358-52186358 on Build GRCh38
rs190164861 Chr.6: 52186480-52186480 on Build GRCh38
rs554117879 Chr.6: 52186532-52186532 on Build GRCh38
rs757792991 Chr.6: 52186555-52186555 on Build GRCh38
rs774834222 Chr.6: 52186505-52186505 on Build GRCh38
rs771662465 Chr.6: 52186504-52186504 on Build GRCh38

View Full Product Details