Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73241233-73241743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDC1
Entrez Gene ID
80759
Gene Name
KH homology domain containing 1
Gene Aliases
C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4
UniGene
Hs.433062
Gene Chromosome Location
Chr.6: 73241314-73310365 on Build GRCh38
Transcripts
NM_001251874.1 NM_030568.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3762182 c.340G>A p.D114N Chr.6: 73241684-73241684 on Build GRCh38
5405813 c.401G>A p.R134K Chr.6: 73241623-73241623 on Build GRCh38
1568272 c.395C>T p.A132V Chr.6: 73241629-73241629 on Build GRCh38
743175 c.331C>G p.L111V Chr.6: 73241693-73241693 on Build GRCh38
4856205 c.490C>T p.P164S Chr.6: 73241534-73241534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765427624 Chr.6: 73241514-73241514 on Build GRCh38
rs761292485 Chr.6: 73241631-73241631 on Build GRCh38
rs1045486858 Chr.6: 73241363-73241363 on Build GRCh38
rs199979331 Chr.6: 73241684-73241684 on Build GRCh38
rs775405936 Chr.6: 73241677-73241677 on Build GRCh38
rs200762693 Chr.6: 73241660-73241660 on Build GRCh38
rs985482455 Chr.6: 73241653-73241653 on Build GRCh38
rs1034454909 Chr.6: 73241422-73241422 on Build GRCh38
rs750865295 Chr.6: 73241293-73241293 on Build GRCh38
rs751917883 Chr.6: 73241721-73241721 on Build GRCh38
rs772596344 Chr.6: 73241483-73241483 on Build GRCh38
rs905343170 Chr.6: 73241624-73241624 on Build GRCh38
rs781617538 Chr.6: 73241665-73241665 on Build GRCh38
rs768707302 Chr.6: 73241705-73241705 on Build GRCh38
rs756908211 Chr.6: 73241550-73241550 on Build GRCh38
rs764925786 Chr.6: 73241641-73241641 on Build GRCh38
rs373674170 Chr.6: 73241597-73241597 on Build GRCh38
rs762613619 Chr.6: 73241717-73241717 on Build GRCh38
rs747755706 Chr.6: 73241400-73241400 on Build GRCh38
rs781270536 Chr.6: 73241577-73241577 on Build GRCh38
rs761326459 Chr.6: 73241710-73241710 on Build GRCh38
rs1031553260 Chr.6: 73241326-73241326 on Build GRCh38
rs1013703800 Chr.6: 73241315-73241315 on Build GRCh38
rs753354797 Chr.6: 73241622-73241622 on Build GRCh38
rs1013063682 Chr.6: 73241267-73241267 on Build GRCh38
rs371388591 Chr.6: 73241571-73241571 on Build GRCh38
rs1014849992 Chr.6: 73241458-73241458 on Build GRCh38
rs530066779 Chr.6: 73241434-73241434 on Build GRCh38
rs955948185 Chr.6: 73241347-73241347 on Build GRCh38
rs776040074 Chr.6: 73241489-73241489 on Build GRCh38
rs904122596 Chr.6: 73241270-73241270 on Build GRCh38
rs779715428 Chr.6: 73241423-73241423 on Build GRCh38
rs367849614 Chr.6: 73241609-73241609 on Build GRCh38
rs769778458 Chr.6: 73241668-73241668 on Build GRCh38
rs755448501 Chr.6: 73241664-73241664 on Build GRCh38
rs1056613087 Chr.6: 73241686-73241686 on Build GRCh38
rs182325320 Chr.6: 73241532-73241532 on Build GRCh38
rs532829513 Chr.6: 73241397-73241397 on Build GRCh38
rs566208834 Chr.6: 73241441-73241441 on Build GRCh38
rs764657829 Chr.6: 73241711-73241711 on Build GRCh38
rs190629651 Chr.6: 73241610-73241610 on Build GRCh38
rs780494238 Chr.6: 73241351-73241351 on Build GRCh38
rs976698998 Chr.6: 73241370-73241370 on Build GRCh38
rs1052621380 Chr.6: 73241305-73241305 on Build GRCh38
rs753386664 Chr.6: 73241542-73241542 on Build GRCh38
rs186941657 Chr.6: 73241557-73241557 on Build GRCh38
rs1891494 Chr.6: 73241302-73241302 on Build GRCh38
rs562732290 Chr.6: 73241433-73241433 on Build GRCh38
rs895522888 Chr.6: 73241263-73241263 on Build GRCh38
rs563152950 Chr.6: 73241674-73241674 on Build GRCh38
rs1012597067 Chr.6: 73241510-73241510 on Build GRCh38
rs955895713 Chr.6: 73241566-73241566 on Build GRCh38
rs368269816 Chr.6: 73241675-73241675 on Build GRCh38
rs199726390 Chr.6: 73241559-73241559 on Build GRCh38
rs780480838 Chr.6: 73241563-73241563 on Build GRCh38
rs117702866 Chr.6: 73241401-73241401 on Build GRCh38
rs962762004 Chr.6: 73241662-73241662 on Build GRCh38
rs375224084 Chr.6: 73241716-73241716 on Build GRCh38
rs1050825945 Chr.6: 73241268-73241268 on Build GRCh38
rs892669092 Chr.6: 73241307-73241307 on Build GRCh38
rs941488701 Chr.6: 73241425-73241425 on Build GRCh38
rs538554497 Chr.6: 73241533-73241533 on Build GRCh38
rs767579637 Chr.6: 73241686-73241686 on Build GRCh38
rs376634472 Chr.6: 73241502-73241502 on Build GRCh38
rs748218067 Chr.6: 73241582-73241582 on Build GRCh38
rs201705217 Chr.6: 73241611-73241611 on Build GRCh38
rs377447679 Chr.6: 73241598-73241598 on Build GRCh38
rs73757029 Chr.6: 73241460-73241460 on Build GRCh38
rs373479803 Chr.6: 73241497-73241497 on Build GRCh38
rs143460400 Chr.6: 73241522-73241522 on Build GRCh38
rs766068547 Chr.6: 73241718-73241718 on Build GRCh38
rs778100429 Chr.6: 73241545-73241545 on Build GRCh38
rs758714404 Chr.6: 73241518-73241518 on Build GRCh38
rs777989124 Chr.6: 73241671-73241671 on Build GRCh38
rs377743187 Chr.6: 73241560-73241560 on Build GRCh38
rs939603983 Chr.6: 73241453-73241453 on Build GRCh38
rs750055770 Chr.6: 73241644-73241644 on Build GRCh38
rs932691641 Chr.6: 73241398-73241398 on Build GRCh38
rs887483060 Chr.6: 73241685-73241685 on Build GRCh38
rs201194392 Chr.6: 73241666-73241666 on Build GRCh38
rs764172574 Chr.6: 73241618-73241618 on Build GRCh38
rs780425911 Chr.6: 73241648-73241648 on Build GRCh38
rs528287481 Chr.6: 73241511-73241511 on Build GRCh38
rs895459650 Chr.6: 73241470-73241470 on Build GRCh38
rs767550112 Chr.6: 73241523-73241523 on Build GRCh38
rs148053198 Chr.6: 73241612-73241612 on Build GRCh38
rs750381929 Chr.6: 73241389-73241389 on Build GRCh38

View Full Product Details