Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73241639-73242147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDC1
Entrez Gene ID
80759
Gene Name
KH homology domain containing 1
Gene Aliases
C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4
UniGene
Hs.433062
Gene Chromosome Location
Chr.6: 73241314-73310365 on Build GRCh38
Transcripts
NM_001251874.1 NM_030568.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
312240 c.293G>T p.R98L Chr.6: 73242057-73242057 on Build GRCh38
3162207 c.203G>A p.R68H Chr.6: 73242147-73242147 on Build GRCh38
4797808 c.273G>A p.Q91Q Chr.6: 73242077-73242077 on Build GRCh38
1445818 c.243G>T p.L81L Chr.6: 73242107-73242107 on Build GRCh38
5061934 c.264_265insTG p.G89fs*24 Chr.6: 73242085-73242086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199979331 Chr.6: 73241684-73241684 on Build GRCh38
rs775405936 Chr.6: 73241677-73241677 on Build GRCh38
rs751917883 Chr.6: 73241721-73241721 on Build GRCh38
rs1007255105 Chr.6: 73241926-73241926 on Build GRCh38
rs781617538 Chr.6: 73241665-73241665 on Build GRCh38
rs768707302 Chr.6: 73241705-73241705 on Build GRCh38
rs777742356 Chr.6: 73241767-73241767 on Build GRCh38
rs762613619 Chr.6: 73241717-73241717 on Build GRCh38
rs564747296 Chr.6: 73241966-73241966 on Build GRCh38
rs761326459 Chr.6: 73241710-73241710 on Build GRCh38
rs750643533 Chr.6: 73242010-73242012 on Build GRCh38
rs997375359 Chr.6: 73241783-73241783 on Build GRCh38
rs988067195 Chr.6: 73242000-73242000 on Build GRCh38
rs181290286 Chr.6: 73242085-73242085 on Build GRCh38
rs546437325 Chr.6: 73241956-73241956 on Build GRCh38
rs772325727 Chr.6: 73241781-73241781 on Build GRCh38
rs527758652 Chr.6: 73241906-73241906 on Build GRCh38
rs763257301 Chr.6: 73241932-73241932 on Build GRCh38
rs572106838 Chr.6: 73242100-73242100 on Build GRCh38
rs140892193 Chr.6: 73241832-73241832 on Build GRCh38
rs748066811 Chr.6: 73242054-73242054 on Build GRCh38
rs368583826 Chr.6: 73241736-73241736 on Build GRCh38
rs555526620 Chr.6: 73241768-73241768 on Build GRCh38
rs769778458 Chr.6: 73241668-73241668 on Build GRCh38
rs541787811 Chr.6: 73241831-73241831 on Build GRCh38
rs755448501 Chr.6: 73241664-73241664 on Build GRCh38
rs1056613087 Chr.6: 73241686-73241686 on Build GRCh38
rs774006126 Chr.6: 73242080-73242080 on Build GRCh38
rs764657829 Chr.6: 73241711-73241711 on Build GRCh38
rs576812355 Chr.6: 73242052-73242052 on Build GRCh38
rs775734336 Chr.6: 73241801-73241801 on Build GRCh38
rs757519552 Chr.6: 73241777-73241777 on Build GRCh38
rs925367878 Chr.6: 73241969-73241969 on Build GRCh38
rs1029968261 Chr.6: 73241830-73241830 on Build GRCh38
rs755395535 Chr.6: 73241730-73241730 on Build GRCh38
rs759829191 Chr.6: 73242083-73242083 on Build GRCh38
rs756214465 Chr.6: 73242099-73242100 on Build GRCh38
rs563152950 Chr.6: 73241674-73241674 on Build GRCh38
rs756183275 Chr.6: 73241755-73241755 on Build GRCh38
rs746783459 Chr.6: 73242018-73242018 on Build GRCh38
rs368269816 Chr.6: 73241675-73241675 on Build GRCh38
rs199949364 Chr.6: 73242057-73242057 on Build GRCh38
rs768344998 Chr.6: 73242035-73242035 on Build GRCh38
rs891619950 Chr.6: 73241774-73241774 on Build GRCh38
rs749431536 Chr.6: 73241775-73241775 on Build GRCh38
rs962762004 Chr.6: 73241662-73241662 on Build GRCh38
rs375224084 Chr.6: 73241716-73241716 on Build GRCh38
rs569098928 Chr.6: 73242106-73242106 on Build GRCh38
rs766005212 Chr.6: 73242069-73242069 on Build GRCh38
rs953996797 Chr.6: 73241840-73241840 on Build GRCh38
rs767579637 Chr.6: 73241686-73241686 on Build GRCh38
rs918684169 Chr.6: 73241968-73241968 on Build GRCh38
rs1052877698 Chr.6: 73241740-73241740 on Build GRCh38
rs766068547 Chr.6: 73241718-73241718 on Build GRCh38
rs998030231 Chr.6: 73241795-73241795 on Build GRCh38
rs532287244 Chr.6: 73241995-73241995 on Build GRCh38
rs753149803 Chr.6: 73241747-73241747 on Build GRCh38
rs777989124 Chr.6: 73241671-73241671 on Build GRCh38
rs780250371 Chr.6: 73242121-73242121 on Build GRCh38
rs961929845 Chr.6: 73241749-73241749 on Build GRCh38
rs1030601678 Chr.6: 73241826-73241826 on Build GRCh38
rs369158621 Chr.6: 73242111-73242111 on Build GRCh38
rs887483060 Chr.6: 73241685-73241685 on Build GRCh38
rs768333060 Chr.6: 73242058-73242058 on Build GRCh38
rs201194392 Chr.6: 73241666-73241666 on Build GRCh38
rs762518328 Chr.6: 73242061-73242061 on Build GRCh38
rs753203314 Chr.6: 73242093-73242093 on Build GRCh38
rs916783826 Chr.6: 73242051-73242051 on Build GRCh38

View Full Product Details