Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125275266-125275769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDDC2
Entrez Gene ID
51020
Gene Name
HD domain containing 2
Gene Aliases
C6orf74, CGI-130, NS5ATP2, dJ167O5.2
UniGene
Hs.32826
Gene Chromosome Location
Chr.6: 125274934-125302141 on Build GRCh38
Transcripts
NM_016063.2

SNP Detail

SNP ID Chromosome Location
rs928278995 Chr.6: 125275683-125275683 on Build GRCh38
rs906723675 Chr.6: 125275365-125275365 on Build GRCh38
rs967418357 Chr.6: 125275409-125275409 on Build GRCh38
rs945591253 Chr.6: 125275640-125275640 on Build GRCh38
rs965970955 Chr.6: 125275486-125275486 on Build GRCh38
rs542324028 Chr.6: 125275592-125275592 on Build GRCh38
rs760899398 Chr.6: 125275357-125275357 on Build GRCh38
rs928216077 Chr.6: 125275723-125275723 on Build GRCh38
rs939529931 Chr.6: 125275370-125275370 on Build GRCh38
rs940357789 Chr.6: 125275319-125275319 on Build GRCh38
rs978342112 Chr.6: 125275690-125275690 on Build GRCh38
rs953230138 Chr.6: 125275563-125275563 on Build GRCh38
rs954539679 Chr.6: 125275501-125275501 on Build GRCh38
rs77956410 Chr.6: 125275332-125275332 on Build GRCh38
rs556585150 Chr.6: 125275523-125275523 on Build GRCh38
rs921788450 Chr.6: 125275553-125275553 on Build GRCh38
rs34648273 Chr.6: 125275406-125275407 on Build GRCh38
rs534478918 Chr.6: 125275397-125275397 on Build GRCh38
rs35994131 Chr.6: 125275320-125275320 on Build GRCh38
rs536749212 Chr.6: 125275315-125275315 on Build GRCh38
rs1017315875 Chr.6: 125275580-125275580 on Build GRCh38
rs765432518 Chr.6: 125275546-125275546 on Build GRCh38
rs1026669670 Chr.6: 125275477-125275477 on Build GRCh38
rs976007827 Chr.6: 125275509-125275509 on Build GRCh38
rs989148624 Chr.6: 125275309-125275309 on Build GRCh38
rs574946351 Chr.6: 125275574-125275574 on Build GRCh38
rs938958869 Chr.6: 125275730-125275730 on Build GRCh38
rs183475296 Chr.6: 125275463-125275463 on Build GRCh38
rs567088680 Chr.6: 125275377-125275377 on Build GRCh38
rs752875969 Chr.6: 125275598-125275598 on Build GRCh38
rs1003048128 Chr.6: 125275368-125275368 on Build GRCh38
rs998865396 Chr.6: 125275418-125275418 on Build GRCh38
rs940408470 Chr.6: 125275593-125275593 on Build GRCh38
rs528107908 Chr.6: 125275577-125275577 on Build GRCh38
rs894632969 Chr.6: 125275382-125275382 on Build GRCh38
rs552930280 Chr.6: 125275391-125275391 on Build GRCh38
rs192104802 Chr.6: 125275424-125275424 on Build GRCh38
rs1016924742 Chr.6: 125275404-125275404 on Build GRCh38
rs776642190 Chr.6: 125275331-125275331 on Build GRCh38
rs907471272 Chr.6: 125275364-125275364 on Build GRCh38
rs1005508850 Chr.6: 125275398-125275398 on Build GRCh38
rs564754466 Chr.6: 125275449-125275449 on Build GRCh38
rs9491352 Chr.6: 125275604-125275604 on Build GRCh38
rs866489799 Chr.6: 125275583-125275583 on Build GRCh38
rs942982502 Chr.6: 125275308-125275308 on Build GRCh38

View Full Product Details