Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154700023-154700538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNN3
Entrez Gene ID
3782
Gene Name
potassium calcium-activated channel subfamily N member 3
Gene Aliases
KCa2.3, SK3, SKCA3, hSK3
UniGene
Hs.490765
Gene Chromosome Location
Chr.1: 154697462-154870278 on Build GRCh38
Transcripts
NM_001204087.1 NM_002249.5 NM_170782.2

SNP Detail

SNP ID Chromosome Location
rs552358152 Chr.1: 154700243-154700243 on Build GRCh38
rs568429091 Chr.1: 154700443-154700443 on Build GRCh38
rs916898938 Chr.1: 154700424-154700424 on Build GRCh38
rs187307782 Chr.1: 154700507-154700507 on Build GRCh38
rs970644741 Chr.1: 154700229-154700229 on Build GRCh38
rs570554440 Chr.1: 154700265-154700265 on Build GRCh38
rs912118966 Chr.1: 154700389-154700389 on Build GRCh38
rs978656275 Chr.1: 154700306-154700306 on Build GRCh38
rs761086990 Chr.1: 154700170-154700170 on Build GRCh38
rs920713471 Chr.1: 154700305-154700305 on Build GRCh38
rs1039217816 Chr.1: 154700145-154700145 on Build GRCh38
rs981916358 Chr.1: 154700235-154700235 on Build GRCh38
rs954611772 Chr.1: 154700208-154700208 on Build GRCh38
rs144000297 Chr.1: 154700494-154700494 on Build GRCh38
rs1009206844 Chr.1: 154700489-154700489 on Build GRCh38
rs996591479 Chr.1: 154700163-154700163 on Build GRCh38
rs145423467 Chr.1: 154700108-154700109 on Build GRCh38
rs893464727 Chr.1: 154700205-154700205 on Build GRCh38
rs766744517 Chr.1: 154700174-154700174 on Build GRCh38
rs755495834 Chr.1: 154700216-154700216 on Build GRCh38
rs925684800 Chr.1: 154700316-154700316 on Build GRCh38
rs944844054 Chr.1: 154700130-154700130 on Build GRCh38
rs767677674 Chr.1: 154700103-154700103 on Build GRCh38
rs1039717449 Chr.1: 154700427-154700427 on Build GRCh38
rs548990336 Chr.1: 154700203-154700203 on Build GRCh38
rs1047966116 Chr.1: 154700071-154700071 on Build GRCh38
rs190820388 Chr.1: 154700120-154700120 on Build GRCh38
rs752188861 Chr.1: 154700250-154700250 on Build GRCh38
rs535488048 Chr.1: 154700493-154700493 on Build GRCh38
rs180684183 Chr.1: 154700326-154700326 on Build GRCh38
rs964201897 Chr.1: 154700159-154700159 on Build GRCh38
rs929340789 Chr.1: 154700054-154700054 on Build GRCh38
rs936200715 Chr.1: 154700468-154700468 on Build GRCh38
rs1020849556 Chr.1: 154700218-154700218 on Build GRCh38
rs1042092967 Chr.1: 154700492-154700492 on Build GRCh38
rs12039698 Chr.1: 154700134-154700134 on Build GRCh38
rs762076815 Chr.1: 154700083-154700083 on Build GRCh38
rs750795225 Chr.1: 154700146-154700146 on Build GRCh38
rs1053538312 Chr.1: 154700480-154700480 on Build GRCh38
rs937133541 Chr.1: 154700371-154700371 on Build GRCh38
rs374503210 Chr.1: 154700276-154700276 on Build GRCh38
rs1047310034 Chr.1: 154700438-154700438 on Build GRCh38
rs527472994 Chr.1: 154700164-154700164 on Build GRCh38
rs973503761 Chr.1: 154700283-154700283 on Build GRCh38
rs369468720 Chr.1: 154700446-154700446 on Build GRCh38
rs777344569 Chr.1: 154700434-154700434 on Build GRCh38
rs893566633 Chr.1: 154700484-154700484 on Build GRCh38
rs944979792 Chr.1: 154700422-154700422 on Build GRCh38
rs758065798 Chr.1: 154700338-154700338 on Build GRCh38

View Full Product Details