Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154701738-154702248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNN3
Entrez Gene ID
3782
Gene Name
potassium calcium-activated channel subfamily N member 3
Gene Aliases
KCa2.3, SK3, SKCA3, hSK3
UniGene
Hs.490765
Gene Chromosome Location
Chr.1: 154697462-154870278 on Build GRCh38
Transcripts
NM_001204087.1 NM_002249.5 NM_170782.2

SNP Detail

SNP ID Chromosome Location
rs931871756 Chr.1: 154701829-154701829 on Build GRCh38
rs940593352 Chr.1: 154701807-154701807 on Build GRCh38
rs145970966 Chr.1: 154702036-154702036 on Build GRCh38
rs936780560 Chr.1: 154702108-154702108 on Build GRCh38
rs747207397 Chr.1: 154701895-154701895 on Build GRCh38
rs1015314926 Chr.1: 154701866-154701866 on Build GRCh38
rs1040296282 Chr.1: 154701812-154701812 on Build GRCh38
rs894379737 Chr.1: 154701781-154701781 on Build GRCh38
rs976401373 Chr.1: 154702181-154702181 on Build GRCh38
rs139897775 Chr.1: 154702210-154702210 on Build GRCh38
rs1023772105 Chr.1: 154702051-154702051 on Build GRCh38
rs6691359 Chr.1: 154701794-154701794 on Build GRCh38
rs893226111 Chr.1: 154701919-154701919 on Build GRCh38
rs931772421 Chr.1: 154702197-154702197 on Build GRCh38
rs549752326 Chr.1: 154701821-154701821 on Build GRCh38
rs1003116285 Chr.1: 154702011-154702011 on Build GRCh38
rs551231347 Chr.1: 154702071-154702071 on Build GRCh38
rs76956551 Chr.1: 154702091-154702091 on Build GRCh38
rs775789705 Chr.1: 154702198-154702198 on Build GRCh38
rs6699979 Chr.1: 154702018-154702018 on Build GRCh38
rs1028427477 Chr.1: 154701902-154701902 on Build GRCh38
rs1041760029 Chr.1: 154701941-154701941 on Build GRCh38
rs182686457 Chr.1: 154701809-154701809 on Build GRCh38
rs907534850 Chr.1: 154702128-154702128 on Build GRCh38
rs1011553010 Chr.1: 154701935-154701935 on Build GRCh38
rs995168175 Chr.1: 154701887-154701887 on Build GRCh38
rs561929443 Chr.1: 154701877-154701877 on Build GRCh38
rs1045697057 Chr.1: 154701799-154701799 on Build GRCh38
rs370030603 Chr.1: 154702032-154702032 on Build GRCh38
rs879652491 Chr.1: 154701873-154701873 on Build GRCh38
rs912822473 Chr.1: 154702012-154702012 on Build GRCh38
rs984527391 Chr.1: 154702102-154702102 on Build GRCh38
rs186847390 Chr.1: 154701966-154701966 on Build GRCh38
rs370648477 Chr.1: 154701768-154701768 on Build GRCh38
rs773313614 Chr.1: 154701828-154701828 on Build GRCh38
rs528995053 Chr.1: 154702187-154702187 on Build GRCh38
rs923089550 Chr.1: 154701827-154701827 on Build GRCh38
rs959125091 Chr.1: 154702029-154702029 on Build GRCh38
rs961721271 Chr.1: 154702130-154702130 on Build GRCh38
rs916546465 Chr.1: 154702150-154702150 on Build GRCh38
rs759955949 Chr.1: 154701776-154701776 on Build GRCh38
rs775656858 Chr.1: 154701876-154701876 on Build GRCh38

View Full Product Details